Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78046233-78046737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC2
Entrez Gene ID
54810
Gene Name
GIPC PDZ domain containing family member 2
Gene Aliases
SEMCAP-2, SEMCAP2
UniGene
Hs.603818
Gene Chromosome Location
Chr.1: 78044962-78138449 on Build GRCh38
Transcripts
NM_001304725.1 NM_017655.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320580 c.170T>A p.V57E Chr.1: 78046264-78046264 on Build GRCh38
292010 c.221T>A p.F74Y Chr.1: 78046315-78046315 on Build GRCh38
1743418 c.230C>T p.S77L Chr.1: 78046324-78046324 on Build GRCh38
5685110 c.167G>A p.R56Q Chr.1: 78046261-78046261 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762511595 Chr.1: 78046317-78046317 on Build GRCh38
rs775053230 Chr.1: 78046309-78046309 on Build GRCh38
rs778802271 Chr.1: 78046349-78046349 on Build GRCh38
rs370127088 Chr.1: 78046259-78046259 on Build GRCh38
rs538252682 Chr.1: 78046347-78046347 on Build GRCh38
rs781655687 Chr.1: 78046384-78046393 on Build GRCh38
rs759433899 Chr.1: 78046306-78046306 on Build GRCh38
rs750714304 Chr.1: 78046369-78046370 on Build GRCh38
rs955477770 Chr.1: 78046567-78046567 on Build GRCh38
rs542285536 Chr.1: 78046370-78046379 on Build GRCh38
rs376997722 Chr.1: 78046258-78046258 on Build GRCh38
rs748261687 Chr.1: 78046355-78046355 on Build GRCh38
rs780313090 Chr.1: 78046345-78046345 on Build GRCh38
rs781317492 Chr.1: 78046360-78046360 on Build GRCh38
rs909022117 Chr.1: 78046319-78046319 on Build GRCh38
rs756290955 Chr.1: 78046262-78046262 on Build GRCh38
rs756523655 Chr.1: 78046339-78046339 on Build GRCh38
rs766589686 Chr.1: 78046383-78046383 on Build GRCh38
rs536041363 Chr.1: 78046419-78046419 on Build GRCh38
rs773767419 Chr.1: 78046378-78046378 on Build GRCh38
rs894927346 Chr.1: 78046707-78046707 on Build GRCh38
rs750252814 Chr.1: 78046451-78046451 on Build GRCh38
rs962920698 Chr.1: 78046295-78046295 on Build GRCh38
rs977462385 Chr.1: 78046616-78046616 on Build GRCh38
rs761864707 Chr.1: 78046362-78046379 on Build GRCh38
rs532208131 Chr.1: 78046445-78046445 on Build GRCh38
rs754245993 Chr.1: 78046385-78046385 on Build GRCh38
rs1045935786 Chr.1: 78046468-78046468 on Build GRCh38
rs768366870 Chr.1: 78046369-78046369 on Build GRCh38
rs746877631 Chr.1: 78046288-78046288 on Build GRCh38
rs950164632 Chr.1: 78046439-78046439 on Build GRCh38
rs1007975649 Chr.1: 78046583-78046583 on Build GRCh38
rs546278344 Chr.1: 78046581-78046581 on Build GRCh38
rs898140661 Chr.1: 78046533-78046533 on Build GRCh38
rs770949826 Chr.1: 78046357-78046357 on Build GRCh38
rs761354882 Chr.1: 78046373-78046373 on Build GRCh38
rs946136644 Chr.1: 78046354-78046354 on Build GRCh38
rs926069755 Chr.1: 78046623-78046623 on Build GRCh38
rs565046927 Chr.1: 78046327-78046327 on Build GRCh38
rs113307583 Chr.1: 78046632-78046632 on Build GRCh38
rs539199310 Chr.1: 78046693-78046693 on Build GRCh38
rs191111075 Chr.1: 78046545-78046545 on Build GRCh38
rs755909724 Chr.1: 78046503-78046503 on Build GRCh38
rs769456024 Chr.1: 78046307-78046307 on Build GRCh38
rs368018968 Chr.1: 78046374-78046374 on Build GRCh38
rs776563650 Chr.1: 78046360-78046377 on Build GRCh38
rs996953616 Chr.1: 78046546-78046546 on Build GRCh38
rs921390436 Chr.1: 78046488-78046488 on Build GRCh38
rs1024584927 Chr.1: 78046429-78046429 on Build GRCh38
rs974853947 Chr.1: 78046375-78046375 on Build GRCh38
rs758726529 Chr.1: 78046370-78046374 on Build GRCh38
rs959113333 Chr.1: 78046409-78046409 on Build GRCh38
rs1009021598 Chr.1: 78046658-78046659 on Build GRCh38
rs147346849 Chr.1: 78046296-78046296 on Build GRCh38
rs374832712 Chr.1: 78046331-78046331 on Build GRCh38
rs916127235 Chr.1: 78046676-78046676 on Build GRCh38
rs749369224 Chr.1: 78046268-78046268 on Build GRCh38
rs1021145130 Chr.1: 78046381-78046381 on Build GRCh38
rs977368756 Chr.1: 78046380-78046380 on Build GRCh38
rs991750984 Chr.1: 78046269-78046269 on Build GRCh38
rs187800751 Chr.1: 78046292-78046292 on Build GRCh38
rs780182588 Chr.1: 78046263-78046263 on Build GRCh38
rs1028232100 Chr.1: 78046551-78046551 on Build GRCh38
rs568793187 Chr.1: 78046400-78046400 on Build GRCh38
rs764792988 Chr.1: 78046594-78046594 on Build GRCh38
rs894754828 Chr.1: 78046429-78046429 on Build GRCh38
rs17101180 Chr.1: 78046276-78046276 on Build GRCh38
rs751081699 Chr.1: 78046325-78046325 on Build GRCh38
rs868514544 Chr.1: 78046683-78046683 on Build GRCh38
rs759917393 Chr.1: 78046379-78046380 on Build GRCh38
rs372492515 Chr.1: 78046382-78046382 on Build GRCh38
rs886837001 Chr.1: 78046480-78046480 on Build GRCh38
rs941867890 Chr.1: 78046462-78046462 on Build GRCh38
rs183708110 Chr.1: 78046593-78046593 on Build GRCh38
rs763950038 Chr.1: 78046351-78046352 on Build GRCh38
rs560737952 Chr.1: 78046375-78046379 on Build GRCh38
rs909103498 Chr.1: 78046440-78046440 on Build GRCh38
rs1020761147 Chr.1: 78046591-78046591 on Build GRCh38
rs908699159 Chr.1: 78046711-78046711 on Build GRCh38
rs370883891 Chr.1: 78046381-78046390 on Build GRCh38
rs983352413 Chr.1: 78046433-78046433 on Build GRCh38
rs200026593 Chr.1: 78046370-78046370 on Build GRCh38
rs904068911 Chr.1: 78046633-78046633 on Build GRCh38
rs1038156998 Chr.1: 78046511-78046511 on Build GRCh38
rs562691981 Chr.1: 78046250-78046250 on Build GRCh38
rs755305218 Chr.1: 78046372-78046373 on Build GRCh38
rs1033854111 Chr.1: 78046403-78046403 on Build GRCh38
rs781465732 Chr.1: 78046257-78046257 on Build GRCh38
rs991908306 Chr.1: 78046668-78046668 on Build GRCh38
rs762445383 Chr.1: 78046364-78046364 on Build GRCh38
rs750757762 Chr.1: 78046334-78046334 on Build GRCh38
rs769534480 Chr.1: 78046362-78046362 on Build GRCh38
rs763574753 Chr.1: 78046323-78046323 on Build GRCh38

View Full Product Details