Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78136522-78137023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC2
Entrez Gene ID
54810
Gene Name
GIPC PDZ domain containing family member 2
Gene Aliases
SEMCAP-2, SEMCAP2
UniGene
Hs.603818
Gene Chromosome Location
Chr.1: 78044962-78138449 on Build GRCh38
Transcripts
NM_001304725.1 NM_017655.5

SNP Detail

SNP ID Chromosome Location
rs74093227 Chr.1: 78136970-78136970 on Build GRCh38
rs556542522 Chr.1: 78136952-78136952 on Build GRCh38
rs13374377 Chr.1: 78136670-78136670 on Build GRCh38
rs1030774947 Chr.1: 78136767-78136767 on Build GRCh38
rs1005804798 Chr.1: 78136564-78136564 on Build GRCh38
rs1010540826 Chr.1: 78136871-78136871 on Build GRCh38
rs627661 Chr.1: 78136807-78136807 on Build GRCh38
rs1052094745 Chr.1: 78136855-78136855 on Build GRCh38
rs187493094 Chr.1: 78136849-78136849 on Build GRCh38
rs866600280 Chr.1: 78136879-78136879 on Build GRCh38
rs368496884 Chr.1: 78136608-78136609 on Build GRCh38
rs992865092 Chr.1: 78136687-78136687 on Build GRCh38
rs568805941 Chr.1: 78136600-78136601 on Build GRCh38
rs557029500 Chr.1: 78136827-78136827 on Build GRCh38
rs628002 Chr.1: 78136777-78136777 on Build GRCh38
rs775447963 Chr.1: 78136837-78136838 on Build GRCh38
rs933738979 Chr.1: 78136834-78136834 on Build GRCh38
rs776035641 Chr.1: 78136613-78136614 on Build GRCh38
rs900621298 Chr.1: 78136669-78136669 on Build GRCh38
rs1014352540 Chr.1: 78136963-78136963 on Build GRCh38
rs879132892 Chr.1: 78136696-78136696 on Build GRCh38
rs960152236 Chr.1: 78136953-78136953 on Build GRCh38
rs147035058 Chr.1: 78136872-78136872 on Build GRCh38
rs1039858531 Chr.1: 78136828-78136828 on Build GRCh38
rs969639161 Chr.1: 78136553-78136554 on Build GRCh38
rs892137438 Chr.1: 78136864-78136864 on Build GRCh38
rs80232408 Chr.1: 78136629-78136629 on Build GRCh38
rs78721412 Chr.1: 78136626-78136626 on Build GRCh38
rs201587350 Chr.1: 78136609-78136613 on Build GRCh38
rs764838891 Chr.1: 78136762-78136762 on Build GRCh38
rs1037782114 Chr.1: 78136581-78136581 on Build GRCh38
rs781181547 Chr.1: 78136962-78136962 on Build GRCh38
rs771065995 Chr.1: 78136961-78136961 on Build GRCh38
rs869130934 Chr.1: 78136624-78136628 on Build GRCh38
rs1034873113 Chr.1: 78136583-78136583 on Build GRCh38
rs747022225 Chr.1: 78136888-78136888 on Build GRCh38
rs1002477380 Chr.1: 78136893-78136893 on Build GRCh38
rs771608745 Chr.1: 78136610-78136613 on Build GRCh38
rs951322022 Chr.1: 78136736-78136736 on Build GRCh38
rs758047549 Chr.1: 78136765-78136765 on Build GRCh38
rs752207497 Chr.1: 78136764-78136765 on Build GRCh38
rs61013502 Chr.1: 78136609-78136612 on Build GRCh38
rs918591789 Chr.1: 78136734-78136734 on Build GRCh38
rs796081664 Chr.1: 78136626-78136629 on Build GRCh38
rs56230848 Chr.1: 78136627-78136628 on Build GRCh38
rs996171491 Chr.1: 78136681-78136681 on Build GRCh38
rs1034845571 Chr.1: 78136894-78136894 on Build GRCh38

View Full Product Details