Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84574161-84574666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
UniGene
Hs.513557
Gene Chromosome Location
Chr.1: 84553121-84574480 on Build GRCh38
Transcripts
NM_004388.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637183 c.33C>T p.L11L Chr.1: 84574383-84574383 on Build GRCh38
1290235 c.92_100delTGGCGCTGC p.L31_L33delLAL Chr.1: 84574316-84574324 on Build GRCh38
3747085 c.30C>T p.R10R Chr.1: 84574386-84574386 on Build GRCh38
7324006 c.150C>T p.R50R Chr.1: 84574266-84574266 on Build GRCh38
4144200 c.96G>C p.A32A Chr.1: 84574320-84574320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539036514 Chr.1: 84574440-84574440 on Build GRCh38
rs767568131 Chr.1: 84574435-84574435 on Build GRCh38
rs1018285387 Chr.1: 84574528-84574528 on Build GRCh38
rs201060055 Chr.1: 84574322-84574322 on Build GRCh38
rs1001911399 Chr.1: 84574394-84574394 on Build GRCh38
rs201734152 Chr.1: 84574350-84574350 on Build GRCh38
rs955524191 Chr.1: 84574519-84574519 on Build GRCh38
rs115347546 Chr.1: 84574387-84574387 on Build GRCh38
rs80026762 Chr.1: 84574210-84574210 on Build GRCh38
rs879527266 Chr.1: 84574429-84574429 on Build GRCh38
rs199701060 Chr.1: 84574324-84574324 on Build GRCh38
rs984822556 Chr.1: 84574457-84574457 on Build GRCh38
rs1020197174 Chr.1: 84574478-84574478 on Build GRCh38
rs377174797 Chr.1: 84574276-84574276 on Build GRCh38
rs934191660 Chr.1: 84574298-84574298 on Build GRCh38
rs754822107 Chr.1: 84574355-84574355 on Build GRCh38
rs756693826 Chr.1: 84574340-84574342 on Build GRCh38
rs566784316 Chr.1: 84574437-84574437 on Build GRCh38
rs985930909 Chr.1: 84574524-84574524 on Build GRCh38
rs745427922 Chr.1: 84574344-84574349 on Build GRCh38
rs1030069841 Chr.1: 84574514-84574514 on Build GRCh38
rs111314592 Chr.1: 84574459-84574459 on Build GRCh38
rs570063308 Chr.1: 84574339-84574339 on Build GRCh38
rs770632973 Chr.1: 84574456-84574456 on Build GRCh38
rs532421838 Chr.1: 84574305-84574305 on Build GRCh38
rs778760119 Chr.1: 84574356-84574356 on Build GRCh38
rs2911592 Chr.1: 84574386-84574386 on Build GRCh38
rs200761866 Chr.1: 84574415-84574415 on Build GRCh38
rs973170228 Chr.1: 84574546-84574546 on Build GRCh38
rs926507834 Chr.1: 84574578-84574578 on Build GRCh38
rs34451108 Chr.1: 84574471-84574471 on Build GRCh38
rs773921710 Chr.1: 84574267-84574267 on Build GRCh38
rs917731310 Chr.1: 84574560-84574560 on Build GRCh38
rs11487943 Chr.1: 84574504-84574504 on Build GRCh38
rs751747964 Chr.1: 84574419-84574419 on Build GRCh38
rs373271011 Chr.1: 84574361-84574361 on Build GRCh38
rs765525422 Chr.1: 84574391-84574391 on Build GRCh38
rs779614755 Chr.1: 84574352-84574353 on Build GRCh38
rs545745384 Chr.1: 84574314-84574314 on Build GRCh38
rs751607399 Chr.1: 84574336-84574336 on Build GRCh38
rs764093334 Chr.1: 84574321-84574321 on Build GRCh38
rs961759402 Chr.1: 84574469-84574469 on Build GRCh38
rs770545131 Chr.1: 84574385-84574385 on Build GRCh38
rs766955313 Chr.1: 84574290-84574290 on Build GRCh38
rs3217269 Chr.1: 84574341-84574349 on Build GRCh38
rs957273756 Chr.1: 84574209-84574209 on Build GRCh38
rs1020462048 Chr.1: 84574416-84574416 on Build GRCh38
rs772631953 Chr.1: 84574382-84574382 on Build GRCh38
rs569575125 Chr.1: 84574589-84574589 on Build GRCh38
rs1054563254 Chr.1: 84574408-84574408 on Build GRCh38
rs774904751 Chr.1: 84574248-84574248 on Build GRCh38
rs748399840 Chr.1: 84574333-84574333 on Build GRCh38
rs369111221 Chr.1: 84574211-84574211 on Build GRCh38
rs181008916 Chr.1: 84574207-84574207 on Build GRCh38
rs771545481 Chr.1: 84574275-84574275 on Build GRCh38
rs781634018 Chr.1: 84574333-84574350 on Build GRCh38
rs765597870 Chr.1: 84574194-84574194 on Build GRCh38
rs1033282820 Chr.1: 84574462-84574462 on Build GRCh38
rs897183596 Chr.1: 84574366-84574366 on Build GRCh38
rs908968581 Chr.1: 84574465-84574465 on Build GRCh38
rs1030087496 Chr.1: 84574592-84574592 on Build GRCh38
rs971908908 Chr.1: 84574498-84574498 on Build GRCh38
rs937367103 Chr.1: 84574590-84574590 on Build GRCh38
rs752929008 Chr.1: 84574310-84574310 on Build GRCh38
rs975743389 Chr.1: 84574559-84574559 on Build GRCh38
rs1054857018 Chr.1: 84574591-84574591 on Build GRCh38
rs749197755 Chr.1: 84574354-84574354 on Build GRCh38
rs142534762 Chr.1: 84574316-84574324 on Build GRCh38
rs1044973679 Chr.1: 84574575-84574575 on Build GRCh38
rs964531026 Chr.1: 84574533-84574533 on Build GRCh38
rs975854792 Chr.1: 84574274-84574274 on Build GRCh38
rs892015067 Chr.1: 84574500-84574500 on Build GRCh38
rs893260464 Chr.1: 84574635-84574635 on Build GRCh38
rs893177818 Chr.1: 84574409-84574409 on Build GRCh38
rs768406558 Chr.1: 84574263-84574263 on Build GRCh38
rs777532510 Chr.1: 84574444-84574444 on Build GRCh38
rs757082869 Chr.1: 84574342-84574350 on Build GRCh38
rs949332772 Chr.1: 84574574-84574574 on Build GRCh38
rs746648737 Chr.1: 84574451-84574451 on Build GRCh38
rs2303307 Chr.1: 84574272-84574272 on Build GRCh38
rs953122325 Chr.1: 84574607-84574607 on Build GRCh38
rs746560768 Chr.1: 84574383-84574383 on Build GRCh38
rs967065469 Chr.1: 84574483-84574483 on Build GRCh38
rs975786089 Chr.1: 84574226-84574226 on Build GRCh38
rs756074317 Chr.1: 84574341-84574341 on Build GRCh38
rs752943680 Chr.1: 84574204-84574204 on Build GRCh38
rs761290807 Chr.1: 84574284-84574284 on Build GRCh38
rs758471823 Chr.1: 84574315-84574316 on Build GRCh38
rs762490549 Chr.1: 84574245-84574245 on Build GRCh38
rs771685426 Chr.1: 84574351-84574353 on Build GRCh38
rs199591724 Chr.1: 84574442-84574442 on Build GRCh38
rs779844966 Chr.1: 84574247-84574247 on Build GRCh38
rs35860428 Chr.1: 84574461-84574461 on Build GRCh38
rs755986468 Chr.1: 84574242-84574242 on Build GRCh38
rs766570971 Chr.1: 84574316-84574333 on Build GRCh38
rs752500407 Chr.1: 84574441-84574441 on Build GRCh38
rs984571218 Chr.1: 84574293-84574293 on Build GRCh38
rs921582783 Chr.1: 84574234-84574234 on Build GRCh38
rs771709818 Chr.1: 84574380-84574380 on Build GRCh38
rs941592740 Chr.1: 84574325-84574325 on Build GRCh38
rs530177467 Chr.1: 84574396-84574396 on Build GRCh38
rs753835378 Chr.1: 84574436-84574436 on Build GRCh38
rs761927268 Chr.1: 84574424-84574424 on Build GRCh38
rs1001778851 Chr.1: 84574405-84574405 on Build GRCh38
rs12079107 Chr.1: 84574529-84574529 on Build GRCh38
rs780004301 Chr.1: 84574349-84574349 on Build GRCh38
rs777248809 Chr.1: 84574304-84574304 on Build GRCh38
rs11487942 Chr.1: 84574502-84574502 on Build GRCh38

View Full Product Details