Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85582338-85582836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYR61
Entrez Gene ID
3491
Gene Name
cysteine rich angiogenic inducer 61
Gene Aliases
CCN1, GIG1, IGFBP10
UniGene
Hs.8867
Gene Chromosome Location
Chr.1: 85580761-85583967 on Build GRCh38
Transcripts
NM_001554.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348936 c.760G>T p.D254Y Chr.1: 85582541-85582541 on Build GRCh38
5449145 c.925T>C p.Y309H Chr.1: 85582821-85582821 on Build GRCh38
2244104 c.912G>C p.L304F Chr.1: 85582808-85582808 on Build GRCh38
4854614 c.796A>T p.I266F Chr.1: 85582577-85582577 on Build GRCh38
1667676 c.743C>T p.S248F Chr.1: 85582524-85582524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774059615 Chr.1: 85582434-85582434 on Build GRCh38
rs144258395 Chr.1: 85582775-85582775 on Build GRCh38
rs777112490 Chr.1: 85582562-85582562 on Build GRCh38
rs761073058 Chr.1: 85582372-85582372 on Build GRCh38
rs777361186 Chr.1: 85582645-85582645 on Build GRCh38
rs752431884 Chr.1: 85582642-85582642 on Build GRCh38
rs140020167 Chr.1: 85582796-85582796 on Build GRCh38
rs372035630 Chr.1: 85582506-85582506 on Build GRCh38
rs1139746 Chr.1: 85582440-85582440 on Build GRCh38
rs369974800 Chr.1: 85582644-85582644 on Build GRCh38
rs752842278 Chr.1: 85582507-85582507 on Build GRCh38
rs919894583 Chr.1: 85582539-85582539 on Build GRCh38
rs1055615804 Chr.1: 85582640-85582640 on Build GRCh38
rs758131582 Chr.1: 85582787-85582787 on Build GRCh38
rs781188033 Chr.1: 85582404-85582404 on Build GRCh38
rs75947336 Chr.1: 85582584-85582584 on Build GRCh38
rs755938076 Chr.1: 85582527-85582527 on Build GRCh38
rs777365838 Chr.1: 85582668-85582669 on Build GRCh38
rs770173391 Chr.1: 85582662-85582662 on Build GRCh38
rs746492611 Chr.1: 85582647-85582647 on Build GRCh38
rs749074854 Chr.1: 85582529-85582529 on Build GRCh38
rs1010598319 Chr.1: 85582812-85582812 on Build GRCh38
rs3211025 Chr.1: 85582378-85582378 on Build GRCh38
rs762879999 Chr.1: 85582471-85582471 on Build GRCh38
rs751124491 Chr.1: 85582813-85582813 on Build GRCh38
rs759768515 Chr.1: 85582495-85582495 on Build GRCh38
rs766218988 Chr.1: 85582610-85582610 on Build GRCh38
rs3211026 Chr.1: 85582384-85582384 on Build GRCh38
rs771284240 Chr.1: 85582561-85582561 on Build GRCh38
rs754796443 Chr.1: 85582622-85582622 on Build GRCh38
rs770045627 Chr.1: 85582588-85582588 on Build GRCh38
rs3211028 Chr.1: 85582393-85582393 on Build GRCh38
rs1046942147 Chr.1: 85582558-85582558 on Build GRCh38
rs779908967 Chr.1: 85582528-85582528 on Build GRCh38
rs373000437 Chr.1: 85582593-85582593 on Build GRCh38
rs1003234344 Chr.1: 85582639-85582639 on Build GRCh38
rs765129171 Chr.1: 85582727-85582727 on Build GRCh38
rs761766701 Chr.1: 85582675-85582675 on Build GRCh38
rs377358947 Chr.1: 85582682-85582682 on Build GRCh38
rs189866774 Chr.1: 85582672-85582672 on Build GRCh38
rs149037713 Chr.1: 85582427-85582427 on Build GRCh38
rs778674730 Chr.1: 85582536-85582536 on Build GRCh38
rs772560592 Chr.1: 85582556-85582556 on Build GRCh38
rs987482581 Chr.1: 85582742-85582742 on Build GRCh38
rs200203130 Chr.1: 85582535-85582535 on Build GRCh38
rs553932011 Chr.1: 85582421-85582421 on Build GRCh38
rs368281986 Chr.1: 85582398-85582398 on Build GRCh38
rs533841417 Chr.1: 85582416-85582416 on Build GRCh38
rs111410670 Chr.1: 85582739-85582739 on Build GRCh38
rs753851213 Chr.1: 85582719-85582719 on Build GRCh38
rs748958257 Chr.1: 85582723-85582723 on Build GRCh38
rs758449064 Chr.1: 85582508-85582508 on Build GRCh38
rs1038453341 Chr.1: 85582483-85582483 on Build GRCh38
rs747698663 Chr.1: 85582403-85582404 on Build GRCh38
rs376325979 Chr.1: 85582388-85582388 on Build GRCh38
rs112232895 Chr.1: 85582737-85582737 on Build GRCh38
rs377332597 Chr.1: 85582723-85582723 on Build GRCh38
rs780418624 Chr.1: 85582402-85582402 on Build GRCh38
rs143007605 Chr.1: 85582430-85582430 on Build GRCh38
rs139284008 Chr.1: 85582524-85582524 on Build GRCh38
rs754116861 Chr.1: 85582376-85582376 on Build GRCh38
rs759000530 Chr.1: 85582387-85582392 on Build GRCh38
rs747654249 Chr.1: 85582439-85582439 on Build GRCh38
rs909867388 Chr.1: 85582591-85582591 on Build GRCh38
rs113320975 Chr.1: 85582488-85582488 on Build GRCh38
rs372341929 Chr.1: 85582377-85582377 on Build GRCh38
rs775761908 Chr.1: 85582663-85582663 on Build GRCh38
rs143178160 Chr.1: 85582468-85582468 on Build GRCh38
rs1011321526 Chr.1: 85582650-85582650 on Build GRCh38
rs3211027 Chr.1: 85582390-85582390 on Build GRCh38
rs759891344 Chr.1: 85582573-85582573 on Build GRCh38
rs755958207 Chr.1: 85582420-85582420 on Build GRCh38
rs1030786882 Chr.1: 85582782-85582782 on Build GRCh38
rs763172280 Chr.1: 85582669-85582669 on Build GRCh38
rs769402244 Chr.1: 85582534-85582535 on Build GRCh38
rs922667068 Chr.1: 85582517-85582517 on Build GRCh38
rs955636453 Chr.1: 85582713-85582713 on Build GRCh38
rs764029932 Chr.1: 85582520-85582520 on Build GRCh38
rs368750850 Chr.1: 85582590-85582590 on Build GRCh38
rs1804662 Chr.1: 85582441-85582441 on Build GRCh38
rs775710634 Chr.1: 85582589-85582589 on Build GRCh38
rs554505515 Chr.1: 85582391-85582391 on Build GRCh38
rs770435967 Chr.1: 85582742-85582742 on Build GRCh38
rs765305490 Chr.1: 85582379-85582379 on Build GRCh38
rs373437549 Chr.1: 85582652-85582652 on Build GRCh38
rs778805266 Chr.1: 85582623-85582623 on Build GRCh38
rs755450244 Chr.1: 85582708-85582708 on Build GRCh38
rs964204654 Chr.1: 85582459-85582459 on Build GRCh38
rs746322786 Chr.1: 85582653-85582653 on Build GRCh38
rs768362037 Chr.1: 85582433-85582433 on Build GRCh38
rs911391798 Chr.1: 85582743-85582743 on Build GRCh38
rs561300948 Chr.1: 85582611-85582611 on Build GRCh38
rs376931984 Chr.1: 85582541-85582541 on Build GRCh38
rs766562788 Chr.1: 85582375-85582375 on Build GRCh38
rs565430789 Chr.1: 85582442-85582442 on Build GRCh38
rs747560642 Chr.1: 85582557-85582557 on Build GRCh38

View Full Product Details