Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85745294-85745806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL24A1
Entrez Gene ID
255631
Gene Name
collagen type XXIV alpha 1
Gene Aliases
-
UniGene
Hs.659516
Gene Chromosome Location
Chr.1: 85728507-86156987 on Build GRCh38
Transcripts
NM_152890.5 XM_017000924.1 XM_017000925.1 XM_017000926.1 XM_017000927.1 XM_017000928.1 XM_017000929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
106682 c.4490C>T p.A1497V Chr.1: 85745454-85745454 on Build GRCh38
4165319 c.4503+1G>A p.? Chr.1: 85745440-85745440 on Build GRCh38
912726 c.4453A>C p.N1485H Chr.1: 85745491-85745491 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371978066 Chr.1: 85745714-85745714 on Build GRCh38
rs773705386 Chr.1: 85745537-85745537 on Build GRCh38
rs149620215 Chr.1: 85745641-85745641 on Build GRCh38
rs79668890 Chr.1: 85745379-85745379 on Build GRCh38
rs770781533 Chr.1: 85745401-85745401 on Build GRCh38
rs982380931 Chr.1: 85745765-85745765 on Build GRCh38
rs1004950653 Chr.1: 85745683-85745683 on Build GRCh38
rs746883067 Chr.1: 85745397-85745397 on Build GRCh38
rs200957951 Chr.1: 85745499-85745499 on Build GRCh38
rs768231957 Chr.1: 85745527-85745527 on Build GRCh38
rs867725788 Chr.1: 85745498-85745498 on Build GRCh38
rs757109417 Chr.1: 85745494-85745494 on Build GRCh38
rs372445647 Chr.1: 85745490-85745490 on Build GRCh38
rs375393256 Chr.1: 85745495-85745495 on Build GRCh38
rs755142867 Chr.1: 85745482-85745482 on Build GRCh38
rs750752771 Chr.1: 85745437-85745437 on Build GRCh38
rs10747324 Chr.1: 85745642-85745642 on Build GRCh38
rs766811364 Chr.1: 85745555-85745555 on Build GRCh38
rs201899812 Chr.1: 85745511-85745511 on Build GRCh38
rs866939847 Chr.1: 85745528-85745528 on Build GRCh38
rs754046703 Chr.1: 85745458-85745458 on Build GRCh38
rs755051153 Chr.1: 85745391-85745391 on Build GRCh38
rs899153528 Chr.1: 85745532-85745532 on Build GRCh38
rs182390734 Chr.1: 85745704-85745704 on Build GRCh38
rs776266265 Chr.1: 85745408-85745408 on Build GRCh38
rs954920435 Chr.1: 85745363-85745363 on Build GRCh38
rs947014827 Chr.1: 85745586-85745586 on Build GRCh38
rs982288764 Chr.1: 85745360-85745360 on Build GRCh38
rs900382997 Chr.1: 85745628-85745628 on Build GRCh38
rs950562049 Chr.1: 85745335-85745335 on Build GRCh38
rs536265783 Chr.1: 85745621-85745621 on Build GRCh38
rs769517118 Chr.1: 85745506-85745506 on Build GRCh38
rs762454250 Chr.1: 85745542-85745542 on Build GRCh38
rs368306760 Chr.1: 85745425-85745425 on Build GRCh38
rs775035773 Chr.1: 85745413-85745413 on Build GRCh38
rs890286396 Chr.1: 85745646-85745646 on Build GRCh38
rs140391595 Chr.1: 85745454-85745454 on Build GRCh38
rs1014943675 Chr.1: 85745701-85745708 on Build GRCh38
rs546550995 Chr.1: 85745347-85745347 on Build GRCh38
rs185786448 Chr.1: 85745415-85745415 on Build GRCh38
rs994780977 Chr.1: 85745533-85745533 on Build GRCh38
rs768190436 Chr.1: 85745557-85745558 on Build GRCh38
rs772860617 Chr.1: 85745325-85745325 on Build GRCh38
rs544377386 Chr.1: 85745743-85745743 on Build GRCh38
rs758873533 Chr.1: 85745343-85745343 on Build GRCh38
rs529662562 Chr.1: 85745334-85745334 on Build GRCh38
rs758681382 Chr.1: 85745639-85745639 on Build GRCh38
rs143321140 Chr.1: 85745660-85745660 on Build GRCh38
rs551833583 Chr.1: 85745433-85745433 on Build GRCh38
rs763525877 Chr.1: 85745417-85745417 on Build GRCh38
rs761125441 Chr.1: 85745554-85745554 on Build GRCh38
rs766723208 Chr.1: 85745452-85745452 on Build GRCh38
rs571556212 Chr.1: 85745485-85745485 on Build GRCh38
rs756466947 Chr.1: 85745438-85745438 on Build GRCh38
rs1030425178 Chr.1: 85745361-85745361 on Build GRCh38
rs766810819 Chr.1: 85745625-85745625 on Build GRCh38
rs947668348 Chr.1: 85745445-85745445 on Build GRCh38
rs773798539 Chr.1: 85745550-85745550 on Build GRCh38
rs779681429 Chr.1: 85745510-85745510 on Build GRCh38
rs978994085 Chr.1: 85745766-85745766 on Build GRCh38
rs753190047 Chr.1: 85745715-85745715 on Build GRCh38
rs1023895102 Chr.1: 85745719-85745719 on Build GRCh38
rs745560603 Chr.1: 85745410-85745410 on Build GRCh38
rs564415114 Chr.1: 85745748-85745748 on Build GRCh38
rs374691211 Chr.1: 85745544-85745544 on Build GRCh38
rs762208092 Chr.1: 85745429-85745429 on Build GRCh38
rs550863773 Chr.1: 85745556-85745556 on Build GRCh38
rs1042991921 Chr.1: 85745592-85745592 on Build GRCh38
rs777773545 Chr.1: 85745392-85745392 on Build GRCh38
rs190294223 Chr.1: 85745603-85745603 on Build GRCh38

View Full Product Details