Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86486448-86486959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA1
Entrez Gene ID
1179
Gene Name
chloride channel accessory 1
Gene Aliases
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1
UniGene
Hs.194659
Gene Chromosome Location
Chr.1: 86468843-86500294 on Build GRCh38
Transcripts
NM_001285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629769 c.955-1G>C p.? Chr.1: 86486525-86486525 on Build GRCh38
3790309 c.1113C>T p.L371L Chr.1: 86486684-86486684 on Build GRCh38
4444282 c.1055C>T p.A352V Chr.1: 86486626-86486626 on Build GRCh38
7361407 c.1015G>T p.E339* Chr.1: 86486586-86486586 on Build GRCh38
1639987 c.1035G>A p.G345G Chr.1: 86486606-86486606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35944380 Chr.1: 86486631-86486632 on Build GRCh38
rs1001078888 Chr.1: 86486840-86486840 on Build GRCh38
rs772377825 Chr.1: 86486743-86486743 on Build GRCh38
rs965907873 Chr.1: 86486535-86486535 on Build GRCh38
rs202202022 Chr.1: 86486499-86486499 on Build GRCh38
rs375936173 Chr.1: 86486719-86486719 on Build GRCh38
rs750957391 Chr.1: 86486647-86486647 on Build GRCh38
rs772061363 Chr.1: 86486571-86486571 on Build GRCh38
rs763207074 Chr.1: 86486601-86486601 on Build GRCh38
rs778883060 Chr.1: 86486545-86486545 on Build GRCh38
rs1054260078 Chr.1: 86486850-86486850 on Build GRCh38
rs775671791 Chr.1: 86486589-86486589 on Build GRCh38
rs967204544 Chr.1: 86486631-86486631 on Build GRCh38
rs1020249502 Chr.1: 86486925-86486925 on Build GRCh38
rs372957890 Chr.1: 86486774-86486774 on Build GRCh38
rs746971261 Chr.1: 86486722-86486722 on Build GRCh38
rs953391303 Chr.1: 86486708-86486708 on Build GRCh38
rs777755432 Chr.1: 86486720-86486720 on Build GRCh38
rs376230857 Chr.1: 86486756-86486756 on Build GRCh38
rs764940063 Chr.1: 86486768-86486768 on Build GRCh38
rs768789097 Chr.1: 86486802-86486802 on Build GRCh38
rs747987194 Chr.1: 86486565-86486565 on Build GRCh38
rs765908821 Chr.1: 86486744-86486744 on Build GRCh38
rs1024514472 Chr.1: 86486517-86486517 on Build GRCh38
rs749005563 Chr.1: 86486537-86486537 on Build GRCh38
rs111541651 Chr.1: 86486624-86486624 on Build GRCh38
rs919054217 Chr.1: 86486605-86486605 on Build GRCh38
rs779180220 Chr.1: 86486803-86486803 on Build GRCh38
rs181631786 Chr.1: 86486894-86486894 on Build GRCh38
rs2734705 Chr.1: 86486641-86486641 on Build GRCh38
rs533096884 Chr.1: 86486895-86486895 on Build GRCh38
rs751463231 Chr.1: 86486775-86486775 on Build GRCh38
rs189130257 Chr.1: 86486701-86486701 on Build GRCh38
rs776509724 Chr.1: 86486575-86486575 on Build GRCh38
rs200291536 Chr.1: 86486724-86486724 on Build GRCh38
rs751982726 Chr.1: 86486489-86486489 on Build GRCh38
rs764398713 Chr.1: 86486614-86486614 on Build GRCh38
rs866759721 Chr.1: 86486643-86486643 on Build GRCh38
rs766034715 Chr.1: 86486655-86486655 on Build GRCh38
rs780047229 Chr.1: 86486787-86486787 on Build GRCh38
rs906971240 Chr.1: 86486839-86486839 on Build GRCh38
rs774797303 Chr.1: 86486629-86486629 on Build GRCh38
rs749960488 Chr.1: 86486508-86486508 on Build GRCh38
rs267598744 Chr.1: 86486697-86486697 on Build GRCh38
rs770839982 Chr.1: 86486502-86486502 on Build GRCh38
rs749370990 Chr.1: 86486796-86486796 on Build GRCh38
rs892812743 Chr.1: 86486888-86486888 on Build GRCh38
rs139300046 Chr.1: 86486739-86486739 on Build GRCh38
rs5744370 Chr.1: 86486767-86486767 on Build GRCh38
rs540776191 Chr.1: 86486834-86486834 on Build GRCh38
rs762231609 Chr.1: 86486740-86486740 on Build GRCh38
rs569872701 Chr.1: 86486929-86486929 on Build GRCh38
rs1050691888 Chr.1: 86486475-86486475 on Build GRCh38
rs997355940 Chr.1: 86486926-86486926 on Build GRCh38
rs745958086 Chr.1: 86486580-86486580 on Build GRCh38
rs146483639 Chr.1: 86486536-86486536 on Build GRCh38
rs753412414 Chr.1: 86486760-86486760 on Build GRCh38
rs753510323 Chr.1: 86486676-86486676 on Build GRCh38
rs762165395 Chr.1: 86486638-86486638 on Build GRCh38
rs140953339 Chr.1: 86486627-86486627 on Build GRCh38
rs1007402120 Chr.1: 86486892-86486892 on Build GRCh38
rs555597850 Chr.1: 86486936-86486936 on Build GRCh38
rs888054345 Chr.1: 86486487-86486487 on Build GRCh38
rs200267383 Chr.1: 86486694-86486694 on Build GRCh38
rs758196276 Chr.1: 86486699-86486699 on Build GRCh38
rs2075630 Chr.1: 86486918-86486918 on Build GRCh38
rs762376347 Chr.1: 86486727-86486727 on Build GRCh38
rs547511215 Chr.1: 86486880-86486880 on Build GRCh38
rs572178650 Chr.1: 86486773-86486773 on Build GRCh38
rs4630108 Chr.1: 86486531-86486531 on Build GRCh38
rs1011512067 Chr.1: 86486582-86486582 on Build GRCh38
rs761303422 Chr.1: 86486749-86486749 on Build GRCh38
rs369109046 Chr.1: 86486493-86486493 on Build GRCh38
rs754798551 Chr.1: 86486544-86486544 on Build GRCh38
rs561895627 Chr.1: 86486778-86486778 on Build GRCh38
rs535596334 Chr.1: 86486934-86486934 on Build GRCh38
rs866152069 Chr.1: 86486745-86486745 on Build GRCh38
rs535298890 Chr.1: 86486479-86486479 on Build GRCh38
rs527779548 Chr.1: 86486861-86486861 on Build GRCh38
rs757204078 Chr.1: 86486786-86486786 on Build GRCh38
rs755670585 Chr.1: 86486649-86486649 on Build GRCh38
rs1001311586 Chr.1: 86486867-86486867 on Build GRCh38
rs780142864 Chr.1: 86486731-86486731 on Build GRCh38
rs769922423 Chr.1: 86486587-86486587 on Build GRCh38
rs1036461195 Chr.1: 86486494-86486494 on Build GRCh38
rs778793374 Chr.1: 86486685-86486685 on Build GRCh38
rs201143830 Chr.1: 86486516-86486516 on Build GRCh38
rs184789973 Chr.1: 86486684-86486684 on Build GRCh38
rs752352591 Chr.1: 86486771-86486771 on Build GRCh38
rs111337245 Chr.1: 86486732-86486732 on Build GRCh38

View Full Product Details