Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86565033-86565535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
UniGene
Hs.567422
Gene Chromosome Location
Chr.1: 86547076-86580758 on Build GRCh38
Transcripts
NM_012128.3 NR_024602.1 XM_011541015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912806 c.651A>G p.K217K Chr.1: 86565367-86565367 on Build GRCh38
912805 c.642T>G p.S214S Chr.1: 86565358-86565358 on Build GRCh38
6064617 c.605G>A p.G202E Chr.1: 86565321-86565321 on Build GRCh38
6127961 c.697G>A p.E233K Chr.1: 86565413-86565413 on Build GRCh38
1201271 c.724A>C p.S242R Chr.1: 86565440-86565440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757463232 Chr.1: 86565282-86565282 on Build GRCh38
rs1008921890 Chr.1: 86565071-86565071 on Build GRCh38
rs766208605 Chr.1: 86565265-86565265 on Build GRCh38
rs1028276935 Chr.1: 86565161-86565161 on Build GRCh38
rs577055430 Chr.1: 86565160-86565160 on Build GRCh38
rs368568589 Chr.1: 86565372-86565372 on Build GRCh38
rs781238968 Chr.1: 86565381-86565381 on Build GRCh38
rs199996343 Chr.1: 86565479-86565479 on Build GRCh38
rs374848221 Chr.1: 86565487-86565487 on Build GRCh38
rs771656460 Chr.1: 86565420-86565420 on Build GRCh38
rs894637928 Chr.1: 86565443-86565443 on Build GRCh38
rs970420963 Chr.1: 86565363-86565363 on Build GRCh38
rs759776397 Chr.1: 86565358-86565358 on Build GRCh38
rs766008951 Chr.1: 86565489-86565489 on Build GRCh38
rs752668233 Chr.1: 86565459-86565459 on Build GRCh38
rs1049325806 Chr.1: 86565504-86565504 on Build GRCh38
rs756850078 Chr.1: 86565284-86565285 on Build GRCh38
rs759470456 Chr.1: 86565364-86565364 on Build GRCh38
rs750181615 Chr.1: 86565243-86565243 on Build GRCh38
rs374879055 Chr.1: 86565397-86565397 on Build GRCh38
rs749833337 Chr.1: 86565297-86565297 on Build GRCh38
rs202050195 Chr.1: 86565280-86565280 on Build GRCh38
rs903173492 Chr.1: 86565195-86565195 on Build GRCh38
rs753729138 Chr.1: 86565362-86565362 on Build GRCh38
rs771611046 Chr.1: 86565330-86565330 on Build GRCh38
rs753858321 Chr.1: 86565277-86565277 on Build GRCh38
rs201651622 Chr.1: 86565373-86565373 on Build GRCh38
rs767278663 Chr.1: 86565234-86565234 on Build GRCh38
rs758374931 Chr.1: 86565450-86565451 on Build GRCh38
rs781530679 Chr.1: 86565289-86565289 on Build GRCh38
rs746192477 Chr.1: 86565290-86565290 on Build GRCh38
rs922283685 Chr.1: 86565361-86565361 on Build GRCh38
rs772967347 Chr.1: 86565337-86565337 on Build GRCh38
rs372259034 Chr.1: 86565281-86565281 on Build GRCh38
rs545639611 Chr.1: 86565169-86565169 on Build GRCh38
rs111681461 Chr.1: 86565419-86565419 on Build GRCh38
rs765078391 Chr.1: 86565285-86565286 on Build GRCh38
rs770576924 Chr.1: 86565430-86565430 on Build GRCh38
rs755996805 Chr.1: 86565264-86565264 on Build GRCh38
rs934496252 Chr.1: 86565388-86565388 on Build GRCh38
rs76950574 Chr.1: 86565064-86565064 on Build GRCh38
rs774004481 Chr.1: 86565313-86565313 on Build GRCh38
rs976515195 Chr.1: 86565354-86565354 on Build GRCh38
rs371872484 Chr.1: 86565380-86565380 on Build GRCh38
rs1003883261 Chr.1: 86565217-86565217 on Build GRCh38
rs200272807 Chr.1: 86565291-86565291 on Build GRCh38
rs1039479646 Chr.1: 86565436-86565436 on Build GRCh38
rs749747365 Chr.1: 86565404-86565404 on Build GRCh38
rs753494354 Chr.1: 86565280-86565281 on Build GRCh38
rs776403617 Chr.1: 86565432-86565432 on Build GRCh38
rs756363123 Chr.1: 86565485-86565485 on Build GRCh38
rs769283564 Chr.1: 86565411-86565411 on Build GRCh38
rs540522500 Chr.1: 86565074-86565074 on Build GRCh38
rs950887296 Chr.1: 86565199-86565199 on Build GRCh38
rs779621917 Chr.1: 86565414-86565414 on Build GRCh38
rs1014340522 Chr.1: 86565170-86565170 on Build GRCh38
rs2231587 Chr.1: 86565345-86565345 on Build GRCh38
rs759382199 Chr.1: 86565454-86565454 on Build GRCh38
rs760951120 Chr.1: 86565423-86565423 on Build GRCh38
rs372831928 Chr.1: 86565457-86565457 on Build GRCh38
rs1022867536 Chr.1: 86565315-86565315 on Build GRCh38
rs572563650 Chr.1: 86565386-86565386 on Build GRCh38
rs912005058 Chr.1: 86565408-86565408 on Build GRCh38
rs756382428 Chr.1: 86565391-86565391 on Build GRCh38
rs900780070 Chr.1: 86565128-86565128 on Build GRCh38
rs746862618 Chr.1: 86565186-86565186 on Build GRCh38
rs560144850 Chr.1: 86565140-86565140 on Build GRCh38
rs780015722 Chr.1: 86565452-86565453 on Build GRCh38
rs760360793 Chr.1: 86565342-86565342 on Build GRCh38
rs773620823 Chr.1: 86565086-86565086 on Build GRCh38
rs761483149 Chr.1: 86565327-86565327 on Build GRCh38
rs750688779 Chr.1: 86565384-86565384 on Build GRCh38
rs767657183 Chr.1: 86565471-86565471 on Build GRCh38
rs750217175 Chr.1: 86565302-86565303 on Build GRCh38
rs2231588 Chr.1: 86565353-86565353 on Build GRCh38
rs781192602 Chr.1: 86565220-86565220 on Build GRCh38
rs768222952 Chr.1: 86565310-86565310 on Build GRCh38
rs1024762332 Chr.1: 86565190-86565190 on Build GRCh38
rs1039270908 Chr.1: 86565083-86565083 on Build GRCh38
rs78872817 Chr.1: 86565407-86565407 on Build GRCh38
rs894510744 Chr.1: 86565138-86565138 on Build GRCh38
rs1055894638 Chr.1: 86565094-86565095 on Build GRCh38
rs1030853147 Chr.1: 86565247-86565247 on Build GRCh38
rs772703816 Chr.1: 86565422-86565422 on Build GRCh38
rs547846462 Chr.1: 86565461-86565461 on Build GRCh38

View Full Product Details