Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62798495-62798992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGBP
Entrez Gene ID
55257
Gene Name
MRG/MORF4L binding protein
Gene Aliases
C20orf20, Eaf7, MRG15BP, URCC4
UniGene
Hs.443260
Gene Chromosome Location
Chr.20: 62796405-62801738 on Build GRCh38
Transcripts
NM_018270.5 NR_136405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198590 c.316G>A p.V106I Chr.20: 62798632-62798632 on Build GRCh38
3548742 c.289C>T p.P97S Chr.20: 62798605-62798605 on Build GRCh38
6041273 c.353-6C>T p.? Chr.20: 62798969-62798969 on Build GRCh38
2930838 c.367G>A p.E123K Chr.20: 62798989-62798989 on Build GRCh38
419309 c.280G>C p.E94Q Chr.20: 62798596-62798596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890637850 Chr.20: 62798756-62798756 on Build GRCh38
rs376801566 Chr.20: 62798677-62798677 on Build GRCh38
rs1050697422 Chr.20: 62798937-62798937 on Build GRCh38
rs774604957 Chr.20: 62798933-62798933 on Build GRCh38
rs371989322 Chr.20: 62798589-62798589 on Build GRCh38
rs202210224 Chr.20: 62798904-62798904 on Build GRCh38
rs533766953 Chr.20: 62798728-62798728 on Build GRCh38
rs200328645 Chr.20: 62798927-62798927 on Build GRCh38
rs371602170 Chr.20: 62798947-62798947 on Build GRCh38
rs771128804 Chr.20: 62798929-62798929 on Build GRCh38
rs893884184 Chr.20: 62798740-62798740 on Build GRCh38
rs915049628 Chr.20: 62798915-62798915 on Build GRCh38
rs753284300 Chr.20: 62798948-62798948 on Build GRCh38
rs746332690 Chr.20: 62798651-62798651 on Build GRCh38
rs949259062 Chr.20: 62798663-62798663 on Build GRCh38
rs6011385 Chr.20: 62798702-62798702 on Build GRCh38
rs750199904 Chr.20: 62798569-62798570 on Build GRCh38
rs761919072 Chr.20: 62798582-62798582 on Build GRCh38
rs201234145 Chr.20: 62798954-62798954 on Build GRCh38
rs1009116717 Chr.20: 62798765-62798765 on Build GRCh38
rs767281518 Chr.20: 62798552-62798559 on Build GRCh38
rs756050824 Chr.20: 62798955-62798955 on Build GRCh38
rs2294982 Chr.20: 62798573-62798573 on Build GRCh38
rs923852122 Chr.20: 62798936-62798936 on Build GRCh38
rs768182100 Chr.20: 62798634-62798634 on Build GRCh38
rs775400241 Chr.20: 62798577-62798577 on Build GRCh38
rs746911210 Chr.20: 62798925-62798925 on Build GRCh38
rs902475751 Chr.20: 62798678-62798678 on Build GRCh38
rs369212207 Chr.20: 62798715-62798715 on Build GRCh38
rs371337407 Chr.20: 62798538-62798538 on Build GRCh38
rs921222120 Chr.20: 62798930-62798932 on Build GRCh38
rs759870218 Chr.20: 62798938-62798938 on Build GRCh38
rs766780072 Chr.20: 62798631-62798631 on Build GRCh38
rs537790334 Chr.20: 62798819-62798819 on Build GRCh38
rs988230217 Chr.20: 62798890-62798890 on Build GRCh38
rs777374298 Chr.20: 62798674-62798674 on Build GRCh38
rs369948386 Chr.20: 62798967-62798967 on Build GRCh38
rs538209955 Chr.20: 62798862-62798862 on Build GRCh38
rs375012435 Chr.20: 62798547-62798547 on Build GRCh38
rs748897215 Chr.20: 62798539-62798539 on Build GRCh38
rs773673511 Chr.20: 62798720-62798720 on Build GRCh38
rs952420312 Chr.20: 62798885-62798885 on Build GRCh38
rs751931246 Chr.20: 62798632-62798632 on Build GRCh38
rs770787749 Chr.20: 62798542-62798542 on Build GRCh38
rs946589303 Chr.20: 62798561-62798561 on Build GRCh38
rs756490940 Chr.20: 62798891-62798891 on Build GRCh38
rs757677847 Chr.20: 62798568-62798568 on Build GRCh38
rs577225408 Chr.20: 62798859-62798859 on Build GRCh38
rs189177039 Chr.20: 62798780-62798780 on Build GRCh38
rs1026681957 Chr.20: 62798866-62798866 on Build GRCh38
rs759944792 Chr.20: 62798633-62798633 on Build GRCh38
rs779991761 Chr.20: 62798924-62798924 on Build GRCh38
rs763078636 Chr.20: 62798928-62798928 on Build GRCh38
rs763019944 Chr.20: 62798618-62798618 on Build GRCh38
rs969779923 Chr.20: 62798753-62798753 on Build GRCh38
rs60815360 Chr.20: 62798777-62798777 on Build GRCh38
rs780037133 Chr.20: 62798831-62798831 on Build GRCh38
rs987948950 Chr.20: 62798516-62798516 on Build GRCh38
rs781439740 Chr.20: 62798572-62798572 on Build GRCh38
rs556038426 Chr.20: 62798822-62798822 on Build GRCh38
rs769818011 Chr.20: 62798583-62798583 on Build GRCh38
rs267606045 Chr.20: 62798617-62798617 on Build GRCh38
rs76584956 Chr.20: 62798751-62798751 on Build GRCh38
rs972803765 Chr.20: 62798861-62798861 on Build GRCh38
rs542337527 Chr.20: 62798918-62798918 on Build GRCh38
rs778614608 Chr.20: 62798700-62798700 on Build GRCh38
rs769868843 Chr.20: 62798926-62798926 on Build GRCh38
rs745831456 Chr.20: 62798569-62798569 on Build GRCh38
rs371948410 Chr.20: 62798643-62798643 on Build GRCh38
rs1036898164 Chr.20: 62798742-62798742 on Build GRCh38
rs1035035683 Chr.20: 62798862-62798865 on Build GRCh38
rs761193016 Chr.20: 62798949-62798949 on Build GRCh38
rs567868558 Chr.20: 62798750-62798750 on Build GRCh38
rs201989646 Chr.20: 62798575-62798575 on Build GRCh38
rs755705192 Chr.20: 62798668-62798668 on Build GRCh38
rs553602361 Chr.20: 62798893-62798893 on Build GRCh38
rs566920623 Chr.20: 62798714-62798714 on Build GRCh38
rs562342206 Chr.20: 62798955-62798955 on Build GRCh38
rs572079326 Chr.20: 62798914-62798914 on Build GRCh38
rs756785638 Chr.20: 62798966-62798966 on Build GRCh38
rs1054953124 Chr.20: 62798605-62798605 on Build GRCh38
rs879041564 Chr.20: 62798644-62798644 on Build GRCh38
rs753346008 Chr.20: 62798962-62798962 on Build GRCh38

View Full Product Details