Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63253743-63254253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ16779
Entrez Gene ID
100192386
Gene Name
uncharacterized LOC100192386
Gene Aliases
-
UniGene
Hs.111539
Gene Chromosome Location
Chr.20: 63253978-63261615 on Build GRCh38
Transcripts
NR_024389.1 NM_152864.3 XM_005260192.1 XM_005260194.3 XM_011528527.1 XM_011528528.2 XM_011528529.2 XM_017027636.1 XM_017027637.1 XM_017027638.1 XM_017027639.1

Gene Symbol
NKAIN4
Entrez Gene ID
128414
Gene Name
Na+/K+ transporting ATPase interacting 4
Gene Aliases
C20orf58, FAM77A, bA261N11.2
UniGene
Hs.46627
Gene Chromosome Location
Chr.20: 63240784-63254904 on Build GRCh38
Transcripts
NR_024389.1 NM_152864.3 XM_005260192.1 XM_005260194.3 XM_011528527.1 XM_011528528.2 XM_011528529.2 XM_017027636.1 XM_017027637.1 XM_017027638.1 XM_017027639.1

SNP Detail

SNP ID Chromosome Location
rs1051743688 Chr.20: 63254114-63254114 on Build GRCh38
rs891383670 Chr.20: 63254125-63254125 on Build GRCh38
rs936110812 Chr.20: 63254091-63254091 on Build GRCh38
rs997001730 Chr.20: 63253766-63253766 on Build GRCh38
rs920488879 Chr.20: 63253871-63253871 on Build GRCh38
rs964415484 Chr.20: 63253943-63253943 on Build GRCh38
rs953449048 Chr.20: 63254138-63254138 on Build GRCh38
rs958576948 Chr.20: 63254206-63254206 on Build GRCh38
rs1027319481 Chr.20: 63254194-63254194 on Build GRCh38
rs868117145 Chr.20: 63253984-63253984 on Build GRCh38
rs954471345 Chr.20: 63253804-63253804 on Build GRCh38
rs914227812 Chr.20: 63253783-63253783 on Build GRCh38
rs1008908129 Chr.20: 63254143-63254143 on Build GRCh38
rs115893263 Chr.20: 63253933-63253933 on Build GRCh38
rs34203226 Chr.20: 63254100-63254100 on Build GRCh38
rs1037228491 Chr.20: 63254160-63254160 on Build GRCh38
rs149519714 Chr.20: 63254228-63254228 on Build GRCh38
rs961809963 Chr.20: 63254034-63254034 on Build GRCh38
rs1014646302 Chr.20: 63254007-63254007 on Build GRCh38
rs1014594573 Chr.20: 63253825-63253825 on Build GRCh38
rs1004363174 Chr.20: 63254132-63254132 on Build GRCh38
rs1002702945 Chr.20: 63253901-63253901 on Build GRCh38
rs897567778 Chr.20: 63254173-63254173 on Build GRCh38
rs1012593587 Chr.20: 63254223-63254223 on Build GRCh38
rs934229178 Chr.20: 63253966-63253966 on Build GRCh38
rs1038402081 Chr.20: 63253873-63253873 on Build GRCh38
rs989811393 Chr.20: 63253780-63253780 on Build GRCh38
rs865940794 Chr.20: 63253840-63253840 on Build GRCh38
rs964810779 Chr.20: 63254092-63254092 on Build GRCh38
rs946983645 Chr.20: 63253789-63253789 on Build GRCh38
rs147378830 Chr.20: 63253878-63253878 on Build GRCh38
rs940179997 Chr.20: 63254196-63254196 on Build GRCh38
rs112145752 Chr.20: 63253821-63253821 on Build GRCh38
rs59774035 Chr.20: 63253852-63253853 on Build GRCh38
rs868625814 Chr.20: 63253826-63253826 on Build GRCh38
rs934176073 Chr.20: 63253822-63253822 on Build GRCh38
rs538986906 Chr.20: 63254106-63254106 on Build GRCh38
rs950161701 Chr.20: 63253894-63253894 on Build GRCh38
rs977237934 Chr.20: 63254081-63254081 on Build GRCh38
rs6090333 Chr.20: 63253823-63253823 on Build GRCh38
rs8119038 Chr.20: 63253854-63253854 on Build GRCh38
rs991327565 Chr.20: 63253882-63253882 on Build GRCh38
rs34830843 Chr.20: 63254107-63254107 on Build GRCh38
rs994488705 Chr.20: 63254037-63254037 on Build GRCh38
rs997536148 Chr.20: 63253976-63253976 on Build GRCh38
rs971682189 Chr.20: 63253892-63253892 on Build GRCh38
rs971605259 Chr.20: 63254079-63254079 on Build GRCh38
rs944646756 Chr.20: 63254029-63254040 on Build GRCh38
rs759743339 Chr.20: 63254133-63254133 on Build GRCh38
rs1031460696 Chr.20: 63253921-63253921 on Build GRCh38
rs552176152 Chr.20: 63254070-63254070 on Build GRCh38
rs374777999 Chr.20: 63253926-63253926 on Build GRCh38
rs1041745875 Chr.20: 63253911-63253911 on Build GRCh38
rs138018661 Chr.20: 63253967-63253967 on Build GRCh38
rs907522677 Chr.20: 63253924-63253935 on Build GRCh38
rs940146943 Chr.20: 63254019-63254020 on Build GRCh38
rs1027354854 Chr.20: 63254048-63254048 on Build GRCh38
rs767750832 Chr.20: 63254200-63254200 on Build GRCh38
rs917176122 Chr.20: 63254072-63254072 on Build GRCh38
rs369233937 Chr.20: 63253813-63253813 on Build GRCh38
rs563077872 Chr.20: 63253835-63253836 on Build GRCh38
rs994624846 Chr.20: 63254174-63254174 on Build GRCh38
rs112990479 Chr.20: 63253880-63253880 on Build GRCh38
rs973080870 Chr.20: 63253866-63253866 on Build GRCh38
rs908691023 Chr.20: 63254015-63254015 on Build GRCh38
rs11696191 Chr.20: 63253853-63253853 on Build GRCh38
rs375293341 Chr.20: 63253851-63253851 on Build GRCh38
rs921567599 Chr.20: 63253962-63253962 on Build GRCh38
rs1038477709 Chr.20: 63254024-63254024 on Build GRCh38
rs987658773 Chr.20: 63253807-63253807 on Build GRCh38
rs891325241 Chr.20: 63253987-63253987 on Build GRCh38
rs530409053 Chr.20: 63254002-63254002 on Build GRCh38
rs565887070 Chr.20: 63254217-63254217 on Build GRCh38
rs1030266337 Chr.20: 63253802-63253802 on Build GRCh38
rs908764491 Chr.20: 63254175-63254175 on Build GRCh38
rs924674475 Chr.20: 63254086-63254086 on Build GRCh38
rs6010885 Chr.20: 63253886-63253886 on Build GRCh38
rs186416769 Chr.20: 63253951-63253951 on Build GRCh38
rs932913291 Chr.20: 63254110-63254110 on Build GRCh38
rs761698318 Chr.20: 63254094-63254094 on Build GRCh38
rs958643449 Chr.20: 63254067-63254067 on Build GRCh38

View Full Product Details