Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63255631-63256135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ16779
Entrez Gene ID
100192386
Gene Name
uncharacterized LOC100192386
Gene Aliases
-
UniGene
Hs.111539
Gene Chromosome Location
Chr.20: 63253978-63261615 on Build GRCh38
Transcripts
NR_024389.1

SNP Detail

SNP ID Chromosome Location
rs993871033 Chr.20: 63255928-63255928 on Build GRCh38
rs913626572 Chr.20: 63256010-63256010 on Build GRCh38
rs76183812 Chr.20: 63255914-63255914 on Build GRCh38
rs114491067 Chr.20: 63255875-63255875 on Build GRCh38
rs993551284 Chr.20: 63256107-63256107 on Build GRCh38
rs553448786 Chr.20: 63255754-63255754 on Build GRCh38
rs770378897 Chr.20: 63255715-63255715 on Build GRCh38
rs531016404 Chr.20: 63255665-63255665 on Build GRCh38
rs545608885 Chr.20: 63255915-63255915 on Build GRCh38
rs751901831 Chr.20: 63256075-63256075 on Build GRCh38
rs552678636 Chr.20: 63255717-63255717 on Build GRCh38
rs755842832 Chr.20: 63255965-63255965 on Build GRCh38
rs965007504 Chr.20: 63255911-63255911 on Build GRCh38
rs534991924 Chr.20: 63255725-63255725 on Build GRCh38
rs1003509673 Chr.20: 63255787-63255787 on Build GRCh38
rs926266964 Chr.20: 63256060-63256060 on Build GRCh38
rs762157836 Chr.20: 63255945-63255945 on Build GRCh38
rs114797814 Chr.20: 63255720-63255720 on Build GRCh38
rs918482181 Chr.20: 63255932-63255932 on Build GRCh38
rs767633090 Chr.20: 63255950-63255950 on Build GRCh38
rs752270577 Chr.20: 63256102-63256102 on Build GRCh38
rs976625440 Chr.20: 63255658-63255658 on Build GRCh38
rs557416642 Chr.20: 63255918-63255918 on Build GRCh38
rs561388136 Chr.20: 63256045-63256045 on Build GRCh38
rs542463880 Chr.20: 63256059-63256059 on Build GRCh38
rs1027575268 Chr.20: 63255943-63255943 on Build GRCh38
rs1045071861 Chr.20: 63255747-63255747 on Build GRCh38
rs534057667 Chr.20: 63255856-63255856 on Build GRCh38
rs944826411 Chr.20: 63256084-63256084 on Build GRCh38
rs554311144 Chr.20: 63255864-63255864 on Build GRCh38
rs928068062 Chr.20: 63255961-63255961 on Build GRCh38
rs1027954540 Chr.20: 63255724-63255724 on Build GRCh38
rs116769520 Chr.20: 63255958-63255958 on Build GRCh38
rs889231205 Chr.20: 63255895-63255895 on Build GRCh38
rs535699232 Chr.20: 63255874-63255874 on Build GRCh38
rs6011703 Chr.20: 63256101-63256101 on Build GRCh38
rs991815815 Chr.20: 63255836-63255836 on Build GRCh38
rs144264151 Chr.20: 63255946-63255946 on Build GRCh38
rs190899537 Chr.20: 63256082-63256082 on Build GRCh38
rs985652685 Chr.20: 63255877-63255877 on Build GRCh38
rs776088427 Chr.20: 63255805-63255805 on Build GRCh38
rs368618433 Chr.20: 63255903-63255903 on Build GRCh38
rs77222937 Chr.20: 63255770-63255770 on Build GRCh38
rs980973998 Chr.20: 63255732-63255732 on Build GRCh38
rs948163139 Chr.20: 63255731-63255731 on Build GRCh38
rs766883343 Chr.20: 63255977-63255977 on Build GRCh38
rs913553086 Chr.20: 63255852-63255852 on Build GRCh38
rs892739283 Chr.20: 63255855-63255855 on Build GRCh38
rs1033461961 Chr.20: 63255957-63255957 on Build GRCh38
rs991889376 Chr.20: 63255999-63255999 on Build GRCh38
rs146582663 Chr.20: 63256058-63256058 on Build GRCh38
rs929899838 Chr.20: 63255940-63255940 on Build GRCh38

View Full Product Details