Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63259375-63259887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ16779
Entrez Gene ID
100192386
Gene Name
uncharacterized LOC100192386
Gene Aliases
-
UniGene
Hs.111539
Gene Chromosome Location
Chr.20: 63253978-63261615 on Build GRCh38
Transcripts
NR_024389.1

SNP Detail

SNP ID Chromosome Location
rs889763174 Chr.20: 63259576-63259576 on Build GRCh38
rs547869852 Chr.20: 63259577-63259577 on Build GRCh38
rs566030723 Chr.20: 63259717-63259717 on Build GRCh38
rs995596990 Chr.20: 63259826-63259826 on Build GRCh38
rs928519887 Chr.20: 63259525-63259525 on Build GRCh38
rs572528686 Chr.20: 63259418-63259418 on Build GRCh38
rs746030117 Chr.20: 63259488-63259488 on Build GRCh38
rs184938598 Chr.20: 63259419-63259419 on Build GRCh38
rs897404951 Chr.20: 63259751-63259751 on Build GRCh38
rs947165604 Chr.20: 63259743-63259743 on Build GRCh38
rs530444014 Chr.20: 63259645-63259645 on Build GRCh38
rs1027608347 Chr.20: 63259608-63259608 on Build GRCh38
rs915672215 Chr.20: 63259637-63259637 on Build GRCh38
rs573767958 Chr.20: 63259700-63259701 on Build GRCh38
rs774296656 Chr.20: 63259642-63259642 on Build GRCh38
rs1045594106 Chr.20: 63259503-63259503 on Build GRCh38
rs891375521 Chr.20: 63259607-63259607 on Build GRCh38
rs948364791 Chr.20: 63259640-63259640 on Build GRCh38
rs551769354 Chr.20: 63259544-63259545 on Build GRCh38
rs1011707275 Chr.20: 63259567-63259567 on Build GRCh38
rs554769576 Chr.20: 63259810-63259810 on Build GRCh38
rs1020068933 Chr.20: 63259720-63259720 on Build GRCh38
rs986979333 Chr.20: 63259453-63259453 on Build GRCh38
rs994790424 Chr.20: 63259773-63259773 on Build GRCh38
rs544618672 Chr.20: 63259815-63259816 on Build GRCh38
rs775216879 Chr.20: 63259565-63259565 on Build GRCh38
rs541263870 Chr.20: 63259557-63259557 on Build GRCh38
rs577094977 Chr.20: 63259442-63259450 on Build GRCh38
rs989105733 Chr.20: 63259860-63259860 on Build GRCh38
rs1027243877 Chr.20: 63259780-63259780 on Build GRCh38
rs559163919 Chr.20: 63259562-63259562 on Build GRCh38
rs964348252 Chr.20: 63259413-63259413 on Build GRCh38
rs1051410593 Chr.20: 63259794-63259794 on Build GRCh38
rs1042837569 Chr.20: 63259752-63259752 on Build GRCh38
rs961544823 Chr.20: 63259424-63259424 on Build GRCh38
rs928555862 Chr.20: 63259648-63259648 on Build GRCh38
rs756278494 Chr.20: 63259682-63259682 on Build GRCh38
rs6122109 Chr.20: 63259714-63259714 on Build GRCh38
rs1042806008 Chr.20: 63259571-63259571 on Build GRCh38
rs207477710 Chr.20: 63259504-63259504 on Build GRCh38
rs766021261 Chr.20: 63259845-63259845 on Build GRCh38
rs576368661 Chr.20: 63259476-63259476 on Build GRCh38
rs973336872 Chr.20: 63259448-63259448 on Build GRCh38
rs1052954320 Chr.20: 63259674-63259674 on Build GRCh38
rs948697790 Chr.20: 63259512-63259512 on Build GRCh38
rs529602162 Chr.20: 63259525-63259525 on Build GRCh38
rs889898580 Chr.20: 63259716-63259716 on Build GRCh38
rs6011704 Chr.20: 63259477-63259477 on Build GRCh38
rs55940436 Chr.20: 63259551-63259551 on Build GRCh38
rs781396231 Chr.20: 63259487-63259487 on Build GRCh38
rs1019847744 Chr.20: 63259585-63259585 on Build GRCh38
rs1015749121 Chr.20: 63259638-63259638 on Build GRCh38
rs892920749 Chr.20: 63259686-63259686 on Build GRCh38
rs1028362198 Chr.20: 63259400-63259400 on Build GRCh38
rs116549677 Chr.20: 63259673-63259673 on Build GRCh38
rs369023710 Chr.20: 63259774-63259774 on Build GRCh38
rs931675232 Chr.20: 63259764-63259764 on Build GRCh38
rs952851148 Chr.20: 63259782-63259782 on Build GRCh38
rs563246033 Chr.20: 63259617-63259617 on Build GRCh38
rs550024996 Chr.20: 63259655-63259655 on Build GRCh38
rs969807537 Chr.20: 63259843-63259843 on Build GRCh38
rs972919555 Chr.20: 63259598-63259598 on Build GRCh38
rs981122626 Chr.20: 63259844-63259844 on Build GRCh38
rs148239296 Chr.20: 63259449-63259449 on Build GRCh38
rs955735042 Chr.20: 63259847-63259847 on Build GRCh38
rs948395472 Chr.20: 63259496-63259496 on Build GRCh38
rs747109322 Chr.20: 63259405-63259405 on Build GRCh38
rs897323419 Chr.20: 63259819-63259819 on Build GRCh38
rs768526446 Chr.20: 63259593-63259593 on Build GRCh38
rs866711747 Chr.20: 63259748-63259748 on Build GRCh38
rs912831347 Chr.20: 63259423-63259423 on Build GRCh38
rs529845816 Chr.20: 63259574-63259574 on Build GRCh38
rs1016235855 Chr.20: 63259818-63259818 on Build GRCh38
rs934491945 Chr.20: 63259532-63259532 on Build GRCh38
rs900245859 Chr.20: 63259761-63259761 on Build GRCh38
rs1028419534 Chr.20: 63259568-63259568 on Build GRCh38
rs984468464 Chr.20: 63259633-63259633 on Build GRCh38
rs141765104 Chr.20: 63259524-63259524 on Build GRCh38
rs538824531 Chr.20: 63259699-63259699 on Build GRCh38
rs6122418 Chr.20: 63259437-63259437 on Build GRCh38
rs760464605 Chr.20: 63259814-63259814 on Build GRCh38
rs952886755 Chr.20: 63259623-63259623 on Build GRCh38

View Full Product Details