Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63309666-63310168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL20A1
Entrez Gene ID
57642
Gene Name
collagen type XX alpha 1 chain
Gene Aliases
bA261N11.4
UniGene
Hs.271285
Gene Chromosome Location
Chr.20: 63293169-63334847 on Build GRCh38
Transcripts
NM_020882.2 XM_011528937.1 XM_011528938.1 XM_011528939.1 XM_011528940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
359727 c.1118C>A p.A373D Chr.20: 63309770-63309770 on Build GRCh38
2931663 c.1125C>G p.D375E Chr.20: 63309777-63309777 on Build GRCh38
5180716 c.1239C>T p.A413A Chr.20: 63309891-63309891 on Build GRCh38
6526076 c.1259C>A p.A420D Chr.20: 63309890-63309890 on Build GRCh38
6526077 c.1238C>A p.A413D Chr.20: 63309890-63309890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768754881 Chr.20: 63309781-63309781 on Build GRCh38
rs986635188 Chr.20: 63309778-63309778 on Build GRCh38
rs575632513 Chr.20: 63310137-63310137 on Build GRCh38
rs370343751 Chr.20: 63309909-63309909 on Build GRCh38
rs1011546109 Chr.20: 63309850-63309850 on Build GRCh38
rs765071435 Chr.20: 63309926-63309926 on Build GRCh38
rs912376488 Chr.20: 63309793-63309793 on Build GRCh38
rs778864152 Chr.20: 63309761-63309761 on Build GRCh38
rs535765739 Chr.20: 63310115-63310115 on Build GRCh38
rs923039373 Chr.20: 63309686-63309687 on Build GRCh38
rs186757692 Chr.20: 63309865-63309865 on Build GRCh38
rs773573190 Chr.20: 63309800-63309800 on Build GRCh38
rs1003007114 Chr.20: 63310028-63310028 on Build GRCh38
rs377073867 Chr.20: 63309851-63309851 on Build GRCh38
rs374436824 Chr.20: 63309709-63309709 on Build GRCh38
rs777540579 Chr.20: 63309818-63309820 on Build GRCh38
rs762608350 Chr.20: 63309960-63309960 on Build GRCh38
rs760071443 Chr.20: 63309893-63309893 on Build GRCh38
rs774321889 Chr.20: 63309809-63309809 on Build GRCh38
rs771294705 Chr.20: 63309887-63309887 on Build GRCh38
rs376170843 Chr.20: 63309944-63309944 on Build GRCh38
rs550314382 Chr.20: 63309949-63309949 on Build GRCh38
rs1044526845 Chr.20: 63309787-63309787 on Build GRCh38
rs755838084 Chr.20: 63309824-63309824 on Build GRCh38
rs568473613 Chr.20: 63310089-63310089 on Build GRCh38
rs1055325742 Chr.20: 63309715-63309715 on Build GRCh38
rs6010897 Chr.20: 63309975-63309975 on Build GRCh38
rs763464712 Chr.20: 63309728-63309728 on Build GRCh38
rs758362254 Chr.20: 63309931-63309931 on Build GRCh38
rs983326841 Chr.20: 63309860-63309860 on Build GRCh38
rs774897563 Chr.20: 63309811-63309811 on Build GRCh38
rs763769391 Chr.20: 63309832-63309832 on Build GRCh38
rs6010896 Chr.20: 63309927-63309927 on Build GRCh38
rs1019938029 Chr.20: 63310098-63310098 on Build GRCh38
rs201122273 Chr.20: 63309744-63309744 on Build GRCh38
rs750264954 Chr.20: 63309959-63309959 on Build GRCh38
rs777692696 Chr.20: 63309746-63309746 on Build GRCh38
rs372427337 Chr.20: 63309752-63309752 on Build GRCh38
rs1011256129 Chr.20: 63310006-63310006 on Build GRCh38
rs768280305 Chr.20: 63309948-63309948 on Build GRCh38
rs752366140 Chr.20: 63309745-63309745 on Build GRCh38
rs907583472 Chr.20: 63309940-63309940 on Build GRCh38
rs752798985 Chr.20: 63309934-63309934 on Build GRCh38
rs759007324 Chr.20: 63309812-63309812 on Build GRCh38
rs774785628 Chr.20: 63309890-63309890 on Build GRCh38
rs895407909 Chr.20: 63309916-63309916 on Build GRCh38
rs778009763 Chr.20: 63309939-63309939 on Build GRCh38
rs753512685 Chr.20: 63309918-63309918 on Build GRCh38
rs757055637 Chr.20: 63309848-63309848 on Build GRCh38
rs781346441 Chr.20: 63309861-63309861 on Build GRCh38
rs367659751 Chr.20: 63309943-63309944 on Build GRCh38
rs6062885 Chr.20: 63310142-63310142 on Build GRCh38
rs552487165 Chr.20: 63310020-63310020 on Build GRCh38
rs555669221 Chr.20: 63310116-63310116 on Build GRCh38
rs779928552 Chr.20: 63309933-63309933 on Build GRCh38
rs73921038 Chr.20: 63310040-63310040 on Build GRCh38
rs769946191 Chr.20: 63309798-63309798 on Build GRCh38
rs771464744 Chr.20: 63309928-63309929 on Build GRCh38
rs1025064451 Chr.20: 63309732-63309732 on Build GRCh38
rs34485195 Chr.20: 63309803-63309804 on Build GRCh38
rs775141604 Chr.20: 63309905-63309905 on Build GRCh38
rs200364190 Chr.20: 63309941-63309941 on Build GRCh38
rs372938275 Chr.20: 63309880-63309880 on Build GRCh38
rs763265567 Chr.20: 63309806-63309806 on Build GRCh38
rs568787488 Chr.20: 63309958-63309958 on Build GRCh38
rs752283267 Chr.20: 63309752-63309754 on Build GRCh38
rs766845299 Chr.20: 63309738-63309738 on Build GRCh38
rs777924541 Chr.20: 63309886-63309886 on Build GRCh38
rs746148096 Chr.20: 63309943-63309943 on Build GRCh38
rs368982947 Chr.20: 63309857-63309857 on Build GRCh38
rs377006506 Chr.20: 63309762-63309762 on Build GRCh38
rs746638053 Chr.20: 63309892-63309893 on Build GRCh38
rs776788181 Chr.20: 63309792-63309792 on Build GRCh38
rs752451966 Chr.20: 63309822-63309822 on Build GRCh38
rs200924087 Chr.20: 63309820-63309820 on Build GRCh38
rs1037457773 Chr.20: 63309688-63309688 on Build GRCh38
rs761582929 Chr.20: 63309921-63309921 on Build GRCh38
rs765585805 Chr.20: 63309717-63309717 on Build GRCh38
rs766815439 Chr.20: 63309807-63309807 on Build GRCh38
rs763890512 Chr.20: 63309913-63309913 on Build GRCh38
rs764730744 Chr.20: 63309953-63309953 on Build GRCh38
rs964604875 Chr.20: 63309879-63309879 on Build GRCh38
rs779345310 Chr.20: 63309942-63309942 on Build GRCh38
rs370160805 Chr.20: 63309810-63309810 on Build GRCh38
rs920055705 Chr.20: 63310136-63310136 on Build GRCh38
rs372246087 Chr.20: 63309713-63309713 on Build GRCh38
rs1053622945 Chr.20: 63310032-63310032 on Build GRCh38
rs757779465 Chr.20: 63309770-63309770 on Build GRCh38
rs989813591 Chr.20: 63309749-63309749 on Build GRCh38
rs201491872 Chr.20: 63309833-63309833 on Build GRCh38
rs375988776 Chr.20: 63309899-63309899 on Build GRCh38
rs748358276 Chr.20: 63309794-63309794 on Build GRCh38
rs376834639 Chr.20: 63309726-63309726 on Build GRCh38
rs745817287 Chr.20: 63309856-63309856 on Build GRCh38
rs566748282 Chr.20: 63310039-63310039 on Build GRCh38
rs535898444 Chr.20: 63310111-63310111 on Build GRCh38
rs570789062 Chr.20: 63310023-63310023 on Build GRCh38
rs762209765 Chr.20: 63309716-63309716 on Build GRCh38
rs995901504 Chr.20: 63309706-63309706 on Build GRCh38
rs756162170 Chr.20: 63309937-63309937 on Build GRCh38
rs761545171 Chr.20: 63309951-63309951 on Build GRCh38
rs546168021 Chr.20: 63309701-63309701 on Build GRCh38

View Full Product Details