Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63343985-63344525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA4
Entrez Gene ID
1137
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4
UniGene
Hs.10734
Gene Chromosome Location
Chr.20: 63343310-63375392 on Build GRCh38
Transcripts
NM_000744.6 NM_001256573.1 NR_046317.1 XM_011528524.1 XM_017027624.1 XM_017027625.1

SNP Detail

SNP ID Chromosome Location
rs1035381401 Chr.20: 63344152-63344152 on Build GRCh38
rs201245835 Chr.20: 63344221-63344221 on Build GRCh38
rs200615176 Chr.20: 63344119-63344119 on Build GRCh38
rs902086464 Chr.20: 63344013-63344013 on Build GRCh38
rs1012080747 Chr.20: 63344261-63344261 on Build GRCh38
rs960641577 Chr.20: 63344153-63344153 on Build GRCh38
rs200979498 Chr.20: 63344199-63344199 on Build GRCh38
rs200412459 Chr.20: 63344241-63344241 on Build GRCh38
rs1016606702 Chr.20: 63344073-63344073 on Build GRCh38
rs764318593 Chr.20: 63344017-63344017 on Build GRCh38
rs199755208 Chr.20: 63344450-63344450 on Build GRCh38
rs568329035 Chr.20: 63344477-63344478 on Build GRCh38
rs959564465 Chr.20: 63344082-63344082 on Build GRCh38
rs775712921 Chr.20: 63344242-63344242 on Build GRCh38
rs199627762 Chr.20: 63344027-63344027 on Build GRCh38
rs200237123 Chr.20: 63344016-63344016 on Build GRCh38
rs199587304 Chr.20: 63344050-63344050 on Build GRCh38
rs199513105 Chr.20: 63344306-63344306 on Build GRCh38
rs200336430 Chr.20: 63344437-63344437 on Build GRCh38
rs199639298 Chr.20: 63344498-63344498 on Build GRCh38
rs200512738 Chr.20: 63344488-63344488 on Build GRCh38
rs201646124 Chr.20: 63344335-63344335 on Build GRCh38
rs201585577 Chr.20: 63344449-63344449 on Build GRCh38
rs967875887 Chr.20: 63344391-63344391 on Build GRCh38
rs943399543 Chr.20: 63344191-63344191 on Build GRCh38
rs200893855 Chr.20: 63344283-63344283 on Build GRCh38
rs398040812 Chr.20: 63344495-63344495 on Build GRCh38
rs758189811 Chr.20: 63344090-63344090 on Build GRCh38
rs199992343 Chr.20: 63344340-63344340 on Build GRCh38
rs780806887 Chr.20: 63344173-63344173 on Build GRCh38
rs201280003 Chr.20: 63344494-63344494 on Build GRCh38
rs774484137 Chr.20: 63344198-63344198 on Build GRCh38
rs201805170 Chr.20: 63344309-63344309 on Build GRCh38
rs201866103 Chr.20: 63344255-63344255 on Build GRCh38
rs200489905 Chr.20: 63344038-63344038 on Build GRCh38
rs189687483 Chr.20: 63344258-63344258 on Build GRCh38
rs200819632 Chr.20: 63344329-63344329 on Build GRCh38
rs942904686 Chr.20: 63344172-63344172 on Build GRCh38
rs989647383 Chr.20: 63344156-63344156 on Build GRCh38
rs751754468 Chr.20: 63344188-63344189 on Build GRCh38
rs1009499235 Chr.20: 63344326-63344326 on Build GRCh38
rs757993356 Chr.20: 63344404-63344404 on Build GRCh38
rs1005066999 Chr.20: 63344251-63344252 on Build GRCh38
rs530341346 Chr.20: 63344149-63344149 on Build GRCh38
rs202107761 Chr.20: 63344215-63344215 on Build GRCh38
rs988401548 Chr.20: 63344159-63344159 on Build GRCh38
rs910599912 Chr.20: 63344190-63344190 on Build GRCh38
rs761584910 Chr.20: 63344388-63344391 on Build GRCh38
rs201603507 Chr.20: 63344305-63344305 on Build GRCh38
rs11480478 Chr.20: 63344478-63344479 on Build GRCh38
rs200534000 Chr.20: 63344058-63344058 on Build GRCh38
rs45537732 Chr.20: 63344321-63344321 on Build GRCh38
rs6090378 Chr.20: 63344026-63344026 on Build GRCh38
rs535104026 Chr.20: 63344105-63344105 on Build GRCh38
rs202178020 Chr.20: 63344224-63344224 on Build GRCh38
rs200499791 Chr.20: 63344216-63344216 on Build GRCh38
rs924042595 Chr.20: 63344115-63344115 on Build GRCh38
rs376422342 Chr.20: 63344420-63344420 on Build GRCh38
rs1051423829 Chr.20: 63344482-63344482 on Build GRCh38
rs772707836 Chr.20: 63344369-63344369 on Build GRCh38
rs201298613 Chr.20: 63344171-63344171 on Build GRCh38
rs535760681 Chr.20: 63344335-63344335 on Build GRCh38
rs201497655 Chr.20: 63344083-63344083 on Build GRCh38
rs774501677 Chr.20: 63344478-63344478 on Build GRCh38
rs202175713 Chr.20: 63344429-63344429 on Build GRCh38
rs768157010 Chr.20: 63344047-63344047 on Build GRCh38
rs747894739 Chr.20: 63344456-63344456 on Build GRCh38
rs199961012 Chr.20: 63344170-63344170 on Build GRCh38
rs199844434 Chr.20: 63344371-63344371 on Build GRCh38
rs528842597 Chr.20: 63344314-63344314 on Build GRCh38
rs45555537 Chr.20: 63344177-63344177 on Build GRCh38
rs202119950 Chr.20: 63344365-63344365 on Build GRCh38
rs557775256 Chr.20: 63344462-63344462 on Build GRCh38
rs200107825 Chr.20: 63344322-63344322 on Build GRCh38
rs201858019 Chr.20: 63344127-63344127 on Build GRCh38
rs181286096 Chr.20: 63344413-63344413 on Build GRCh38
rs199849183 Chr.20: 63344189-63344189 on Build GRCh38
rs1023570380 Chr.20: 63344074-63344074 on Build GRCh38
rs551959449 Chr.20: 63344494-63344495 on Build GRCh38
rs565780113 Chr.20: 63344336-63344336 on Build GRCh38
rs200852579 Chr.20: 63344381-63344381 on Build GRCh38
rs557180407 Chr.20: 63344214-63344214 on Build GRCh38
rs201157455 Chr.20: 63344249-63344249 on Build GRCh38
rs556065563 Chr.20: 63344256-63344256 on Build GRCh38
rs45617937 Chr.20: 63344412-63344412 on Build GRCh38
rs200660084 Chr.20: 63344308-63344308 on Build GRCh38
rs201095357 Chr.20: 63344268-63344268 on Build GRCh38
rs201615724 Chr.20: 63344039-63344039 on Build GRCh38
rs199833642 Chr.20: 63344100-63344100 on Build GRCh38
rs201294271 Chr.20: 63344357-63344357 on Build GRCh38
rs45494793 Chr.20: 63344272-63344272 on Build GRCh38
rs570983745 Chr.20: 63344045-63344045 on Build GRCh38
rs559194411 Chr.20: 63344044-63344044 on Build GRCh38
rs989987538 Chr.20: 63344311-63344311 on Build GRCh38
rs201255747 Chr.20: 63344421-63344421 on Build GRCh38
rs200751902 Chr.20: 63344501-63344501 on Build GRCh38
rs199666656 Chr.20: 63344283-63344283 on Build GRCh38
rs921562138 Chr.20: 63344237-63344237 on Build GRCh38

View Full Product Details