Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63350558-63351048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA4
Entrez Gene ID
1137
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4
UniGene
Hs.10734
Gene Chromosome Location
Chr.20: 63343310-63375392 on Build GRCh38
Transcripts
NM_000744.6 NM_001256573.1 NR_046317.1 XM_011528524.1 XM_017027624.1 XM_017027625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1483837 c.650C>G p.T217S Chr.20: 63350761-63350761 on Build GRCh38
5707511 c.622G>A p.E208K Chr.20: 63350789-63350789 on Build GRCh38
2931875 c.461delC p.P154fs*24 Chr.20: 63350950-63350950 on Build GRCh38
3548974 c.578A>G p.N193S Chr.20: 63350833-63350833 on Build GRCh38
1413084 c.681C>T p.A227A Chr.20: 63350730-63350730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61734382 Chr.20: 63350922-63350922 on Build GRCh38
rs200259564 Chr.20: 63350919-63350919 on Build GRCh38
rs776183357 Chr.20: 63350873-63350873 on Build GRCh38
rs201977125 Chr.20: 63351015-63351015 on Build GRCh38
rs772720363 Chr.20: 63350819-63350819 on Build GRCh38
rs121912254 Chr.20: 63350658-63350658 on Build GRCh38
rs755825569 Chr.20: 63350683-63350683 on Build GRCh38
rs758348186 Chr.20: 63350771-63350771 on Build GRCh38
rs754895704 Chr.20: 63350945-63350945 on Build GRCh38
rs777698611 Chr.20: 63350625-63350625 on Build GRCh38
rs761516841 Chr.20: 63350840-63350840 on Build GRCh38
rs121912255 Chr.20: 63350636-63350636 on Build GRCh38
rs770005515 Chr.20: 63350712-63350712 on Build GRCh38
rs121912251 Chr.20: 63350706-63350706 on Build GRCh38
rs1012796618 Chr.20: 63350759-63350759 on Build GRCh38
rs199735480 Chr.20: 63350939-63350939 on Build GRCh38
rs918550714 Chr.20: 63350768-63350768 on Build GRCh38
rs150082508 Chr.20: 63350627-63350627 on Build GRCh38
rs769287364 Chr.20: 63350997-63350997 on Build GRCh38
rs121912245 Chr.20: 63350949-63350949 on Build GRCh38
rs776204052 Chr.20: 63350887-63350887 on Build GRCh38
rs78695425 Chr.20: 63350692-63350692 on Build GRCh38
rs770780958 Chr.20: 63350633-63350633 on Build GRCh38
rs776092779 Chr.20: 63350820-63350820 on Build GRCh38
rs376632839 Chr.20: 63350814-63350814 on Build GRCh38
rs768343743 Chr.20: 63350884-63350884 on Build GRCh38
rs1044393 Chr.20: 63350772-63350772 on Build GRCh38
rs1048677024 Chr.20: 63350805-63350805 on Build GRCh38
rs763234634 Chr.20: 63350668-63350668 on Build GRCh38
rs759753426 Chr.20: 63350802-63350802 on Build GRCh38
rs201530542 Chr.20: 63350857-63350857 on Build GRCh38
rs796052317 Chr.20: 63350867-63350867 on Build GRCh38
rs200747769 Chr.20: 63350963-63350963 on Build GRCh38
rs981113016 Chr.20: 63350595-63350595 on Build GRCh38
rs1044394 Chr.20: 63350733-63350733 on Build GRCh38
rs576426437 Chr.20: 63350960-63350960 on Build GRCh38
rs756012418 Chr.20: 63350622-63350622 on Build GRCh38
rs750232429 Chr.20: 63350843-63350843 on Build GRCh38
rs138926102 Chr.20: 63350779-63350779 on Build GRCh38
rs201645533 Chr.20: 63350755-63350755 on Build GRCh38
rs752054605 Chr.20: 63350679-63350679 on Build GRCh38
rs985276958 Chr.20: 63350982-63350982 on Build GRCh38
rs121912252 Chr.20: 63350704-63350704 on Build GRCh38
rs776213268 Chr.20: 63350902-63350915 on Build GRCh38
rs757236120 Chr.20: 63350956-63350956 on Build GRCh38
rs147853603 Chr.20: 63350973-63350973 on Build GRCh38
rs554282916 Chr.20: 63350841-63350841 on Build GRCh38
rs796052322 Chr.20: 63351004-63351004 on Build GRCh38
rs373516964 Chr.20: 63350821-63350821 on Build GRCh38
rs199939003 Chr.20: 63350769-63350769 on Build GRCh38
rs201376529 Chr.20: 63351010-63351010 on Build GRCh38
rs745790876 Chr.20: 63350699-63350699 on Build GRCh38
rs121912256 Chr.20: 63350612-63350612 on Build GRCh38
rs941063587 Chr.20: 63350968-63350968 on Build GRCh38
rs199650427 Chr.20: 63350628-63350628 on Build GRCh38
rs758128052 Chr.20: 63350844-63350844 on Build GRCh38
rs149407396 Chr.20: 63350790-63350790 on Build GRCh38
rs749493840 Chr.20: 63350780-63350780 on Build GRCh38
rs144446511 Chr.20: 63350700-63350700 on Build GRCh38
rs201467277 Chr.20: 63351000-63351000 on Build GRCh38
rs766218638 Chr.20: 63351012-63351012 on Build GRCh38
rs112301452 Chr.20: 63350859-63350859 on Build GRCh38
rs773557088 Chr.20: 63350665-63350665 on Build GRCh38
rs121912248 Chr.20: 63350826-63350826 on Build GRCh38
rs758885593 Chr.20: 63350594-63350594 on Build GRCh38
rs138547414 Chr.20: 63350664-63350664 on Build GRCh38
rs121912246 Chr.20: 63350907-63350907 on Build GRCh38
rs538111736 Chr.20: 63350739-63350739 on Build GRCh38
rs112368488 Chr.20: 63350592-63350592 on Build GRCh38
rs780556996 Chr.20: 63350972-63350972 on Build GRCh38
rs553582391 Chr.20: 63350860-63350860 on Build GRCh38
rs200811782 Chr.20: 63350940-63350940 on Build GRCh38
rs121912249 Chr.20: 63350794-63350794 on Build GRCh38
rs200550248 Chr.20: 63350986-63350986 on Build GRCh38
rs776675247 Chr.20: 63350703-63350703 on Build GRCh38
rs774288629 Chr.20: 63350643-63350643 on Build GRCh38
rs769813725 Chr.20: 63350778-63350778 on Build GRCh38
rs61739212 Chr.20: 63350862-63350862 on Build GRCh38
rs777500590 Chr.20: 63350691-63350691 on Build GRCh38
rs772526356 Chr.20: 63350863-63350863 on Build GRCh38
rs79068736 Chr.20: 63350742-63350742 on Build GRCh38
rs762201442 Chr.20: 63350580-63350580 on Build GRCh38
rs144716263 Chr.20: 63350822-63350822 on Build GRCh38
rs121912243 Chr.20: 63350969-63350969 on Build GRCh38
rs201488442 Chr.20: 63351009-63351009 on Build GRCh38
rs762764855 Chr.20: 63350918-63350918 on Build GRCh38
rs757030850 Chr.20: 63350693-63350693 on Build GRCh38
rs778749351 Chr.20: 63350694-63350694 on Build GRCh38
rs201421555 Chr.20: 63350932-63350932 on Build GRCh38
rs747499031 Chr.20: 63350874-63350874 on Build GRCh38
rs757045864 Chr.20: 63350762-63350762 on Build GRCh38
rs796052316 Chr.20: 63350890-63350890 on Build GRCh38
rs762004535 Chr.20: 63350661-63350661 on Build GRCh38
rs759269817 Chr.20: 63351020-63351020 on Build GRCh38
rs121912244 Chr.20: 63350950-63350950 on Build GRCh38
rs200979226 Chr.20: 63350858-63350858 on Build GRCh38
rs910734867 Chr.20: 63350617-63350617 on Build GRCh38
rs780729214 Chr.20: 63350609-63350609 on Build GRCh38
rs763863668 Chr.20: 63350754-63350754 on Build GRCh38
rs374072103 Chr.20: 63350674-63350674 on Build GRCh38
rs200197645 Chr.20: 63350851-63350851 on Build GRCh38
rs201865234 Chr.20: 63350657-63350657 on Build GRCh38
rs748237979 Chr.20: 63350777-63350777 on Build GRCh38
rs746049095 Chr.20: 63350967-63350967 on Build GRCh38
rs796052321 Chr.20: 63351011-63351011 on Build GRCh38
rs200063611 Chr.20: 63350724-63350724 on Build GRCh38
rs779985852 Chr.20: 63350850-63350850 on Build GRCh38
rs759579810 Chr.20: 63350663-63350663 on Build GRCh38
rs121912253 Chr.20: 63350682-63350682 on Build GRCh38
rs201335931 Chr.20: 63350604-63350604 on Build GRCh38
rs112748821 Chr.20: 63350897-63350897 on Build GRCh38
rs374602963 Chr.20: 63350910-63350910 on Build GRCh38
rs372063405 Chr.20: 63350999-63350999 on Build GRCh38
rs572569890 Chr.20: 63350853-63350853 on Build GRCh38
rs121912247 Chr.20: 63350901-63350901 on Build GRCh38
rs796052323 Chr.20: 63350584-63350584 on Build GRCh38
rs778751703 Chr.20: 63350764-63350764 on Build GRCh38
rs199915426 Chr.20: 63350976-63350976 on Build GRCh38
rs375014053 Chr.20: 63351021-63351021 on Build GRCh38
rs200741008 Chr.20: 63350649-63350649 on Build GRCh38
rs763248360 Chr.20: 63350719-63350719 on Build GRCh38
rs750974344 Chr.20: 63350586-63350586 on Build GRCh38
rs199519224 Chr.20: 63350905-63350905 on Build GRCh38
rs747083728 Chr.20: 63350653-63350653 on Build GRCh38
rs199972913 Chr.20: 63350763-63350763 on Build GRCh38
rs2234932 Chr.20: 63350583-63350583 on Build GRCh38
rs776882201 Chr.20: 63350660-63350660 on Build GRCh38
rs765977956 Chr.20: 63350603-63350603 on Build GRCh38
rs748432058 Chr.20: 63350709-63350709 on Build GRCh38
rs1035741624 Chr.20: 63350597-63350597 on Build GRCh38
rs1012408774 Chr.20: 63350825-63350825 on Build GRCh38
rs747762347 Chr.20: 63350610-63350610 on Build GRCh38
rs121912250 Chr.20: 63350718-63350718 on Build GRCh38
rs771284636 Chr.20: 63350721-63350721 on Build GRCh38
rs141909279 Chr.20: 63350895-63350895 on Build GRCh38
rs76830527 Chr.20: 63350624-63350624 on Build GRCh38
rs45588436 Chr.20: 63350730-63350730 on Build GRCh38
rs771249249 Chr.20: 63350789-63350789 on Build GRCh38

View Full Product Details