Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63355611-63356097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA4
Entrez Gene ID
1137
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4
UniGene
Hs.10734
Gene Chromosome Location
Chr.20: 63343310-63375392 on Build GRCh38
Transcripts
NM_000744.6 NM_001256573.1 NR_046317.1 XM_011528524.1 XM_017027624.1 XM_017027625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3548977 c.274G>A p.E92K Chr.20: 63356084-63356084 on Build GRCh38
5584791 c.340C>T p.P114S Chr.20: 63356018-63356018 on Build GRCh38
5392587 c.300G>A p.W100* Chr.20: 63356058-63356058 on Build GRCh38
6628718 c.283G>A p.D95N Chr.20: 63356075-63356075 on Build GRCh38
2931880 c.369C>T p.I123I Chr.20: 63355989-63355989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035455129 Chr.20: 63355664-63355664 on Build GRCh38
rs199761642 Chr.20: 63355962-63355962 on Build GRCh38
rs572394984 Chr.20: 63355683-63355683 on Build GRCh38
rs145017594 Chr.20: 63356033-63356033 on Build GRCh38
rs1047922821 Chr.20: 63355833-63355833 on Build GRCh38
rs998054994 Chr.20: 63355888-63355888 on Build GRCh38
rs376139218 Chr.20: 63355770-63355770 on Build GRCh38
rs201166208 Chr.20: 63355947-63355947 on Build GRCh38
rs201160663 Chr.20: 63356063-63356063 on Build GRCh38
rs200010568 Chr.20: 63356000-63356000 on Build GRCh38
rs375654386 Chr.20: 63356024-63356024 on Build GRCh38
rs202055640 Chr.20: 63356066-63356066 on Build GRCh38
rs867890473 Chr.20: 63355948-63355948 on Build GRCh38
rs200801426 Chr.20: 63355939-63355939 on Build GRCh38
rs768495909 Chr.20: 63355954-63355954 on Build GRCh38
rs558874684 Chr.20: 63355942-63355942 on Build GRCh38
rs377114983 Chr.20: 63355996-63355996 on Build GRCh38
rs201386851 Chr.20: 63355986-63355986 on Build GRCh38
rs139129197 Chr.20: 63355974-63355974 on Build GRCh38
rs141864244 Chr.20: 63355870-63355870 on Build GRCh38
rs201033859 Chr.20: 63356013-63356013 on Build GRCh38
rs759593635 Chr.20: 63356053-63356053 on Build GRCh38
rs149055490 Chr.20: 63356043-63356043 on Build GRCh38
rs143103435 Chr.20: 63356062-63356062 on Build GRCh38
rs754312855 Chr.20: 63355999-63355999 on Build GRCh38
rs984096354 Chr.20: 63355918-63355918 on Build GRCh38
rs45462695 Chr.20: 63355696-63355696 on Build GRCh38
rs199507174 Chr.20: 63355935-63355935 on Build GRCh38
rs866334493 Chr.20: 63356010-63356010 on Build GRCh38
rs764421351 Chr.20: 63356068-63356068 on Build GRCh38
rs772347854 Chr.20: 63355907-63355907 on Build GRCh38
rs988855080 Chr.20: 63355686-63355686 on Build GRCh38
rs752089579 Chr.20: 63355942-63355943 on Build GRCh38
rs200690355 Chr.20: 63355968-63355968 on Build GRCh38
rs775147001 Chr.20: 63355950-63355950 on Build GRCh38
rs202042826 Chr.20: 63356047-63356047 on Build GRCh38
rs546771660 Chr.20: 63355943-63355960 on Build GRCh38
rs578019255 Chr.20: 63355639-63355639 on Build GRCh38
rs763234844 Chr.20: 63355938-63355938 on Build GRCh38
rs903454076 Chr.20: 63355697-63355697 on Build GRCh38
rs911475985 Chr.20: 63355757-63355757 on Build GRCh38
rs767647066 Chr.20: 63356055-63356055 on Build GRCh38
rs565024139 Chr.20: 63355828-63355828 on Build GRCh38
rs1041236621 Chr.20: 63355782-63355782 on Build GRCh38
rs149936966 Chr.20: 63355995-63355995 on Build GRCh38
rs759624178 Chr.20: 63355891-63355891 on Build GRCh38
rs1035528942 Chr.20: 63355957-63355957 on Build GRCh38
rs2273501 Chr.20: 63355777-63355777 on Build GRCh38
rs200639077 Chr.20: 63355827-63355827 on Build GRCh38
rs539072096 Chr.20: 63355657-63355657 on Build GRCh38
rs760455864 Chr.20: 63355952-63355952 on Build GRCh38
rs757345344 Chr.20: 63355959-63355959 on Build GRCh38
rs929074970 Chr.20: 63355820-63355820 on Build GRCh38
rs200741415 Chr.20: 63356031-63356031 on Build GRCh38
rs887475986 Chr.20: 63355864-63355864 on Build GRCh38
rs1053405214 Chr.20: 63355718-63355718 on Build GRCh38
rs543249857 Chr.20: 63355795-63355795 on Build GRCh38
rs897324943 Chr.20: 63355805-63355805 on Build GRCh38
rs532073622 Chr.20: 63355865-63355865 on Build GRCh38
rs768527293 Chr.20: 63356032-63356032 on Build GRCh38
rs770388417 Chr.20: 63355938-63355938 on Build GRCh38
rs200089220 Chr.20: 63356067-63356067 on Build GRCh38
rs781305602 Chr.20: 63355973-63355976 on Build GRCh38
rs985452934 Chr.20: 63355735-63355735 on Build GRCh38
rs554177070 Chr.20: 63355673-63355673 on Build GRCh38
rs905797402 Chr.20: 63355897-63355897 on Build GRCh38
rs771191068 Chr.20: 63356044-63356044 on Build GRCh38
rs767990721 Chr.20: 63355988-63355988 on Build GRCh38
rs771963046 Chr.20: 63355944-63355944 on Build GRCh38
rs955857261 Chr.20: 63355961-63355961 on Build GRCh38
rs763152627 Chr.20: 63355965-63355965 on Build GRCh38
rs754402670 Chr.20: 63356076-63356076 on Build GRCh38
rs779702890 Chr.20: 63356026-63356026 on Build GRCh38
rs761317422 Chr.20: 63356075-63356075 on Build GRCh38
rs1022602660 Chr.20: 63355874-63355874 on Build GRCh38
rs991964086 Chr.20: 63355955-63355958 on Build GRCh38
rs376069741 Chr.20: 63355684-63355689 on Build GRCh38
rs1018048819 Chr.20: 63355835-63355835 on Build GRCh38
rs561073250 Chr.20: 63355717-63355717 on Build GRCh38
rs745604520 Chr.20: 63356020-63356020 on Build GRCh38
rs774567134 Chr.20: 63356046-63356046 on Build GRCh38

View Full Product Details