Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63490043-63490574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1A2
Entrez Gene ID
1917
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Gene Aliases
EEF1AL, EF-1-alpha-2, EF1A, EIEE33, HS1, MRD38, STN, STNL
UniGene
Hs.433839
Gene Chromosome Location
Chr.20: 63488012-63499315 on Build GRCh38
Transcripts
NM_001958.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4773301 c.997C>A p.P333T Chr.20: 63490511-63490511 on Build GRCh38
4100401 c.962G>A p.R321Q Chr.20: 63490546-63490546 on Build GRCh38
6041452 c.1001C>T p.P334L Chr.20: 63490507-63490507 on Build GRCh38
1615863 c.1012G>T p.A338S Chr.20: 63490496-63490496 on Build GRCh38
1250950 c.999G>A p.P333P Chr.20: 63490509-63490509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940701157 Chr.20: 63490081-63490081 on Build GRCh38
rs1036345223 Chr.20: 63490227-63490227 on Build GRCh38
rs1014889787 Chr.20: 63490121-63490121 on Build GRCh38
rs917960235 Chr.20: 63490468-63490468 on Build GRCh38
rs796558317 Chr.20: 63490080-63490080 on Build GRCh38
rs992932736 Chr.20: 63490061-63490061 on Build GRCh38
rs1054278909 Chr.20: 63490551-63490551 on Build GRCh38
rs761090978 Chr.20: 63490403-63490403 on Build GRCh38
rs962995958 Chr.20: 63490213-63490213 on Build GRCh38
rs988387826 Chr.20: 63490221-63490221 on Build GRCh38
rs926645906 Chr.20: 63490234-63490234 on Build GRCh38
rs945481319 Chr.20: 63490475-63490475 on Build GRCh38
rs111392970 Chr.20: 63490476-63490476 on Build GRCh38
rs752443173 Chr.20: 63490470-63490471 on Build GRCh38
rs574384755 Chr.20: 63490507-63490507 on Build GRCh38
rs996488810 Chr.20: 63490357-63490357 on Build GRCh38
rs61414752 Chr.20: 63490186-63490186 on Build GRCh38
rs935904739 Chr.20: 63490550-63490550 on Build GRCh38
rs310607 Chr.20: 63490349-63490349 on Build GRCh38
rs370600415 Chr.20: 63490446-63490446 on Build GRCh38
rs549958091 Chr.20: 63490178-63490178 on Build GRCh38
rs57130528 Chr.20: 63490068-63490068 on Build GRCh38
rs577079221 Chr.20: 63490447-63490447 on Build GRCh38
rs529629975 Chr.20: 63490115-63490115 on Build GRCh38
rs571434862 Chr.20: 63490188-63490188 on Build GRCh38
rs779722584 Chr.20: 63490452-63490452 on Build GRCh38
rs1045520862 Chr.20: 63490396-63490396 on Build GRCh38
rs944637496 Chr.20: 63490316-63490316 on Build GRCh38
rs778258442 Chr.20: 63490448-63490449 on Build GRCh38
rs202227388 Chr.20: 63490513-63490513 on Build GRCh38
rs561382298 Chr.20: 63490149-63490149 on Build GRCh38
rs945798921 Chr.20: 63490072-63490072 on Build GRCh38
rs771937333 Chr.20: 63490153-63490153 on Build GRCh38
rs143644219 Chr.20: 63490090-63490090 on Build GRCh38
rs372861286 Chr.20: 63490297-63490297 on Build GRCh38
rs75499147 Chr.20: 63490069-63490069 on Build GRCh38
rs994883567 Chr.20: 63490136-63490136 on Build GRCh38
rs903113040 Chr.20: 63490108-63490108 on Build GRCh38
rs541540087 Chr.20: 63490098-63490098 on Build GRCh38
rs760721599 Chr.20: 63490509-63490509 on Build GRCh38
rs310608 Chr.20: 63490373-63490373 on Build GRCh38
rs559317496 Chr.20: 63490114-63490114 on Build GRCh38
rs1033660535 Chr.20: 63490449-63490449 on Build GRCh38
rs754338238 Chr.20: 63490516-63490516 on Build GRCh38
rs779441972 Chr.20: 63490546-63490546 on Build GRCh38
rs374272155 Chr.20: 63490466-63490466 on Build GRCh38
rs141887309 Chr.20: 63490312-63490312 on Build GRCh38
rs544115703 Chr.20: 63490345-63490345 on Build GRCh38
rs1032641434 Chr.20: 63490129-63490129 on Build GRCh38
rs890390020 Chr.20: 63490104-63490104 on Build GRCh38
rs192151392 Chr.20: 63490128-63490128 on Build GRCh38
rs931844369 Chr.20: 63490091-63490091 on Build GRCh38
rs1036196302 Chr.20: 63490344-63490344 on Build GRCh38
rs370457251 Chr.20: 63490473-63490473 on Build GRCh38
rs1027660779 Chr.20: 63490137-63490137 on Build GRCh38
rs912873467 Chr.20: 63490064-63490067 on Build GRCh38
rs558771513 Chr.20: 63490374-63490374 on Build GRCh38
rs1050272353 Chr.20: 63490101-63490101 on Build GRCh38
rs759770756 Chr.20: 63490506-63490506 on Build GRCh38
rs903057706 Chr.20: 63490317-63490317 on Build GRCh38
rs748781344 Chr.20: 63490335-63490335 on Build GRCh38
rs912906670 Chr.20: 63490239-63490239 on Build GRCh38
rs370482820 Chr.20: 63490430-63490430 on Build GRCh38
rs373731830 Chr.20: 63490536-63490536 on Build GRCh38
rs748110210 Chr.20: 63490384-63490386 on Build GRCh38
rs955706679 Chr.20: 63490170-63490170 on Build GRCh38
rs938063565 Chr.20: 63490255-63490255 on Build GRCh38
rs925733655 Chr.20: 63490256-63490256 on Build GRCh38
rs531827185 Chr.20: 63490161-63490161 on Build GRCh38
rs45510898 Chr.20: 63490467-63490467 on Build GRCh38
rs974068538 Chr.20: 63490223-63490223 on Build GRCh38
rs576256590 Chr.20: 63490359-63490359 on Build GRCh38
rs935957038 Chr.20: 63490330-63490330 on Build GRCh38
rs949894300 Chr.20: 63490308-63490308 on Build GRCh38
rs1004750167 Chr.20: 63490388-63490388 on Build GRCh38
rs918026662 Chr.20: 63490301-63490301 on Build GRCh38
rs1042789804 Chr.20: 63490073-63490073 on Build GRCh38
rs899533646 Chr.20: 63490350-63490350 on Build GRCh38
rs547450062 Chr.20: 63490224-63490224 on Build GRCh38
rs945661768 Chr.20: 63490252-63490252 on Build GRCh38
rs981385928 Chr.20: 63490195-63490195 on Build GRCh38
rs200662984 Chr.20: 63490500-63490500 on Build GRCh38
rs373337082 Chr.20: 63490433-63490433 on Build GRCh38
rs376449261 Chr.20: 63490444-63490444 on Build GRCh38
rs754214863 Chr.20: 63490431-63490431 on Build GRCh38
rs555114811 Chr.20: 63490193-63490193 on Build GRCh38
rs538729380 Chr.20: 63490214-63490214 on Build GRCh38
rs1008799927 Chr.20: 63490109-63490109 on Build GRCh38
rs963282819 Chr.20: 63490412-63490412 on Build GRCh38
rs1035181601 Chr.20: 63490207-63490207 on Build GRCh38
rs536203214 Chr.20: 63490347-63490347 on Build GRCh38
rs575077061 Chr.20: 63490204-63490204 on Build GRCh38
rs996582213 Chr.20: 63490152-63490152 on Build GRCh38
rs550785848 Chr.20: 63490352-63490352 on Build GRCh38
rs866984659 Chr.20: 63490526-63490526 on Build GRCh38

View Full Product Details