Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63706271-63706767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
UniGene
Hs.389277
Gene Chromosome Location
Chr.20: 63698642-63708013 on Build GRCh38
Transcripts
NM_001134758.3 NM_001267544.2 NM_001267545.2 NM_001267546.2 NM_001267547.2 NM_001267548.2 NM_001267549.2 NM_003224.5 NR_051954.2 NR_051955.2 NR_051956.2 NR_051957.2 NR_051958.2 XM_011528482.2 XM_011528483.1 XM_017027576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725118 c.182-2A>T p.? Chr.20: 63706441-63706441 on Build GRCh38
7341113 c.230G>T p.G77V Chr.20: 63706391-63706391 on Build GRCh38
1029170 c.253T>C p.L85L Chr.20: 63706368-63706368 on Build GRCh38
478398 c.215T>A p.M72K Chr.20: 63706406-63706406 on Build GRCh38
1029171 c.110C>T p.S37L Chr.20: 63706722-63706722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543502182 Chr.20: 63706450-63706450 on Build GRCh38
rs746051037 Chr.20: 63706698-63706698 on Build GRCh38
rs368887277 Chr.20: 63706624-63706624 on Build GRCh38
rs754237422 Chr.20: 63706473-63706473 on Build GRCh38
rs879127624 Chr.20: 63706426-63706426 on Build GRCh38
rs761022856 Chr.20: 63706631-63706631 on Build GRCh38
rs145809158 Chr.20: 63706587-63706587 on Build GRCh38
rs141232776 Chr.20: 63706665-63706665 on Build GRCh38
rs774751038 Chr.20: 63706610-63706610 on Build GRCh38
rs751696324 Chr.20: 63706384-63706384 on Build GRCh38
rs749931171 Chr.20: 63706340-63706340 on Build GRCh38
rs771999150 Chr.20: 63706315-63706315 on Build GRCh38
rs764838441 Chr.20: 63706661-63706661 on Build GRCh38
rs751025946 Chr.20: 63706745-63706745 on Build GRCh38
rs749662460 Chr.20: 63706447-63706447 on Build GRCh38
rs373946175 Chr.20: 63706453-63706453 on Build GRCh38
rs775674404 Chr.20: 63706630-63706630 on Build GRCh38
rs779337271 Chr.20: 63706562-63706562 on Build GRCh38
rs996729557 Chr.20: 63706578-63706578 on Build GRCh38
rs528376535 Chr.20: 63706721-63706721 on Build GRCh38
rs368456471 Chr.20: 63706687-63706687 on Build GRCh38
rs546042210 Chr.20: 63706594-63706594 on Build GRCh38
rs772667823 Chr.20: 63706312-63706312 on Build GRCh38
rs758076846 Chr.20: 63706669-63706669 on Build GRCh38
rs764328829 Chr.20: 63706471-63706471 on Build GRCh38
rs955202756 Chr.20: 63706605-63706605 on Build GRCh38
rs772347254 Chr.20: 63706703-63706703 on Build GRCh38
rs756846463 Chr.20: 63706474-63706474 on Build GRCh38
rs777737516 Chr.20: 63706696-63706696 on Build GRCh38
rs757943900 Chr.20: 63706483-63706483 on Build GRCh38
rs771270734 Chr.20: 63706609-63706609 on Build GRCh38
rs765545157 Chr.20: 63706733-63706733 on Build GRCh38
rs546482299 Chr.20: 63706551-63706552 on Build GRCh38
rs778595611 Chr.20: 63706476-63706476 on Build GRCh38
rs370321126 Chr.20: 63706603-63706603 on Build GRCh38
rs34843664 Chr.20: 63706715-63706715 on Build GRCh38
rs201367545 Chr.20: 63706437-63706437 on Build GRCh38
rs376804407 Chr.20: 63706339-63706339 on Build GRCh38
rs749086905 Chr.20: 63706700-63706700 on Build GRCh38
rs375948751 Chr.20: 63706619-63706619 on Build GRCh38
rs148848505 Chr.20: 63706424-63706424 on Build GRCh38
rs748030926 Chr.20: 63706606-63706606 on Build GRCh38
rs762232409 Chr.20: 63706640-63706640 on Build GRCh38
rs774556927 Chr.20: 63706463-63706463 on Build GRCh38
rs766047766 Chr.20: 63706551-63706551 on Build GRCh38
rs753196918 Chr.20: 63706469-63706469 on Build GRCh38
rs896773594 Chr.20: 63706446-63706446 on Build GRCh38
rs370708627 Chr.20: 63706634-63706634 on Build GRCh38
rs112794876 Chr.20: 63706536-63706536 on Build GRCh38
rs371997885 Chr.20: 63706633-63706633 on Build GRCh38
rs756480274 Chr.20: 63706407-63706407 on Build GRCh38
rs145589861 Chr.20: 63706402-63706402 on Build GRCh38
rs768102583 Chr.20: 63706468-63706468 on Build GRCh38
rs762534440 Chr.20: 63706741-63706741 on Build GRCh38
rs80322094 Chr.20: 63706743-63706743 on Build GRCh38
rs766719308 Chr.20: 63706459-63706459 on Build GRCh38
rs771982698 Chr.20: 63706445-63706445 on Build GRCh38
rs548213874 Chr.20: 63706727-63706727 on Build GRCh38
rs149028886 Chr.20: 63706567-63706567 on Build GRCh38
rs749932216 Chr.20: 63706477-63706477 on Build GRCh38
rs937756948 Chr.20: 63706672-63706672 on Build GRCh38
rs760444676 Chr.20: 63706334-63706334 on Build GRCh38
rs747917536 Chr.20: 63706691-63706691 on Build GRCh38
rs573225049 Chr.20: 63706293-63706293 on Build GRCh38
rs375086901 Chr.20: 63706638-63706638 on Build GRCh38
rs754855495 Chr.20: 63706684-63706684 on Build GRCh38
rs779362957 Chr.20: 63706670-63706670 on Build GRCh38
rs759753683 Chr.20: 63706465-63706465 on Build GRCh38
rs767032612 Chr.20: 63706741-63706743 on Build GRCh38
rs750028516 Chr.20: 63706664-63706664 on Build GRCh38
rs574377812 Chr.20: 63706427-63706427 on Build GRCh38
rs1015497877 Chr.20: 63706620-63706620 on Build GRCh38
rs370693784 Chr.20: 63706438-63706438 on Build GRCh38
rs746798202 Chr.20: 63706449-63706449 on Build GRCh38
rs775471873 Chr.20: 63706448-63706448 on Build GRCh38
rs369464988 Chr.20: 63706320-63706320 on Build GRCh38
rs766528995 Chr.20: 63706349-63706349 on Build GRCh38
rs755488304 Chr.20: 63706386-63706386 on Build GRCh38
rs762144078 Chr.20: 63706732-63706732 on Build GRCh38
rs900585654 Chr.20: 63706568-63706568 on Build GRCh38
rs926344492 Chr.20: 63706294-63706294 on Build GRCh38
rs773355649 Chr.20: 63706346-63706346 on Build GRCh38
rs181865540 Chr.20: 63706679-63706679 on Build GRCh38
rs553206884 Chr.20: 63706291-63706291 on Build GRCh38
rs377397111 Chr.20: 63706484-63706484 on Build GRCh38
rs763260169 Chr.20: 63706348-63706348 on Build GRCh38
rs554534156 Chr.20: 63706418-63706418 on Build GRCh38
rs11547196 Chr.20: 63706399-63706399 on Build GRCh38
rs73319723 Chr.20: 63706332-63706332 on Build GRCh38
rs745393886 Chr.20: 63706443-63706443 on Build GRCh38
rs756805303 Chr.20: 63706723-63706763 on Build GRCh38
rs780790599 Chr.20: 63706604-63706604 on Build GRCh38
rs777456674 Chr.20: 63706413-63706413 on Build GRCh38
rs757503056 Chr.20: 63706517-63706517 on Build GRCh38

View Full Product Details