Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63706895-63707391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
UniGene
Hs.389277
Gene Chromosome Location
Chr.20: 63698642-63708013 on Build GRCh38
Transcripts
NM_001134758.3 NM_001267544.2 NM_001267545.2 NM_001267546.2 NM_001267547.2 NM_001267548.2 NM_001267549.2 NM_003224.5 NR_051954.2 NR_051955.2 NR_051956.2 NR_051957.2 NR_051958.2 XM_011528482.2 XM_011528483.1 XM_017027576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390014 c.60C>T p.I20I Chr.20: 63707032-63707032 on Build GRCh38
5780052 c.5A>G p.Y2C Chr.20: 63707087-63707087 on Build GRCh38
5858351 c.31T>C p.Y11H Chr.20: 63707061-63707061 on Build GRCh38
4923105 c.28A>G p.K10E Chr.20: 63707064-63707064 on Build GRCh38
1307639 c.10C>G p.L4V Chr.20: 63707082-63707082 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756061238 Chr.20: 63706976-63706976 on Build GRCh38
rs771274335 Chr.20: 63707059-63707059 on Build GRCh38
rs759166110 Chr.20: 63707084-63707084 on Build GRCh38
rs747205794 Chr.20: 63707013-63707013 on Build GRCh38
rs559563921 Chr.20: 63707194-63707194 on Build GRCh38
rs910555874 Chr.20: 63706939-63706939 on Build GRCh38
rs774605679 Chr.20: 63707064-63707064 on Build GRCh38
rs907991952 Chr.20: 63707256-63707256 on Build GRCh38
rs971904930 Chr.20: 63707244-63707244 on Build GRCh38
rs557064908 Chr.20: 63706932-63706932 on Build GRCh38
rs867230800 Chr.20: 63707175-63707175 on Build GRCh38
rs933313969 Chr.20: 63706934-63706934 on Build GRCh38
rs755212995 Chr.20: 63707122-63707122 on Build GRCh38
rs942232050 Chr.20: 63707363-63707363 on Build GRCh38
rs931114485 Chr.20: 63707307-63707307 on Build GRCh38
rs886514585 Chr.20: 63707023-63707023 on Build GRCh38
rs775797096 Chr.20: 63707336-63707336 on Build GRCh38
rs925461776 Chr.20: 63707162-63707162 on Build GRCh38
rs940758493 Chr.20: 63707271-63707271 on Build GRCh38
rs1052307288 Chr.20: 63706956-63706956 on Build GRCh38
rs752739881 Chr.20: 63707131-63707131 on Build GRCh38
rs749758912 Chr.20: 63707143-63707143 on Build GRCh38
rs781459401 Chr.20: 63707127-63707127 on Build GRCh38
rs189261646 Chr.20: 63707232-63707232 on Build GRCh38
rs895055091 Chr.20: 63707328-63707328 on Build GRCh38
rs923303563 Chr.20: 63707330-63707330 on Build GRCh38
rs143388894 Chr.20: 63707029-63707029 on Build GRCh38
rs146966742 Chr.20: 63707025-63707025 on Build GRCh38
rs764092865 Chr.20: 63707111-63707111 on Build GRCh38
rs543053254 Chr.20: 63706930-63706930 on Build GRCh38
rs1033572030 Chr.20: 63707093-63707093 on Build GRCh38
rs753683308 Chr.20: 63706985-63706985 on Build GRCh38
rs922190810 Chr.20: 63707230-63707230 on Build GRCh38
rs1037756590 Chr.20: 63707278-63707278 on Build GRCh38
rs181914983 Chr.20: 63707352-63707352 on Build GRCh38
rs756233430 Chr.20: 63707135-63707135 on Build GRCh38
rs773746876 Chr.20: 63707046-63707046 on Build GRCh38
rs1040617964 Chr.20: 63706979-63706979 on Build GRCh38
rs561842508 Chr.20: 63707353-63707353 on Build GRCh38
rs749590691 Chr.20: 63707058-63707058 on Build GRCh38
rs910670461 Chr.20: 63707294-63707294 on Build GRCh38
rs760331612 Chr.20: 63707110-63707110 on Build GRCh38
rs752263927 Chr.20: 63706975-63706975 on Build GRCh38
rs771869403 Chr.20: 63707105-63707105 on Build GRCh38
rs748284689 Chr.20: 63707028-63707028 on Build GRCh38
rs777848469 Chr.20: 63707141-63707141 on Build GRCh38
rs1051765858 Chr.20: 63707246-63707246 on Build GRCh38
rs1055075516 Chr.20: 63707321-63707321 on Build GRCh38
rs137883585 Chr.20: 63707026-63707026 on Build GRCh38
rs977374565 Chr.20: 63707204-63707204 on Build GRCh38
rs990228527 Chr.20: 63707280-63707280 on Build GRCh38
rs774098901 Chr.20: 63706957-63706957 on Build GRCh38
rs371346755 Chr.20: 63706973-63706973 on Build GRCh38
rs765015965 Chr.20: 63707121-63707121 on Build GRCh38
rs755724981 Chr.20: 63707058-63707063 on Build GRCh38
rs957232407 Chr.20: 63707261-63707261 on Build GRCh38
rs936924646 Chr.20: 63706921-63706921 on Build GRCh38
rs367672959 Chr.20: 63706928-63706928 on Build GRCh38
rs553612498 Chr.20: 63706948-63706948 on Build GRCh38
rs749909286 Chr.20: 63706966-63706966 on Build GRCh38
rs958169644 Chr.20: 63707172-63707172 on Build GRCh38
rs902117986 Chr.20: 63706991-63706991 on Build GRCh38
rs6011041 Chr.20: 63707200-63707200 on Build GRCh38
rs943366503 Chr.20: 63707303-63707303 on Build GRCh38
rs143886078 Chr.20: 63707176-63707177 on Build GRCh38
rs971693524 Chr.20: 63707196-63707196 on Build GRCh38
rs995431347 Chr.20: 63707077-63707077 on Build GRCh38
rs764186946 Chr.20: 63706983-63706983 on Build GRCh38
rs753875890 Chr.20: 63707114-63707114 on Build GRCh38
rs761678778 Chr.20: 63707116-63707116 on Build GRCh38
rs759129647 Chr.20: 63706967-63706967 on Build GRCh38
rs780117354 Chr.20: 63707010-63707010 on Build GRCh38
rs775129483 Chr.20: 63707108-63707108 on Build GRCh38
rs999243318 Chr.20: 63707357-63707357 on Build GRCh38
rs898346001 Chr.20: 63707291-63707291 on Build GRCh38
rs111259883 Chr.20: 63707086-63707086 on Build GRCh38
rs757117033 Chr.20: 63706987-63706987 on Build GRCh38

View Full Product Details