Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63894938-63895432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5
Entrez Gene ID
80331
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
Gene Aliases
CLN4, CLN4B, CSP, DNAJC5A, NCL, mir-941-2, mir-941-3, mir-941-4, mir-941-5
UniGene
Hs.164419
Gene Chromosome Location
Chr.20: 63895102-63936031 on Build GRCh38
Transcripts
NM_025219.2

SNP Detail

SNP ID Chromosome Location
rs886056928 Chr.20: 63895150-63895150 on Build GRCh38
rs1001499013 Chr.20: 63895184-63895184 on Build GRCh38
rs926467122 Chr.20: 63895263-63895263 on Build GRCh38
rs945377973 Chr.20: 63895173-63895173 on Build GRCh38
rs537670690 Chr.20: 63895124-63895124 on Build GRCh38
rs917952143 Chr.20: 63895169-63895170 on Build GRCh38
rs904060246 Chr.20: 63895034-63895034 on Build GRCh38
rs993661788 Chr.20: 63894969-63894969 on Build GRCh38
rs531246320 Chr.20: 63895194-63895195 on Build GRCh38
rs577550567 Chr.20: 63895156-63895156 on Build GRCh38
rs1018701451 Chr.20: 63895202-63895202 on Build GRCh38
rs904094797 Chr.20: 63895178-63895178 on Build GRCh38
rs917983556 Chr.20: 63895298-63895301 on Build GRCh38
rs965724653 Chr.20: 63895203-63895203 on Build GRCh38
rs1001025435 Chr.20: 63895045-63895045 on Build GRCh38
rs886056932 Chr.20: 63895240-63895240 on Build GRCh38
rs540212627 Chr.20: 63895167-63895167 on Build GRCh38
rs886056929 Chr.20: 63895180-63895181 on Build GRCh38
rs1007348593 Chr.20: 63895196-63895196 on Build GRCh38
rs981723471 Chr.20: 63895396-63895396 on Build GRCh38
rs965692513 Chr.20: 63895128-63895128 on Build GRCh38
rs750302498 Chr.20: 63895226-63895226 on Build GRCh38
rs993165610 Chr.20: 63895205-63895205 on Build GRCh38
rs539422683 Chr.20: 63894999-63894999 on Build GRCh38
rs925233366 Chr.20: 63895375-63895375 on Build GRCh38
rs889921666 Chr.20: 63895195-63895206 on Build GRCh38
rs886056927 Chr.20: 63895149-63895149 on Build GRCh38
rs6089779 Chr.20: 63894981-63894981 on Build GRCh38
rs1033207180 Chr.20: 63895053-63895053 on Build GRCh38
rs915929426 Chr.20: 63895093-63895093 on Build GRCh38
rs972428929 Chr.20: 63895133-63895133 on Build GRCh38
rs960598331 Chr.20: 63895022-63895022 on Build GRCh38
rs115636427 Chr.20: 63895020-63895020 on Build GRCh38
rs953124172 Chr.20: 63895080-63895080 on Build GRCh38
rs553030156 Chr.20: 63895005-63895005 on Build GRCh38
rs563818595 Chr.20: 63895147-63895148 on Build GRCh38
rs1023402472 Chr.20: 63895002-63895002 on Build GRCh38
rs992243199 Chr.20: 63895289-63895289 on Build GRCh38
rs946469707 Chr.20: 63895072-63895072 on Build GRCh38
rs941287791 Chr.20: 63895157-63895158 on Build GRCh38
rs924733226 Chr.20: 63895403-63895403 on Build GRCh38
rs886056931 Chr.20: 63895215-63895216 on Build GRCh38
rs566439711 Chr.20: 63895016-63895016 on Build GRCh38
rs192730887 Chr.20: 63895048-63895048 on Build GRCh38
rs886056926 Chr.20: 63895174-63895175 on Build GRCh38
rs909535456 Chr.20: 63895148-63895165 on Build GRCh38
rs1049925548 Chr.20: 63895195-63895200 on Build GRCh38
rs919161449 Chr.20: 63895148-63895153 on Build GRCh38
rs1056440806 Chr.20: 63895168-63895168 on Build GRCh38
rs575349152 Chr.20: 63895092-63895092 on Build GRCh38
rs959192520 Chr.20: 63895276-63895276 on Build GRCh38
rs926425639 Chr.20: 63895149-63895149 on Build GRCh38
rs913709370 Chr.20: 63895068-63895068 on Build GRCh38
rs987994819 Chr.20: 63895028-63895028 on Build GRCh38
rs919117473 Chr.20: 63894966-63894966 on Build GRCh38
rs185056589 Chr.20: 63895366-63895366 on Build GRCh38
rs936945280 Chr.20: 63895397-63895397 on Build GRCh38
rs1026429541 Chr.20: 63894977-63894977 on Build GRCh38
rs952015679 Chr.20: 63895238-63895238 on Build GRCh38
rs985074150 Chr.20: 63895256-63895256 on Build GRCh38
rs927907039 Chr.20: 63895021-63895021 on Build GRCh38
rs952000255 Chr.20: 63895148-63895159 on Build GRCh38
rs886056933 Chr.20: 63895246-63895246 on Build GRCh38
rs1056408784 Chr.20: 63895008-63895008 on Build GRCh38
rs1043057997 Chr.20: 63895176-63895176 on Build GRCh38
rs971939360 Chr.20: 63894956-63894956 on Build GRCh38
rs557596205 Chr.20: 63895127-63895127 on Build GRCh38
rs886056930 Chr.20: 63895175-63895175 on Build GRCh38
rs984870227 Chr.20: 63894985-63894985 on Build GRCh38
rs886056934 Chr.20: 63895302-63895302 on Build GRCh38
rs886056925 Chr.20: 63895141-63895141 on Build GRCh38
rs560162132 Chr.20: 63895343-63895343 on Build GRCh38
rs926714989 Chr.20: 63895003-63895003 on Build GRCh38

View Full Product Details