Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63929404-63929911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5
Entrez Gene ID
80331
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
Gene Aliases
CLN4, CLN4B, CSP, DNAJC5A, NCL, mir-941-2, mir-941-3, mir-941-4, mir-941-5
UniGene
Hs.164419
Gene Chromosome Location
Chr.20: 63895102-63936031 on Build GRCh38
Transcripts
NM_025219.2 XM_011529048.2 XM_011529049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516884 c.237C>T p.Y79Y Chr.20: 63929441-63929441 on Build GRCh38
5516883 c.237C>T p.Y79Y Chr.20: 63929441-63929441 on Build GRCh38
1204215 c.208G>A p.A70T Chr.20: 63929412-63929412 on Build GRCh38
5000895 c.282C>A p.N94K Chr.20: 63929486-63929486 on Build GRCh38
6647362 c.226T>C p.Y76H Chr.20: 63929430-63929430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367766641 Chr.20: 63929539-63929539 on Build GRCh38
rs777619967 Chr.20: 63929553-63929553 on Build GRCh38
rs35311419 Chr.20: 63929684-63929684 on Build GRCh38
rs759202388 Chr.20: 63929546-63929546 on Build GRCh38
rs749077830 Chr.20: 63929459-63929459 on Build GRCh38
rs762638579 Chr.20: 63929433-63929433 on Build GRCh38
rs536802709 Chr.20: 63929606-63929606 on Build GRCh38
rs74337839 Chr.20: 63929765-63929765 on Build GRCh38
rs777583663 Chr.20: 63929454-63929454 on Build GRCh38
rs563480286 Chr.20: 63929766-63929766 on Build GRCh38
rs201495666 Chr.20: 63929432-63929432 on Build GRCh38
rs763367010 Chr.20: 63929534-63929534 on Build GRCh38
rs376163421 Chr.20: 63929429-63929429 on Build GRCh38
rs930452906 Chr.20: 63929750-63929750 on Build GRCh38
rs570181415 Chr.20: 63929812-63929812 on Build GRCh38
rs370198236 Chr.20: 63929757-63929757 on Build GRCh38
rs934029432 Chr.20: 63929680-63929680 on Build GRCh38
rs773593782 Chr.20: 63929818-63929818 on Build GRCh38
rs749801865 Chr.20: 63929572-63929572 on Build GRCh38
rs33998408 Chr.20: 63929667-63929667 on Build GRCh38
rs113080441 Chr.20: 63929598-63929598 on Build GRCh38
rs138086896 Chr.20: 63929731-63929731 on Build GRCh38
rs112959403 Chr.20: 63929736-63929736 on Build GRCh38
rs953203419 Chr.20: 63929536-63929536 on Build GRCh38
rs940576822 Chr.20: 63929706-63929706 on Build GRCh38
rs1047502569 Chr.20: 63929702-63929702 on Build GRCh38
rs189188563 Chr.20: 63929679-63929679 on Build GRCh38
rs756026221 Chr.20: 63929453-63929453 on Build GRCh38
rs752178736 Chr.20: 63929549-63929549 on Build GRCh38
rs879788909 Chr.20: 63929766-63929767 on Build GRCh38
rs926905239 Chr.20: 63929636-63929636 on Build GRCh38
rs565725554 Chr.20: 63929557-63929557 on Build GRCh38
rs111838668 Chr.20: 63929889-63929889 on Build GRCh38
rs1021150330 Chr.20: 63929877-63929877 on Build GRCh38
rs768786863 Chr.20: 63929495-63929495 on Build GRCh38
rs964479182 Chr.20: 63929633-63929633 on Build GRCh38
rs781303835 Chr.20: 63929565-63929565 on Build GRCh38
rs560879349 Chr.20: 63929751-63929751 on Build GRCh38
rs553743982 Chr.20: 63929689-63929689 on Build GRCh38
rs35630509 Chr.20: 63929889-63929890 on Build GRCh38
rs972372661 Chr.20: 63929687-63929687 on Build GRCh38
rs150357637 Chr.20: 63929719-63929719 on Build GRCh38
rs576774539 Chr.20: 63929641-63929641 on Build GRCh38
rs75457861 Chr.20: 63929811-63929811 on Build GRCh38
rs536758837 Chr.20: 63929754-63929754 on Build GRCh38
rs975493683 Chr.20: 63929634-63929634 on Build GRCh38
rs1056482390 Chr.20: 63929814-63929814 on Build GRCh38
rs572067675 Chr.20: 63929701-63929701 on Build GRCh38
rs964273440 Chr.20: 63929774-63929774 on Build GRCh38
rs2295436 Chr.20: 63929545-63929545 on Build GRCh38
rs187798588 Chr.20: 63929728-63929728 on Build GRCh38
rs770247745 Chr.20: 63929510-63929510 on Build GRCh38
rs560607645 Chr.20: 63929676-63929676 on Build GRCh38
rs145704049 Chr.20: 63929685-63929686 on Build GRCh38
rs752620792 Chr.20: 63929449-63929449 on Build GRCh38
rs762344309 Chr.20: 63929502-63929502 on Build GRCh38
rs372259352 Chr.20: 63929571-63929571 on Build GRCh38
rs2295439 Chr.20: 63929739-63929739 on Build GRCh38
rs755284297 Chr.20: 63929564-63929564 on Build GRCh38
rs982753636 Chr.20: 63929874-63929874 on Build GRCh38
rs190651531 Chr.20: 63929693-63929693 on Build GRCh38
rs559173027 Chr.20: 63929875-63929875 on Build GRCh38
rs749945571 Chr.20: 63929684-63929685 on Build GRCh38
rs780477656 Chr.20: 63929468-63929468 on Build GRCh38
rs753649103 Chr.20: 63929555-63929555 on Build GRCh38
rs534739204 Chr.20: 63929563-63929563 on Build GRCh38
rs940629101 Chr.20: 63929890-63929890 on Build GRCh38
rs776598152 Chr.20: 63929501-63929501 on Build GRCh38
rs556799650 Chr.20: 63929632-63929632 on Build GRCh38
rs532423762 Chr.20: 63929705-63929705 on Build GRCh38
rs762228571 Chr.20: 63929426-63929426 on Build GRCh38
rs558502866 Chr.20: 63929653-63929653 on Build GRCh38
rs149971662 Chr.20: 63929441-63929441 on Build GRCh38
rs557501238 Chr.20: 63929628-63929628 on Build GRCh38
rs771228555 Chr.20: 63929575-63929575 on Build GRCh38
rs933246467 Chr.20: 63929446-63929446 on Build GRCh38
rs766072943 Chr.20: 63929435-63929435 on Build GRCh38
rs1037497030 Chr.20: 63929727-63929727 on Build GRCh38
rs955633373 Chr.20: 63929851-63929851 on Build GRCh38
rs532410241 Chr.20: 63929447-63929447 on Build GRCh38
rs772397861 Chr.20: 63929465-63929465 on Build GRCh38
rs145127044 Chr.20: 63929836-63929836 on Build GRCh38
rs2295437 Chr.20: 63929658-63929658 on Build GRCh38
rs748228389 Chr.20: 63929567-63929567 on Build GRCh38
rs182943774 Chr.20: 63929710-63929710 on Build GRCh38
rs532246570 Chr.20: 63929768-63929768 on Build GRCh38
rs770896397 Chr.20: 63929572-63929573 on Build GRCh38
rs113207069 Chr.20: 63929486-63929486 on Build GRCh38
rs554543887 Chr.20: 63929569-63929569 on Build GRCh38
rs773607766 Chr.20: 63929533-63929533 on Build GRCh38
rs568175223 Chr.20: 63929583-63929583 on Build GRCh38
rs371244374 Chr.20: 63929824-63929824 on Build GRCh38
rs926076708 Chr.20: 63929627-63929627 on Build GRCh38
rs756046159 Chr.20: 63929550-63929550 on Build GRCh38
rs190682209 Chr.20: 63929650-63929650 on Build GRCh38

View Full Product Details