Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63935214-63935725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5
Entrez Gene ID
80331
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
Gene Aliases
CLN4, CLN4B, CSP, DNAJC5A, NCL, mir-941-2, mir-941-3, mir-941-4, mir-941-5
UniGene
Hs.164419
Gene Chromosome Location
Chr.20: 63895102-63936031 on Build GRCh38
Transcripts
NM_025219.2 XM_011529048.2 XM_011529049.1

SNP Detail

SNP ID Chromosome Location
rs886056953 Chr.20: 63935438-63935438 on Build GRCh38
rs115323539 Chr.20: 63935563-63935563 on Build GRCh38
rs191412647 Chr.20: 63935697-63935697 on Build GRCh38
rs11554630 Chr.20: 63935685-63935685 on Build GRCh38
rs1040085624 Chr.20: 63935360-63935360 on Build GRCh38
rs1013607138 Chr.20: 63935497-63935497 on Build GRCh38
rs993856048 Chr.20: 63935389-63935389 on Build GRCh38
rs754459457 Chr.20: 63935498-63935498 on Build GRCh38
rs762613624 Chr.20: 63935270-63935270 on Build GRCh38
rs910315120 Chr.20: 63935254-63935254 on Build GRCh38
rs943074392 Chr.20: 63935329-63935329 on Build GRCh38
rs901495442 Chr.20: 63935363-63935363 on Build GRCh38
rs886056954 Chr.20: 63935625-63935625 on Build GRCh38
rs758582743 Chr.20: 63935604-63935604 on Build GRCh38
rs1006062790 Chr.20: 63935453-63935453 on Build GRCh38
rs576531330 Chr.20: 63935475-63935475 on Build GRCh38
rs146360147 Chr.20: 63935699-63935699 on Build GRCh38
rs887651921 Chr.20: 63935438-63935438 on Build GRCh38
rs928991131 Chr.20: 63935625-63935625 on Build GRCh38
rs138866062 Chr.20: 63935574-63935574 on Build GRCh38
rs958501503 Chr.20: 63935566-63935566 on Build GRCh38
rs564527134 Chr.20: 63935686-63935686 on Build GRCh38
rs768172464 Chr.20: 63935313-63935313 on Build GRCh38
rs369634697 Chr.20: 63935539-63935539 on Build GRCh38
rs3810500 Chr.20: 63935638-63935638 on Build GRCh38
rs773737847 Chr.20: 63935348-63935348 on Build GRCh38
rs542135679 Chr.20: 63935603-63935603 on Build GRCh38
rs1047637366 Chr.20: 63935406-63935406 on Build GRCh38
rs966730386 Chr.20: 63935508-63935508 on Build GRCh38
rs766779440 Chr.20: 63935483-63935483 on Build GRCh38
rs77107548 Chr.20: 63935238-63935238 on Build GRCh38
rs556682543 Chr.20: 63935425-63935425 on Build GRCh38
rs373099874 Chr.20: 63935633-63935633 on Build GRCh38
rs145028189 Chr.20: 63935407-63935407 on Build GRCh38
rs568079996 Chr.20: 63935382-63935382 on Build GRCh38
rs528703267 Chr.20: 63935567-63935567 on Build GRCh38
rs767414680 Chr.20: 63935639-63935639 on Build GRCh38
rs549826139 Chr.20: 63935239-63935241 on Build GRCh38
rs77777268 Chr.20: 63935575-63935575 on Build GRCh38

View Full Product Details