Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17561198-17561738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001207063.1 NM_001207064.1 NM_001207065.1 NM_001207066.1 NM_001338.4 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713857 c.822T>A p.H274Q Chr.21: 17561465-17561465 on Build GRCh38
5392721 c.814G>A p.E272K Chr.21: 17561457-17561457 on Build GRCh38
6587429 c.770T>A p.I257N Chr.21: 17561413-17561413 on Build GRCh38
1713856 c.814G>A p.E272K Chr.21: 17561457-17561457 on Build GRCh38
3764905 c.753G>A p.A251A Chr.21: 17561396-17561396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186357304 Chr.21: 17561598-17561598 on Build GRCh38
rs557805960 Chr.21: 17561277-17561277 on Build GRCh38
rs56962067 Chr.21: 17561243-17561244 on Build GRCh38
rs1042999616 Chr.21: 17561530-17561530 on Build GRCh38
rs533643575 Chr.21: 17561410-17561410 on Build GRCh38
rs756589633 Chr.21: 17561343-17561343 on Build GRCh38
rs778349868 Chr.21: 17561428-17561428 on Build GRCh38
rs760582039 Chr.21: 17561377-17561377 on Build GRCh38
rs1010913610 Chr.21: 17561689-17561689 on Build GRCh38
rs931098824 Chr.21: 17561269-17561269 on Build GRCh38
rs757637973 Chr.21: 17561433-17561433 on Build GRCh38
rs754261158 Chr.21: 17561432-17561432 on Build GRCh38
rs767607873 Chr.21: 17561406-17561406 on Build GRCh38
rs759787298 Chr.21: 17561503-17561503 on Build GRCh38
rs200024200 Chr.21: 17561239-17561240 on Build GRCh38
rs543846381 Chr.21: 17561250-17561250 on Build GRCh38
rs183522625 Chr.21: 17561554-17561554 on Build GRCh38
rs765611350 Chr.21: 17561521-17561521 on Build GRCh38
rs137939398 Chr.21: 17561477-17561477 on Build GRCh38
rs779083564 Chr.21: 17561308-17561309 on Build GRCh38
rs559458863 Chr.21: 17561599-17561599 on Build GRCh38
rs772388424 Chr.21: 17561323-17561325 on Build GRCh38
rs1027880100 Chr.21: 17561668-17561669 on Build GRCh38
rs768582795 Chr.21: 17561472-17561472 on Build GRCh38
rs757895680 Chr.21: 17561448-17561448 on Build GRCh38
rs951391329 Chr.21: 17561610-17561610 on Build GRCh38
rs912371525 Chr.21: 17561365-17561365 on Build GRCh38
rs77939418 Chr.21: 17561586-17561586 on Build GRCh38
rs148881358 Chr.21: 17561395-17561395 on Build GRCh38
rs573934328 Chr.21: 17561663-17561663 on Build GRCh38
rs980429796 Chr.21: 17561219-17561219 on Build GRCh38
rs372351020 Chr.21: 17561221-17561224 on Build GRCh38
rs199863730 Chr.21: 17561246-17561246 on Build GRCh38
rs1041934785 Chr.21: 17561707-17561707 on Build GRCh38
rs201516962 Chr.21: 17561244-17561244 on Build GRCh38
rs1053602327 Chr.21: 17561581-17561581 on Build GRCh38
rs781568544 Chr.21: 17561309-17561309 on Build GRCh38
rs771570606 Chr.21: 17561375-17561375 on Build GRCh38
rs760771144 Chr.21: 17561512-17561512 on Build GRCh38
rs112723687 Chr.21: 17561243-17561243 on Build GRCh38
rs774185810 Chr.21: 17561271-17561271 on Build GRCh38
rs17760603 Chr.21: 17561668-17561668 on Build GRCh38
rs750100259 Chr.21: 17561526-17561526 on Build GRCh38
rs536612224 Chr.21: 17561681-17561681 on Build GRCh38
rs201624272 Chr.21: 17561391-17561391 on Build GRCh38
rs372788152 Chr.21: 17561469-17561469 on Build GRCh38
rs748426780 Chr.21: 17561318-17561321 on Build GRCh38
rs143478011 Chr.21: 17561436-17561436 on Build GRCh38
rs971906499 Chr.21: 17561248-17561248 on Build GRCh38
rs921047397 Chr.21: 17561671-17561671 on Build GRCh38
rs750692034 Chr.21: 17561292-17561292 on Build GRCh38
rs982690033 Chr.21: 17561648-17561648 on Build GRCh38
rs908332571 Chr.21: 17561288-17561288 on Build GRCh38
rs766509347 Chr.21: 17561299-17561299 on Build GRCh38
rs760896186 Chr.21: 17561537-17561537 on Build GRCh38
rs759569781 Chr.21: 17561397-17561397 on Build GRCh38
rs1053403826 Chr.21: 17561673-17561673 on Build GRCh38
rs138289666 Chr.21: 17561489-17561489 on Build GRCh38
rs537375317 Chr.21: 17561376-17561376 on Build GRCh38
rs764103508 Chr.21: 17561546-17561546 on Build GRCh38
rs989417806 Chr.21: 17561227-17561227 on Build GRCh38
rs191106225 Chr.21: 17561672-17561672 on Build GRCh38
rs76069758 Chr.21: 17561242-17561243 on Build GRCh38
rs2824359 Chr.21: 17561510-17561510 on Build GRCh38
rs778263762 Chr.21: 17561344-17561344 on Build GRCh38
rs572463186 Chr.21: 17561616-17561616 on Build GRCh38
rs751593551 Chr.21: 17561304-17561304 on Build GRCh38
rs764713992 Chr.21: 17561516-17561516 on Build GRCh38
rs899509385 Chr.21: 17561425-17561425 on Build GRCh38
rs778561658 Chr.21: 17561374-17561374 on Build GRCh38
rs562154685 Chr.21: 17561274-17561274 on Build GRCh38
rs965196573 Chr.21: 17561688-17561688 on Build GRCh38
rs749007607 Chr.21: 17561362-17561362 on Build GRCh38
rs779600277 Chr.21: 17561468-17561468 on Build GRCh38
rs751752573 Chr.21: 17561396-17561396 on Build GRCh38
rs958658291 Chr.21: 17561273-17561273 on Build GRCh38
rs893417982 Chr.21: 17561492-17561492 on Build GRCh38
rs150493172 Chr.21: 17561318-17561318 on Build GRCh38
rs770704671 Chr.21: 17561373-17561373 on Build GRCh38
rs1047358778 Chr.21: 17561338-17561338 on Build GRCh38
rs549818224 Chr.21: 17561218-17561221 on Build GRCh38
rs535178575 Chr.21: 17561540-17561540 on Build GRCh38
rs141988381 Chr.21: 17561308-17561308 on Build GRCh38
rs1014454826 Chr.21: 17561400-17561400 on Build GRCh38
rs191648127 Chr.21: 17561520-17561520 on Build GRCh38
rs912390454 Chr.21: 17561662-17561662 on Build GRCh38
rs28595698 Chr.21: 17561678-17561678 on Build GRCh38
rs756679498 Chr.21: 17561424-17561424 on Build GRCh38
rs776490788 Chr.21: 17561483-17561483 on Build GRCh38
rs200360457 Chr.21: 17561393-17561393 on Build GRCh38
rs567772789 Chr.21: 17561528-17561528 on Build GRCh38
rs763147801 Chr.21: 17561298-17561298 on Build GRCh38
rs778453029 Chr.21: 17561434-17561434 on Build GRCh38
rs186473514 Chr.21: 17561306-17561306 on Build GRCh38
rs776403577 Chr.21: 17561379-17561379 on Build GRCh38
rs141516762 Chr.21: 17561244-17561245 on Build GRCh38
rs377231273 Chr.21: 17561497-17561497 on Build GRCh38
rs1003373738 Chr.21: 17561544-17561544 on Build GRCh38
rs927287712 Chr.21: 17561266-17561266 on Build GRCh38
rs201106254 Chr.21: 17561239-17561239 on Build GRCh38

View Full Product Details