Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17567624-17568128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001207063.1 NM_001207064.1 NM_001207065.1 NM_001207066.1 NM_001338.4 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

SNP Detail

SNP ID Chromosome Location
rs889646503 Chr.21: 17567890-17567890 on Build GRCh38
rs183719649 Chr.21: 17567847-17567847 on Build GRCh38
rs1036763923 Chr.21: 17567844-17567844 on Build GRCh38
rs2824361 Chr.21: 17567657-17567657 on Build GRCh38
rs985077824 Chr.21: 17567755-17567755 on Build GRCh38
rs926757900 Chr.21: 17567986-17567986 on Build GRCh38
rs897908318 Chr.21: 17568014-17568014 on Build GRCh38
rs1045278516 Chr.21: 17567942-17567942 on Build GRCh38
rs918198957 Chr.21: 17567925-17567925 on Build GRCh38
rs955935421 Chr.21: 17567845-17567845 on Build GRCh38
rs62239919 Chr.21: 17567875-17567875 on Build GRCh38
rs1032921067 Chr.21: 17567895-17567895 on Build GRCh38
rs189557666 Chr.21: 17568096-17568096 on Build GRCh38
rs397815769 Chr.21: 17567872-17567873 on Build GRCh38
rs187257859 Chr.21: 17567680-17567680 on Build GRCh38
rs576482300 Chr.21: 17567808-17567808 on Build GRCh38
rs201263478 Chr.21: 17567863-17567863 on Build GRCh38
rs548328022 Chr.21: 17567862-17567863 on Build GRCh38
rs183989507 Chr.21: 17567660-17567660 on Build GRCh38
rs779523118 Chr.21: 17567827-17567829 on Build GRCh38
rs2824362 Chr.21: 17568012-17568012 on Build GRCh38
rs541217767 Chr.21: 17567982-17567982 on Build GRCh38
rs144960946 Chr.21: 17567879-17567879 on Build GRCh38
rs535284574 Chr.21: 17567673-17567673 on Build GRCh38
rs1017444527 Chr.21: 17568104-17568104 on Build GRCh38
rs1051702738 Chr.21: 17568061-17568061 on Build GRCh38
rs993493484 Chr.21: 17568045-17568045 on Build GRCh38
rs565359390 Chr.21: 17567724-17567725 on Build GRCh38
rs543186109 Chr.21: 17568008-17568009 on Build GRCh38
rs949599670 Chr.21: 17567927-17567927 on Build GRCh38
rs942136245 Chr.21: 17567987-17567987 on Build GRCh38
rs939696762 Chr.21: 17567800-17567800 on Build GRCh38
rs11418205 Chr.21: 17567873-17567874 on Build GRCh38
rs1000494157 Chr.21: 17567893-17567893 on Build GRCh38
rs578160415 Chr.21: 17568044-17568044 on Build GRCh38
rs754988139 Chr.21: 17568043-17568043 on Build GRCh38
rs924302715 Chr.21: 17567831-17567831 on Build GRCh38
rs113927263 Chr.21: 17567684-17567684 on Build GRCh38
rs890390909 Chr.21: 17568089-17568089 on Build GRCh38
rs891782215 Chr.21: 17567906-17567906 on Build GRCh38
rs909518671 Chr.21: 17567758-17567758 on Build GRCh38
rs930849607 Chr.21: 17567744-17567745 on Build GRCh38
rs557801491 Chr.21: 17567770-17567770 on Build GRCh38
rs553943023 Chr.21: 17567678-17567678 on Build GRCh38
rs78888925 Chr.21: 17567873-17567873 on Build GRCh38
rs1049987904 Chr.21: 17567883-17567883 on Build GRCh38

View Full Product Details