Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17593495-17594007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG3
Entrez Gene ID
10950
Gene Name
BTG anti-proliferation factor 3
Gene Aliases
ANA, APRO4, TOB5, TOB55, TOFA
UniGene
Hs.473420
Gene Chromosome Location
Chr.21: 17593650-17612950 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 NM_001207066.1 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 NM_001207066.1 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

SNP Detail

SNP ID Chromosome Location
rs552951198 Chr.21: 17593708-17593710 on Build GRCh38
rs1032376921 Chr.21: 17593804-17593804 on Build GRCh38
rs28512353 Chr.21: 17593600-17593600 on Build GRCh38
rs949707834 Chr.21: 17593650-17593650 on Build GRCh38
rs1040038940 Chr.21: 17593835-17593835 on Build GRCh38
rs15459 Chr.21: 17593818-17593818 on Build GRCh38
rs886222977 Chr.21: 17593806-17593806 on Build GRCh38
rs190677133 Chr.21: 17593607-17593607 on Build GRCh38
rs988744150 Chr.21: 17593630-17593630 on Build GRCh38
rs925390693 Chr.21: 17593619-17593619 on Build GRCh38
rs867242330 Chr.21: 17593828-17593828 on Build GRCh38
rs887522228 Chr.21: 17593931-17593931 on Build GRCh38
rs937950022 Chr.21: 17593713-17593713 on Build GRCh38
rs958397948 Chr.21: 17593824-17593825 on Build GRCh38
rs1008597445 Chr.21: 17593811-17593811 on Build GRCh38
rs958200464 Chr.21: 17593646-17593646 on Build GRCh38
rs1014828175 Chr.21: 17593533-17593533 on Build GRCh38
rs186080055 Chr.21: 17593552-17593552 on Build GRCh38
rs149725830 Chr.21: 17593707-17593710 on Build GRCh38
rs539442992 Chr.21: 17593593-17593593 on Build GRCh38
rs1028548057 Chr.21: 17593920-17593920 on Build GRCh38
rs182507220 Chr.21: 17593609-17593609 on Build GRCh38
rs997518519 Chr.21: 17593908-17593908 on Build GRCh38
rs146798740 Chr.21: 17593686-17593688 on Build GRCh38
rs982336835 Chr.21: 17593844-17593844 on Build GRCh38
rs966710564 Chr.21: 17593666-17593666 on Build GRCh38
rs913951245 Chr.21: 17593649-17593649 on Build GRCh38
rs982145102 Chr.21: 17593683-17593683 on Build GRCh38
rs1021129305 Chr.21: 17593978-17593978 on Build GRCh38
rs143261518 Chr.21: 17593761-17593765 on Build GRCh38
rs1015209614 Chr.21: 17593725-17593725 on Build GRCh38
rs367675835 Chr.21: 17593917-17593917 on Build GRCh38
rs997810853 Chr.21: 17593754-17593754 on Build GRCh38
rs869211999 Chr.21: 17593765-17593769 on Build GRCh38
rs563413894 Chr.21: 17593929-17593929 on Build GRCh38
rs578146672 Chr.21: 17593940-17593940 on Build GRCh38
rs188071848 Chr.21: 17593833-17593833 on Build GRCh38
rs528767000 Chr.21: 17593535-17593535 on Build GRCh38
rs1008482936 Chr.21: 17593594-17593594 on Build GRCh38
rs897658073 Chr.21: 17593751-17593751 on Build GRCh38
rs898981226 Chr.21: 17593524-17593524 on Build GRCh38
rs1047442098 Chr.21: 17593558-17593558 on Build GRCh38
rs573786795 Chr.21: 17593719-17593719 on Build GRCh38
rs920437291 Chr.21: 17593755-17593755 on Build GRCh38
rs1046645514 Chr.21: 17593653-17593653 on Build GRCh38
rs114741737 Chr.21: 17593648-17593648 on Build GRCh38
rs555513644 Chr.21: 17593712-17593712 on Build GRCh38
rs1028936586 Chr.21: 17593628-17593628 on Build GRCh38
rs537213577 Chr.21: 17593711-17593711 on Build GRCh38
rs886108464 Chr.21: 17593559-17593559 on Build GRCh38
rs750353511 Chr.21: 17593596-17593596 on Build GRCh38
rs547018640 Chr.21: 17593539-17593539 on Build GRCh38
rs976104710 Chr.21: 17593579-17593579 on Build GRCh38
rs988324434 Chr.21: 17593824-17593824 on Build GRCh38
rs936977210 Chr.21: 17593914-17593914 on Build GRCh38
rs374710526 Chr.21: 17593801-17593801 on Build GRCh38
rs751383282 Chr.21: 17593799-17593799 on Build GRCh38
rs552998857 Chr.21: 17593785-17593785 on Build GRCh38
rs577577077 Chr.21: 17593802-17593802 on Build GRCh38
rs1055039268 Chr.21: 17593714-17593714 on Build GRCh38
rs900151319 Chr.21: 17593881-17593881 on Build GRCh38
rs2824386 Chr.21: 17593771-17593771 on Build GRCh38

View Full Product Details