Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17594201-17594632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG3
Entrez Gene ID
10950
Gene Name
BTG anti-proliferation factor 3
Gene Aliases
ANA, APRO4, TOB5, TOB55, TOFA
UniGene
Hs.473420
Gene Chromosome Location
Chr.21: 17593650-17612950 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 XM_011529476.2 XM_011529477.2 XM_011529478.2 XM_011529479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5858449 c.721C>T p.R241* Chr.21: 17594263-17594263 on Build GRCh38
373069 c.577C>T p.P193S Chr.21: 17594275-17594275 on Build GRCh38
265615 c.575_576insA p.P193fs*29 Chr.21: 17594276-17594277 on Build GRCh38
5784496 c.724G>C p.G242R Chr.21: 17594260-17594260 on Build GRCh38
2840959 c.629C>T p.P210L Chr.21: 17594223-17594223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750033952 Chr.21: 17594249-17594254 on Build GRCh38
rs766724664 Chr.21: 17594352-17594356 on Build GRCh38
rs765374841 Chr.21: 17594237-17594237 on Build GRCh38
rs865991301 Chr.21: 17594314-17594314 on Build GRCh38
rs145339937 Chr.21: 17594265-17594265 on Build GRCh38
rs1035088072 Chr.21: 17594420-17594420 on Build GRCh38
rs770438493 Chr.21: 17594293-17594293 on Build GRCh38
rs760402424 Chr.21: 17594310-17594310 on Build GRCh38
rs574628649 Chr.21: 17594376-17594376 on Build GRCh38
rs983318240 Chr.21: 17594490-17594490 on Build GRCh38
rs754321733 Chr.21: 17594364-17594364 on Build GRCh38
rs534529331 Chr.21: 17594417-17594417 on Build GRCh38
rs747791396 Chr.21: 17594506-17594506 on Build GRCh38
rs759495841 Chr.21: 17594301-17594301 on Build GRCh38
rs758454807 Chr.21: 17594241-17594241 on Build GRCh38
rs762011087 Chr.21: 17594235-17594235 on Build GRCh38
rs532920211 Chr.21: 17594294-17594294 on Build GRCh38
rs201393065 Chr.21: 17594413-17594413 on Build GRCh38
rs941716098 Chr.21: 17594239-17594239 on Build GRCh38
rs753128595 Chr.21: 17594347-17594347 on Build GRCh38
rs5842649 Chr.21: 17594407-17594410 on Build GRCh38
rs907724782 Chr.21: 17594494-17594494 on Build GRCh38
rs200170210 Chr.21: 17594228-17594228 on Build GRCh38
rs762983299 Chr.21: 17594331-17594331 on Build GRCh38
rs772358756 Chr.21: 17594477-17594477 on Build GRCh38
rs746230646 Chr.21: 17594467-17594467 on Build GRCh38
rs755502179 Chr.21: 17594339-17594339 on Build GRCh38
rs140021036 Chr.21: 17594262-17594262 on Build GRCh38
rs202093449 Chr.21: 17594410-17594410 on Build GRCh38
rs765466233 Chr.21: 17594315-17594315 on Build GRCh38
rs771821699 Chr.21: 17594302-17594302 on Build GRCh38
rs532673402 Chr.21: 17594538-17594538 on Build GRCh38
rs756766246 Chr.21: 17594356-17594356 on Build GRCh38
rs748774075 Chr.21: 17594280-17594280 on Build GRCh38
rs745861748 Chr.21: 17594371-17594371 on Build GRCh38
rs367639121 Chr.21: 17594345-17594345 on Build GRCh38
rs538575992 Chr.21: 17594602-17594602 on Build GRCh38
rs35351204 Chr.21: 17594415-17594416 on Build GRCh38
rs931624914 Chr.21: 17594242-17594242 on Build GRCh38
rs567409990 Chr.21: 17594384-17594384 on Build GRCh38
rs569694722 Chr.21: 17594335-17594335 on Build GRCh38
rs777512668 Chr.21: 17594277-17594277 on Build GRCh38
rs571230538 Chr.21: 17594517-17594517 on Build GRCh38
rs182523222 Chr.21: 17594257-17594257 on Build GRCh38
rs767142948 Chr.21: 17594228-17594230 on Build GRCh38
rs772753453 Chr.21: 17594554-17594554 on Build GRCh38
rs3833345 Chr.21: 17594412-17594415 on Build GRCh38
rs551416462 Chr.21: 17594323-17594323 on Build GRCh38
rs1003643532 Chr.21: 17594389-17594389 on Build GRCh38
rs963356761 Chr.21: 17594601-17594601 on Build GRCh38
rs902671516 Chr.21: 17594368-17594368 on Build GRCh38
rs769223899 Chr.21: 17594526-17594526 on Build GRCh38
rs976237783 Chr.21: 17594512-17594512 on Build GRCh38
rs1033806192 Chr.21: 17594544-17594547 on Build GRCh38
rs776165689 Chr.21: 17594499-17594499 on Build GRCh38
rs1054168248 Chr.21: 17594579-17594579 on Build GRCh38
rs951987301 Chr.21: 17594487-17594487 on Build GRCh38
rs750514745 Chr.21: 17594238-17594238 on Build GRCh38
rs894094772 Chr.21: 17594259-17594259 on Build GRCh38
rs548853527 Chr.21: 17594365-17594365 on Build GRCh38
rs373951463 Chr.21: 17594263-17594263 on Build GRCh38
rs755516239 Chr.21: 17594374-17594376 on Build GRCh38
rs2824387 Chr.21: 17594465-17594465 on Build GRCh38
rs879207913 Chr.21: 17594281-17594281 on Build GRCh38
rs760950585 Chr.21: 17594347-17594350 on Build GRCh38
rs984341924 Chr.21: 17594556-17594556 on Build GRCh38
rs1036501191 Chr.21: 17594247-17594247 on Build GRCh38
rs892777962 Chr.21: 17594608-17594608 on Build GRCh38
rs192593680 Chr.21: 17594227-17594227 on Build GRCh38
rs959534481 Chr.21: 17594552-17594552 on Build GRCh38
rs537124505 Chr.21: 17594355-17594355 on Build GRCh38
rs769576476 Chr.21: 17594268-17594268 on Build GRCh38
rs775262362 Chr.21: 17594304-17594304 on Build GRCh38
rs147482425 Chr.21: 17594254-17594254 on Build GRCh38
rs768067837 Chr.21: 17594340-17594340 on Build GRCh38
rs200554253 Chr.21: 17594352-17594352 on Build GRCh38
rs756501688 Chr.21: 17594350-17594350 on Build GRCh38
rs3074937 Chr.21: 17594409-17594412 on Build GRCh38
rs946982141 Chr.21: 17594609-17594609 on Build GRCh38
rs756169731 Chr.21: 17594232-17594232 on Build GRCh38
rs373867022 Chr.21: 17594336-17594336 on Build GRCh38

View Full Product Details