Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 18245794-18246290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHODL
Entrez Gene ID
140578
Gene Name
chondrolectin
Gene Aliases
C21orf68, MT75, PRED12
UniGene
Hs.283725
Gene Chromosome Location
Chr.21: 17819329-18267371 on Build GRCh38
Transcripts
NM_001204174.1 NM_001204175.1 NM_001204176.1 NM_001204177.1 NM_001204178.1 NM_024944.2 XM_011529453.2 XM_011529457.2 XM_017028273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200850 c.22+6T>C p.? Chr.21: 18245975-18245975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs8129797 Chr.21: 18245945-18245945 on Build GRCh38
rs74889189 Chr.21: 18246154-18246154 on Build GRCh38
rs145550037 Chr.21: 18246082-18246082 on Build GRCh38
rs911582283 Chr.21: 18245891-18245891 on Build GRCh38
rs1007394994 Chr.21: 18246087-18246087 on Build GRCh38
rs530821187 Chr.21: 18246169-18246169 on Build GRCh38
rs973273239 Chr.21: 18246084-18246084 on Build GRCh38
rs8129796 Chr.21: 18245944-18245944 on Build GRCh38
rs531919580 Chr.21: 18246033-18246033 on Build GRCh38
rs888588868 Chr.21: 18245989-18245989 on Build GRCh38
rs1043920017 Chr.21: 18246239-18246239 on Build GRCh38
rs558464359 Chr.21: 18245886-18245886 on Build GRCh38
rs992951800 Chr.21: 18246246-18246246 on Build GRCh38
rs564653345 Chr.21: 18246024-18246024 on Build GRCh38
rs9980998 Chr.21: 18246004-18246004 on Build GRCh38
rs999719580 Chr.21: 18246263-18246263 on Build GRCh38
rs71318134 Chr.21: 18246245-18246245 on Build GRCh38
rs1036090520 Chr.21: 18246046-18246046 on Build GRCh38
rs924497386 Chr.21: 18246177-18246177 on Build GRCh38
rs1021972046 Chr.21: 18246150-18246150 on Build GRCh38
rs75025284 Chr.21: 18246086-18246086 on Build GRCh38
rs577484944 Chr.21: 18245871-18245871 on Build GRCh38
rs1057141520 Chr.21: 18245902-18245902 on Build GRCh38
rs76025562 Chr.21: 18245981-18245981 on Build GRCh38
rs950643380 Chr.21: 18246120-18246120 on Build GRCh38
rs915874575 Chr.21: 18246255-18246255 on Build GRCh38
rs759935109 Chr.21: 18245955-18245955 on Build GRCh38
rs8130922 Chr.21: 18246012-18246012 on Build GRCh38
rs960724896 Chr.21: 18245985-18245985 on Build GRCh38
rs747553502 Chr.21: 18245972-18245972 on Build GRCh38
rs8129789 Chr.21: 18245935-18245935 on Build GRCh38
rs1012283540 Chr.21: 18245990-18245990 on Build GRCh38
rs533723758 Chr.21: 18245824-18245824 on Build GRCh38
rs946847643 Chr.21: 18246237-18246237 on Build GRCh38
rs924705433 Chr.21: 18245841-18245841 on Build GRCh38
rs921711476 Chr.21: 18246232-18246232 on Build GRCh38
rs542368938 Chr.21: 18245957-18245957 on Build GRCh38
rs769792407 Chr.21: 18245973-18245973 on Build GRCh38
rs570652640 Chr.21: 18245930-18245930 on Build GRCh38
rs900668616 Chr.21: 18245931-18245931 on Build GRCh38
rs897552193 Chr.21: 18246054-18246054 on Build GRCh38
rs755582172 Chr.21: 18246079-18246079 on Build GRCh38
rs554291404 Chr.21: 18245963-18245963 on Build GRCh38
rs915948093 Chr.21: 18245959-18245959 on Build GRCh38
rs938802119 Chr.21: 18245889-18245889 on Build GRCh38
rs944422557 Chr.21: 18245895-18245895 on Build GRCh38
rs561784143 Chr.21: 18246160-18246160 on Build GRCh38
rs1038834314 Chr.21: 18245926-18245926 on Build GRCh38
rs770453363 Chr.21: 18245970-18245971 on Build GRCh38
rs779390028 Chr.21: 18246208-18246208 on Build GRCh38

View Full Product Details