Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26467906-26468445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
UniGene
Hs.37445
Gene Chromosome Location
Chr.21: 26466209-26573404 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 NR_135515.1 NR_135516.1

Gene Symbol
CYYR1-AS1
Entrez Gene ID
100996571
Gene Name
cysteine and tyrosine rich 1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.570432
Gene Chromosome Location
Chr.21: 26393635-26569252 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 NR_135515.1 NR_135516.1

SNP Detail

SNP ID Chromosome Location
rs183308924 Chr.21: 26468402-26468402 on Build GRCh38
rs1005134929 Chr.21: 26468278-26468278 on Build GRCh38
rs996328145 Chr.21: 26468254-26468254 on Build GRCh38
rs915081410 Chr.21: 26468417-26468417 on Build GRCh38
rs115409952 Chr.21: 26467945-26467945 on Build GRCh38
rs981874387 Chr.21: 26468376-26468376 on Build GRCh38
rs568868439 Chr.21: 26468188-26468188 on Build GRCh38
rs765427597 Chr.21: 26468308-26468308 on Build GRCh38
rs753967800 Chr.21: 26468197-26468197 on Build GRCh38
rs1034416169 Chr.21: 26468382-26468382 on Build GRCh38
rs569024240 Chr.21: 26468415-26468415 on Build GRCh38
rs1036467296 Chr.21: 26468360-26468360 on Build GRCh38
rs952285194 Chr.21: 26468044-26468044 on Build GRCh38
rs887989439 Chr.21: 26468273-26468273 on Build GRCh38
rs887813594 Chr.21: 26468351-26468351 on Build GRCh38
rs931986979 Chr.21: 26467939-26467939 on Build GRCh38
rs971375227 Chr.21: 26468314-26468314 on Build GRCh38
rs554451224 Chr.21: 26467993-26467993 on Build GRCh38
rs957331733 Chr.21: 26468072-26468072 on Build GRCh38
rs981540394 Chr.21: 26468080-26468080 on Build GRCh38
rs562238685 Chr.21: 26468176-26468176 on Build GRCh38
rs773348781 Chr.21: 26468084-26468084 on Build GRCh38
rs767529730 Chr.21: 26468052-26468052 on Build GRCh38
rs191048525 Chr.21: 26468275-26468275 on Build GRCh38
rs191044338 Chr.21: 26467990-26467990 on Build GRCh38
rs576597536 Chr.21: 26468043-26468043 on Build GRCh38
rs753078419 Chr.21: 26468341-26468341 on Build GRCh38
rs1009006767 Chr.21: 26468223-26468223 on Build GRCh38
rs1014230985 Chr.21: 26468027-26468027 on Build GRCh38
rs564978852 Chr.21: 26468300-26468300 on Build GRCh38
rs2830239 Chr.21: 26467978-26467978 on Build GRCh38
rs988605710 Chr.21: 26468134-26468134 on Build GRCh38
rs375596323 Chr.21: 26468322-26468322 on Build GRCh38
rs576779627 Chr.21: 26468309-26468309 on Build GRCh38
rs758830765 Chr.21: 26468408-26468408 on Build GRCh38
rs185954373 Chr.21: 26468165-26468165 on Build GRCh38
rs927399786 Chr.21: 26468132-26468132 on Build GRCh38
rs959056023 Chr.21: 26468395-26468395 on Build GRCh38
rs1027832231 Chr.21: 26468269-26468271 on Build GRCh38
rs946593047 Chr.21: 26468420-26468420 on Build GRCh38
rs1032719415 Chr.21: 26468066-26468066 on Build GRCh38
rs760824288 Chr.21: 26468111-26468111 on Build GRCh38
rs1055388004 Chr.21: 26468142-26468142 on Build GRCh38
rs921859918 Chr.21: 26468240-26468240 on Build GRCh38
rs546801413 Chr.21: 26467975-26467975 on Build GRCh38
rs553346111 Chr.21: 26467935-26467935 on Build GRCh38
rs887854045 Chr.21: 26467969-26467969 on Build GRCh38
rs902186851 Chr.21: 26468214-26468214 on Build GRCh38
rs182829858 Chr.21: 26468065-26468065 on Build GRCh38
rs931933172 Chr.21: 26468245-26468245 on Build GRCh38
rs1040103092 Chr.21: 26468229-26468229 on Build GRCh38
rs219643 Chr.21: 26468248-26468248 on Build GRCh38
rs915130704 Chr.21: 26468155-26468157 on Build GRCh38
rs1005005674 Chr.21: 26467973-26467973 on Build GRCh38
rs940743068 Chr.21: 26468352-26468352 on Build GRCh38
rs143100369 Chr.21: 26468056-26468056 on Build GRCh38
rs533321787 Chr.21: 26468401-26468401 on Build GRCh38
rs142060580 Chr.21: 26468020-26468020 on Build GRCh38

View Full Product Details