Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29326305-29326817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6161063 c.955G>T p.G319* Chr.21: 29326779-29326779 on Build GRCh38
1029784 c.488G>T p.R163M Chr.21: 29326312-29326312 on Build GRCh38
4818995 c.965C>G p.S322C Chr.21: 29326789-29326789 on Build GRCh38
1029785 c.683G>T p.R228I Chr.21: 29326507-29326507 on Build GRCh38
7287244 c.949C>A p.P317T Chr.21: 29326773-29326773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146651304 Chr.21: 29326411-29326411 on Build GRCh38
rs545357951 Chr.21: 29326574-29326574 on Build GRCh38
rs374723065 Chr.21: 29326417-29326417 on Build GRCh38
rs757377063 Chr.21: 29326631-29326631 on Build GRCh38
rs574088955 Chr.21: 29326496-29326496 on Build GRCh38
rs764486069 Chr.21: 29326737-29326737 on Build GRCh38
rs755532442 Chr.21: 29326356-29326356 on Build GRCh38
rs573908981 Chr.21: 29326657-29326657 on Build GRCh38
rs970456121 Chr.21: 29326474-29326474 on Build GRCh38
rs35474725 Chr.21: 29326764-29326764 on Build GRCh38
rs768118683 Chr.21: 29326463-29326463 on Build GRCh38
rs369485706 Chr.21: 29326753-29326753 on Build GRCh38
rs77477102 Chr.21: 29326664-29326664 on Build GRCh38
rs368476331 Chr.21: 29326542-29326542 on Build GRCh38
rs755790815 Chr.21: 29326650-29326650 on Build GRCh38
rs374802824 Chr.21: 29326626-29326626 on Build GRCh38
rs780874382 Chr.21: 29326774-29326774 on Build GRCh38
rs542608015 Chr.21: 29326513-29326513 on Build GRCh38
rs769864742 Chr.21: 29326783-29326783 on Build GRCh38
rs147272592 Chr.21: 29326677-29326677 on Build GRCh38
rs571532697 Chr.21: 29326373-29326373 on Build GRCh38
rs914848850 Chr.21: 29326368-29326368 on Build GRCh38
rs772331744 Chr.21: 29326413-29326415 on Build GRCh38
rs140102895 Chr.21: 29326425-29326425 on Build GRCh38
rs750517099 Chr.21: 29326759-29326759 on Build GRCh38
rs372369161 Chr.21: 29326628-29326628 on Build GRCh38
rs372118522 Chr.21: 29326549-29326549 on Build GRCh38
rs372287138 Chr.21: 29326696-29326696 on Build GRCh38
rs780500015 Chr.21: 29326346-29326346 on Build GRCh38
rs536766184 Chr.21: 29326453-29326453 on Build GRCh38
rs544317112 Chr.21: 29326639-29326639 on Build GRCh38
rs745638584 Chr.21: 29326539-29326539 on Build GRCh38
rs747643943 Chr.21: 29326684-29326684 on Build GRCh38
rs147328273 Chr.21: 29326427-29326427 on Build GRCh38
rs201844220 Chr.21: 29326506-29326506 on Build GRCh38
rs957755223 Chr.21: 29326497-29326497 on Build GRCh38
rs375506236 Chr.21: 29326697-29326697 on Build GRCh38
rs375713948 Chr.21: 29326456-29326456 on Build GRCh38
rs751986135 Chr.21: 29326352-29326352 on Build GRCh38
rs1046350996 Chr.21: 29326752-29326752 on Build GRCh38
rs1001859623 Chr.21: 29326493-29326493 on Build GRCh38
rs553355487 Chr.21: 29326543-29326543 on Build GRCh38
rs762633394 Chr.21: 29326705-29326705 on Build GRCh38
rs61735773 Chr.21: 29326451-29326451 on Build GRCh38
rs779145687 Chr.21: 29326537-29326537 on Build GRCh38
rs757462849 Chr.21: 29326521-29326521 on Build GRCh38
rs749402266 Chr.21: 29326392-29326392 on Build GRCh38
rs746048388 Chr.21: 29326633-29326633 on Build GRCh38
rs761138517 Chr.21: 29326736-29326736 on Build GRCh38
rs753098656 Chr.21: 29326481-29326481 on Build GRCh38
rs138981213 Chr.21: 29326452-29326452 on Build GRCh38
rs777938054 Chr.21: 29326384-29326384 on Build GRCh38
rs754294251 Chr.21: 29326746-29326746 on Build GRCh38
rs933278998 Chr.21: 29326387-29326387 on Build GRCh38
rs746856749 Chr.21: 29326547-29326547 on Build GRCh38
rs961392975 Chr.21: 29326692-29326692 on Build GRCh38
rs112330292 Chr.21: 29326399-29326399 on Build GRCh38
rs754856199 Chr.21: 29326768-29326768 on Build GRCh38
rs148287786 Chr.21: 29326725-29326725 on Build GRCh38
rs769815586 Chr.21: 29326383-29326383 on Build GRCh38
rs756562023 Chr.21: 29326627-29326627 on Build GRCh38
rs771966161 Chr.21: 29326423-29326423 on Build GRCh38
rs771150831 Chr.21: 29326393-29326393 on Build GRCh38
rs764495595 Chr.21: 29326611-29326611 on Build GRCh38
rs748061224 Chr.21: 29326782-29326782 on Build GRCh38
rs371081000 Chr.21: 29326442-29326442 on Build GRCh38
rs4817282 Chr.21: 29326634-29326634 on Build GRCh38
rs762721028 Chr.21: 29326619-29326619 on Build GRCh38
rs147096876 Chr.21: 29326562-29326562 on Build GRCh38
rs779658191 Chr.21: 29326649-29326649 on Build GRCh38
rs915682429 Chr.21: 29326751-29326751 on Build GRCh38
rs200303423 Chr.21: 29326460-29326460 on Build GRCh38
rs776674031 Chr.21: 29326680-29326680 on Build GRCh38
rs34811108 Chr.21: 29326553-29326553 on Build GRCh38
rs774116353 Chr.21: 29326403-29326403 on Build GRCh38
rs748231244 Chr.21: 29326374-29326374 on Build GRCh38
rs764957109 Chr.21: 29326572-29326572 on Build GRCh38
rs775709740 Chr.21: 29326726-29326726 on Build GRCh38
rs751284536 Chr.21: 29326767-29326767 on Build GRCh38
rs61735774 Chr.21: 29326632-29326632 on Build GRCh38
rs767704566 Chr.21: 29326624-29326624 on Build GRCh38
rs780390713 Chr.21: 29326766-29326766 on Build GRCh38
rs763472575 Chr.21: 29326437-29326437 on Build GRCh38

View Full Product Details