Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29327050-29327548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6161062 c.1417T>A p.S473T Chr.21: 29327241-29327241 on Build GRCh38
1737754 c.1543T>C p.C515R Chr.21: 29327367-29327367 on Build GRCh38
6190165 c.1561G>C p.E521Q Chr.21: 29327385-29327385 on Build GRCh38
7428661 c.1325delT p.S443fs*14 Chr.21: 29327149-29327149 on Build GRCh38
7429885 c.1390A>G p.I464V Chr.21: 29327214-29327214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761553877 Chr.21: 29327242-29327242 on Build GRCh38
rs762912319 Chr.21: 29327258-29327258 on Build GRCh38
rs201222202 Chr.21: 29327161-29327161 on Build GRCh38
rs775802435 Chr.21: 29327152-29327152 on Build GRCh38
rs1035248328 Chr.21: 29327109-29327109 on Build GRCh38
rs749678444 Chr.21: 29327209-29327209 on Build GRCh38
rs575620663 Chr.21: 29327230-29327230 on Build GRCh38
rs200791937 Chr.21: 29327132-29327132 on Build GRCh38
rs199690512 Chr.21: 29327131-29327131 on Build GRCh38
rs973795883 Chr.21: 29327487-29327487 on Build GRCh38
rs779363849 Chr.21: 29327302-29327302 on Build GRCh38
rs941994895 Chr.21: 29327125-29327125 on Build GRCh38
rs746309353 Chr.21: 29327303-29327303 on Build GRCh38
rs753767412 Chr.21: 29327078-29327078 on Build GRCh38
rs200802905 Chr.21: 29327203-29327203 on Build GRCh38
rs775873520 Chr.21: 29327483-29327483 on Build GRCh38
rs1030605246 Chr.21: 29327075-29327075 on Build GRCh38
rs749919709 Chr.21: 29327187-29327187 on Build GRCh38
rs762823839 Chr.21: 29327360-29327360 on Build GRCh38
rs145991657 Chr.21: 29327192-29327192 on Build GRCh38
rs544651776 Chr.21: 29327375-29327375 on Build GRCh38
rs540348624 Chr.21: 29327458-29327458 on Build GRCh38
rs199802427 Chr.21: 29327244-29327244 on Build GRCh38
rs779086628 Chr.21: 29327411-29327411 on Build GRCh38
rs986315548 Chr.21: 29327089-29327089 on Build GRCh38
rs140585235 Chr.21: 29327336-29327336 on Build GRCh38
rs2239572 Chr.21: 29327521-29327521 on Build GRCh38
rs139219595 Chr.21: 29327105-29327105 on Build GRCh38
rs780267098 Chr.21: 29327317-29327317 on Build GRCh38
rs367693467 Chr.21: 29327358-29327358 on Build GRCh38
rs751034308 Chr.21: 29327371-29327371 on Build GRCh38
rs1012459797 Chr.21: 29327490-29327490 on Build GRCh38
rs748491382 Chr.21: 29327122-29327122 on Build GRCh38
rs371330530 Chr.21: 29327366-29327366 on Build GRCh38
rs752248312 Chr.21: 29327382-29327382 on Build GRCh38
rs778144906 Chr.21: 29327207-29327207 on Build GRCh38
rs559888699 Chr.21: 29327484-29327484 on Build GRCh38
rs143227772 Chr.21: 29327227-29327227 on Build GRCh38
rs780747281 Chr.21: 29327444-29327444 on Build GRCh38
rs112285327 Chr.21: 29327486-29327486 on Build GRCh38
rs750875639 Chr.21: 29327415-29327415 on Build GRCh38
rs1049633413 Chr.21: 29327267-29327267 on Build GRCh38
rs757513028 Chr.21: 29327301-29327301 on Build GRCh38
rs769438465 Chr.21: 29327348-29327348 on Build GRCh38
rs963378730 Chr.21: 29327455-29327455 on Build GRCh38
rs765154710 Chr.21: 29327186-29327186 on Build GRCh38
rs149911725 Chr.21: 29327216-29327216 on Build GRCh38
rs775714503 Chr.21: 29327235-29327235 on Build GRCh38
rs769056682 Chr.21: 29327238-29327238 on Build GRCh38
rs758968799 Chr.21: 29327443-29327443 on Build GRCh38
rs780743906 Chr.21: 29327283-29327283 on Build GRCh38
rs745428812 Chr.21: 29327106-29327106 on Build GRCh38
rs768892267 Chr.21: 29327335-29327335 on Build GRCh38
rs771359504 Chr.21: 29327141-29327141 on Build GRCh38
rs375374696 Chr.21: 29327376-29327376 on Build GRCh38
rs755962041 Chr.21: 29327460-29327461 on Build GRCh38
rs754615744 Chr.21: 29327274-29327274 on Build GRCh38
rs139083953 Chr.21: 29327118-29327118 on Build GRCh38
rs754204780 Chr.21: 29327296-29327296 on Build GRCh38
rs752679001 Chr.21: 29327182-29327182 on Build GRCh38
rs911049235 Chr.21: 29327299-29327299 on Build GRCh38
rs144158872 Chr.21: 29327142-29327142 on Build GRCh38
rs761007030 Chr.21: 29327158-29327158 on Build GRCh38
rs759570045 Chr.21: 29327145-29327145 on Build GRCh38
rs142524709 Chr.21: 29327264-29327264 on Build GRCh38
rs751087323 Chr.21: 29327265-29327265 on Build GRCh38
rs765294150 Chr.21: 29327479-29327479 on Build GRCh38
rs902242107 Chr.21: 29327261-29327261 on Build GRCh38
rs760753557 Chr.21: 29327198-29327198 on Build GRCh38
rs755794107 Chr.21: 29327404-29327404 on Build GRCh38
rs757204896 Chr.21: 29327185-29327185 on Build GRCh38
rs771433838 Chr.21: 29327214-29327214 on Build GRCh38
rs759528868 Chr.21: 29327461-29327461 on Build GRCh38
rs770098097 Chr.21: 29327130-29327130 on Build GRCh38
rs150888228 Chr.21: 29327079-29327079 on Build GRCh38
rs185275351 Chr.21: 29327100-29327100 on Build GRCh38
rs530151866 Chr.21: 29327247-29327247 on Build GRCh38
rs189708113 Chr.21: 29327189-29327189 on Build GRCh38
rs754634127 Chr.21: 29327204-29327204 on Build GRCh38
rs148401992 Chr.21: 29327239-29327239 on Build GRCh38
rs201560153 Chr.21: 29327330-29327330 on Build GRCh38
rs35766001 Chr.21: 29327236-29327236 on Build GRCh38
rs753587832 Chr.21: 29327165-29327165 on Build GRCh38
rs151321858 Chr.21: 29327316-29327316 on Build GRCh38
rs762119140 Chr.21: 29327342-29327342 on Build GRCh38
rs779724096 Chr.21: 29327119-29327119 on Build GRCh38
rs919335895 Chr.21: 29327169-29327169 on Build GRCh38
rs767823826 Chr.21: 29327151-29327151 on Build GRCh38

View Full Product Details