Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30215409-30215912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN8
Entrez Gene ID
9073
Gene Name
claudin 8
Gene Aliases
HEL-S-79
UniGene
Hs.162209
Gene Chromosome Location
Chr.21: 30214006-30216151 on Build GRCh38
Transcripts
NM_199328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3550130 c.120C>T p.I40I Chr.21: 30215806-30215806 on Build GRCh38
6094344 c.428C>A p.A143D Chr.21: 30215498-30215498 on Build GRCh38
6094342 c.185T>A p.M62K Chr.21: 30215741-30215741 on Build GRCh38
3423892 c.156G>C p.M52I Chr.21: 30215770-30215770 on Build GRCh38
6004708 c.454A>G p.I152V Chr.21: 30215472-30215472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376757104 Chr.21: 30215564-30215564 on Build GRCh38
rs372970119 Chr.21: 30215662-30215662 on Build GRCh38
rs141620647 Chr.21: 30215794-30215794 on Build GRCh38
rs761949265 Chr.21: 30215601-30215601 on Build GRCh38
rs762203558 Chr.21: 30215597-30215597 on Build GRCh38
rs775856986 Chr.21: 30215491-30215491 on Build GRCh38
rs555436710 Chr.21: 30215540-30215540 on Build GRCh38
rs761303990 Chr.21: 30215499-30215499 on Build GRCh38
rs778164419 Chr.21: 30215805-30215805 on Build GRCh38
rs763174117 Chr.21: 30215635-30215635 on Build GRCh38
rs748719176 Chr.21: 30215713-30215713 on Build GRCh38
rs766571800 Chr.21: 30215726-30215726 on Build GRCh38
rs767844489 Chr.21: 30215681-30215681 on Build GRCh38
rs762973038 Chr.21: 30215720-30215720 on Build GRCh38
rs685967 Chr.21: 30215541-30215541 on Build GRCh38
rs753772677 Chr.21: 30215844-30215844 on Build GRCh38
rs370704016 Chr.21: 30215568-30215568 on Build GRCh38
rs986413133 Chr.21: 30215831-30215831 on Build GRCh38
rs773835736 Chr.21: 30215517-30215518 on Build GRCh38
rs200767039 Chr.21: 30215553-30215553 on Build GRCh38
rs756482500 Chr.21: 30215869-30215869 on Build GRCh38
rs764337784 Chr.21: 30215435-30215435 on Build GRCh38
rs755617586 Chr.21: 30215674-30215674 on Build GRCh38
rs749038670 Chr.21: 30215563-30215563 on Build GRCh38
rs764708430 Chr.21: 30215855-30215855 on Build GRCh38
rs763946058 Chr.21: 30215771-30215771 on Build GRCh38
rs61743791 Chr.21: 30215645-30215645 on Build GRCh38
rs765475207 Chr.21: 30215625-30215625 on Build GRCh38
rs756745321 Chr.21: 30215786-30215786 on Build GRCh38
rs762018445 Chr.21: 30215512-30215512 on Build GRCh38
rs868192260 Chr.21: 30215629-30215629 on Build GRCh38
rs931878628 Chr.21: 30215535-30215535 on Build GRCh38
rs369970339 Chr.21: 30215472-30215472 on Build GRCh38
rs749962253 Chr.21: 30215857-30215857 on Build GRCh38
rs753906623 Chr.21: 30215506-30215506 on Build GRCh38
rs370248819 Chr.21: 30215859-30215859 on Build GRCh38
rs775201255 Chr.21: 30215827-30215827 on Build GRCh38
rs963177454 Chr.21: 30215868-30215868 on Build GRCh38
rs978915979 Chr.21: 30215766-30215766 on Build GRCh38
rs754103569 Chr.21: 30215448-30215448 on Build GRCh38
rs539459302 Chr.21: 30215458-30215458 on Build GRCh38
rs752619315 Chr.21: 30215763-30215763 on Build GRCh38
rs201941862 Chr.21: 30215688-30215688 on Build GRCh38
rs387907424 Chr.21: 30215764-30215764 on Build GRCh38
rs146174025 Chr.21: 30215537-30215537 on Build GRCh38
rs55884670 Chr.21: 30215636-30215636 on Build GRCh38
rs781640763 Chr.21: 30215800-30215800 on Build GRCh38
rs748763576 Chr.21: 30215677-30215677 on Build GRCh38
rs967083664 Chr.21: 30215704-30215704 on Build GRCh38
rs567735724 Chr.21: 30215701-30215701 on Build GRCh38
rs769169043 Chr.21: 30215758-30215758 on Build GRCh38
rs768034106 Chr.21: 30215468-30215468 on Build GRCh38
rs772187336 Chr.21: 30215579-30215579 on Build GRCh38
rs761359261 Chr.21: 30215848-30215848 on Build GRCh38
rs893943148 Chr.21: 30215661-30215661 on Build GRCh38
rs141376936 Chr.21: 30215546-30215546 on Build GRCh38
rs1030402102 Chr.21: 30215738-30215738 on Build GRCh38
rs79452452 Chr.21: 30215723-30215723 on Build GRCh38
rs778463287 Chr.21: 30215692-30215692 on Build GRCh38
rs752672377 Chr.21: 30215670-30215670 on Build GRCh38
rs757500351 Chr.21: 30215450-30215450 on Build GRCh38
rs686364 Chr.21: 30215475-30215475 on Build GRCh38
rs528758738 Chr.21: 30215741-30215741 on Build GRCh38
rs767367630 Chr.21: 30215750-30215750 on Build GRCh38
rs778285222 Chr.21: 30215498-30215498 on Build GRCh38
rs200097700 Chr.21: 30215679-30215679 on Build GRCh38
rs185171814 Chr.21: 30215582-30215582 on Build GRCh38
rs755377935 Chr.21: 30215797-30215797 on Build GRCh38
rs759813417 Chr.21: 30215819-30215819 on Build GRCh38
rs748525098 Chr.21: 30215803-30215803 on Build GRCh38
rs750983909 Chr.21: 30215796-30215797 on Build GRCh38
rs953112188 Chr.21: 30215813-30215813 on Build GRCh38
rs751327021 Chr.21: 30215642-30215642 on Build GRCh38
rs1032600168 Chr.21: 30215653-30215653 on Build GRCh38
rs994122095 Chr.21: 30215640-30215640 on Build GRCh38
rs772528868 Chr.21: 30215489-30215489 on Build GRCh38
rs796717455 Chr.21: 30215762-30215762 on Build GRCh38
rs201870752 Chr.21: 30215514-30215514 on Build GRCh38
rs765509944 Chr.21: 30215517-30215517 on Build GRCh38
rs201177228 Chr.21: 30215694-30215694 on Build GRCh38
rs769259130 Chr.21: 30215474-30215474 on Build GRCh38
rs747608175 Chr.21: 30215543-30215543 on Build GRCh38
rs778312220 Chr.21: 30215872-30215872 on Build GRCh38
rs770295035 Chr.21: 30215804-30215804 on Build GRCh38
rs774553337 Chr.21: 30215818-30215818 on Build GRCh38
rs373504217 Chr.21: 30215526-30215526 on Build GRCh38
rs138394812 Chr.21: 30215867-30215867 on Build GRCh38
rs975739466 Chr.21: 30215522-30215522 on Build GRCh38
rs375641409 Chr.21: 30215806-30215806 on Build GRCh38
rs747333331 Chr.21: 30215693-30215693 on Build GRCh38
rs770434883 Chr.21: 30215717-30215717 on Build GRCh38
rs531956336 Chr.21: 30215832-30215832 on Build GRCh38
rs749828592 Chr.21: 30215883-30215883 on Build GRCh38
rs772297302 Chr.21: 30215826-30215826 on Build GRCh38
rs568681833 Chr.21: 30215515-30215515 on Build GRCh38
rs760549552 Chr.21: 30215765-30215765 on Build GRCh38
rs543328603 Chr.21: 30215587-30215587 on Build GRCh38
rs777401334 Chr.21: 30215675-30215675 on Build GRCh38
rs769250676 Chr.21: 30215577-30215577 on Build GRCh38
rs561390736 Chr.21: 30215634-30215634 on Build GRCh38
rs145203913 Chr.21: 30215455-30215455 on Build GRCh38
rs770955180 Chr.21: 30215810-30215810 on Build GRCh38
rs867441012 Chr.21: 30215531-30215531 on Build GRCh38
rs779821920 Chr.21: 30215462-30215462 on Build GRCh38
rs759798441 Chr.21: 30215476-30215476 on Build GRCh38
rs747030038 Chr.21: 30215463-30215463 on Build GRCh38
rs758122461 Chr.21: 30215532-30215532 on Build GRCh38
rs949339666 Chr.21: 30215598-30215598 on Build GRCh38
rs764714350 Chr.21: 30215501-30215501 on Build GRCh38
rs753350719 Chr.21: 30215779-30215779 on Build GRCh38
rs150078407 Chr.21: 30215858-30215858 on Build GRCh38
rs764047012 Chr.21: 30215843-30215843 on Build GRCh38
rs1557294 Chr.21: 30215853-30215853 on Build GRCh38
rs778608955 Chr.21: 30215792-30215792 on Build GRCh38
rs774484538 Chr.21: 30215740-30215740 on Build GRCh38

View Full Product Details