Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32737019-32737515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189888 c.2610_2611insA p.S871fs*2 Chr.21: 32737279-32737280 on Build GRCh38
6738315 c.2591A>C p.Y864S Chr.21: 32737299-32737299 on Build GRCh38
6306220 c.2633C>A p.A878E Chr.21: 32737257-32737257 on Build GRCh38
1161403 c.2569G>A p.V857I Chr.21: 32737321-32737321 on Build GRCh38
1030240 c.2505A>C p.Q835H Chr.21: 32737385-32737385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769978056 Chr.21: 32737224-32737224 on Build GRCh38
rs760041666 Chr.21: 32737422-32737422 on Build GRCh38
rs761318734 Chr.21: 32737412-32737412 on Build GRCh38
rs779176388 Chr.21: 32737445-32737445 on Build GRCh38
rs544078703 Chr.21: 32737150-32737150 on Build GRCh38
rs947590297 Chr.21: 32737479-32737479 on Build GRCh38
rs551236608 Chr.21: 32737295-32737295 on Build GRCh38
rs745722467 Chr.21: 32737349-32737349 on Build GRCh38
rs746856382 Chr.21: 32737428-32737428 on Build GRCh38
rs563031399 Chr.21: 32737304-32737304 on Build GRCh38
rs370603447 Chr.21: 32737283-32737283 on Build GRCh38
rs760246998 Chr.21: 32737286-32737286 on Build GRCh38
rs1049444035 Chr.21: 32737307-32737307 on Build GRCh38
rs746060549 Chr.21: 32737446-32737446 on Build GRCh38
rs757348900 Chr.21: 32737441-32737441 on Build GRCh38
rs138511262 Chr.21: 32737329-32737329 on Build GRCh38
rs190439966 Chr.21: 32737195-32737195 on Build GRCh38
rs770804239 Chr.21: 32737242-32737242 on Build GRCh38
rs755335296 Chr.21: 32737317-32737317 on Build GRCh38
rs564128254 Chr.21: 32737130-32737130 on Build GRCh38
rs750084791 Chr.21: 32737417-32737417 on Build GRCh38
rs199811323 Chr.21: 32737334-32737334 on Build GRCh38
rs376033585 Chr.21: 32737415-32737415 on Build GRCh38
rs755227711 Chr.21: 32737417-32737420 on Build GRCh38
rs575981751 Chr.21: 32737113-32737113 on Build GRCh38
rs751976064 Chr.21: 32737310-32737310 on Build GRCh38
rs190913591 Chr.21: 32737425-32737425 on Build GRCh38
rs376912985 Chr.21: 32737365-32737365 on Build GRCh38
rs778279679 Chr.21: 32737438-32737438 on Build GRCh38
rs749337356 Chr.21: 32737332-32737332 on Build GRCh38
rs779077920 Chr.21: 32737427-32737431 on Build GRCh38
rs756597122 Chr.21: 32737432-32737432 on Build GRCh38
rs564083427 Chr.21: 32737079-32737079 on Build GRCh38
rs774050399 Chr.21: 32737247-32737247 on Build GRCh38
rs748249291 Chr.21: 32737220-32737220 on Build GRCh38
rs143382738 Chr.21: 32737271-32737271 on Build GRCh38
rs549339725 Chr.21: 32737458-32737458 on Build GRCh38
rs775040197 Chr.21: 32737376-32737376 on Build GRCh38
rs914738015 Chr.21: 32737452-32737452 on Build GRCh38
rs572714432 Chr.21: 32737162-32737162 on Build GRCh38
rs763482590 Chr.21: 32737293-32737293 on Build GRCh38
rs749435435 Chr.21: 32737234-32737234 on Build GRCh38
rs770259842 Chr.21: 32737321-32737321 on Build GRCh38
rs997492835 Chr.21: 32737111-32737111 on Build GRCh38
rs773546406 Chr.21: 32737225-32737225 on Build GRCh38
rs775447295 Chr.21: 32737394-32737394 on Build GRCh38
rs753881379 Chr.21: 32737439-32737439 on Build GRCh38
rs370348660 Chr.21: 32737401-32737401 on Build GRCh38
rs776036603 Chr.21: 32737405-32737405 on Build GRCh38
rs562272048 Chr.21: 32737253-32737253 on Build GRCh38
rs781736352 Chr.21: 32737214-32737214 on Build GRCh38
rs144102538 Chr.21: 32737403-32737403 on Build GRCh38
rs903174229 Chr.21: 32737106-32737106 on Build GRCh38
rs147145324 Chr.21: 32737319-32737319 on Build GRCh38
rs779053019 Chr.21: 32737347-32737347 on Build GRCh38
rs199655078 Chr.21: 32737257-32737257 on Build GRCh38
rs199507974 Chr.21: 32737330-32737330 on Build GRCh38
rs755573141 Chr.21: 32737210-32737210 on Build GRCh38
rs753868359 Chr.21: 32737217-32737218 on Build GRCh38
rs867702535 Chr.21: 32737285-32737285 on Build GRCh38
rs375035076 Chr.21: 32737426-32737426 on Build GRCh38

View Full Product Details