Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32793272-32793817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf62
Entrez Gene ID
56245
Gene Name
chromosome 21 open reading frame 62
Gene Aliases
B37, C21orf120, PRED81
UniGene
Hs.517235
Gene Chromosome Location
Chr.21: 32790673-32813808 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
UniGene
Hs.54725
Gene Chromosome Location
Chr.21: 32772100-32797707 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030255 c.626G>T p.S209I Chr.21: 32793796-32793796 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375024046 Chr.21: 32793649-32793649 on Build GRCh38
rs985635612 Chr.21: 32793592-32793592 on Build GRCh38
rs766964726 Chr.21: 32793647-32793647 on Build GRCh38
rs769143331 Chr.21: 32793698-32793698 on Build GRCh38
rs1021847273 Chr.21: 32793427-32793427 on Build GRCh38
rs113251491 Chr.21: 32793523-32793523 on Build GRCh38
rs781148411 Chr.21: 32793700-32793700 on Build GRCh38
rs372493378 Chr.21: 32793786-32793786 on Build GRCh38
rs548906741 Chr.21: 32793494-32793494 on Build GRCh38
rs948643899 Chr.21: 32793556-32793556 on Build GRCh38
rs759881459 Chr.21: 32793398-32793398 on Build GRCh38
rs770869916 Chr.21: 32793720-32793720 on Build GRCh38
rs1002568758 Chr.21: 32793358-32793358 on Build GRCh38
rs759495278 Chr.21: 32793646-32793646 on Build GRCh38
rs767430057 Chr.21: 32793741-32793741 on Build GRCh38
rs1044212648 Chr.21: 32793348-32793348 on Build GRCh38
rs371362639 Chr.21: 32793313-32793313 on Build GRCh38
rs751866590 Chr.21: 32793785-32793785 on Build GRCh38
rs751517863 Chr.21: 32793645-32793645 on Build GRCh38
rs62214287 Chr.21: 32793298-32793298 on Build GRCh38
rs941523038 Chr.21: 32793603-32793603 on Build GRCh38
rs773203324 Chr.21: 32793638-32793638 on Build GRCh38
rs1034151392 Chr.21: 32793462-32793462 on Build GRCh38
rs370140004 Chr.21: 32793639-32793639 on Build GRCh38
rs552831765 Chr.21: 32793587-32793587 on Build GRCh38
rs752201249 Chr.21: 32793748-32793749 on Build GRCh38
rs758740610 Chr.21: 32793668-32793668 on Build GRCh38
rs559343672 Chr.21: 32793403-32793403 on Build GRCh38
rs372600850 Chr.21: 32793686-32793686 on Build GRCh38
rs1052779189 Chr.21: 32793593-32793593 on Build GRCh38
rs529641762 Chr.21: 32793577-32793577 on Build GRCh38
rs747372360 Chr.21: 32793452-32793452 on Build GRCh38
rs540756086 Chr.21: 32793318-32793318 on Build GRCh38
rs1001335694 Chr.21: 32793435-32793435 on Build GRCh38
rs756864702 Chr.21: 32793781-32793781 on Build GRCh38
rs769981563 Chr.21: 32793713-32793713 on Build GRCh38
rs907104922 Chr.21: 32793410-32793410 on Build GRCh38
rs892878547 Chr.21: 32793467-32793467 on Build GRCh38
rs975730511 Chr.21: 32793527-32793527 on Build GRCh38
rs548971378 Chr.21: 32793522-32793522 on Build GRCh38
rs574473392 Chr.21: 32793576-32793576 on Build GRCh38
rs780348377 Chr.21: 32793685-32793685 on Build GRCh38
rs972797395 Chr.21: 32793513-32793513 on Build GRCh38
rs1011809864 Chr.21: 32793744-32793744 on Build GRCh38
rs937218888 Chr.21: 32793677-32793677 on Build GRCh38
rs1025694172 Chr.21: 32793712-32793712 on Build GRCh38
rs2231391 Chr.21: 32793553-32793553 on Build GRCh38
rs138552991 Chr.21: 32793631-32793631 on Build GRCh38
rs918687683 Chr.21: 32793516-32793516 on Build GRCh38
rs535418618 Chr.21: 32793740-32793740 on Build GRCh38
rs376124156 Chr.21: 32793715-32793715 on Build GRCh38
rs766456419 Chr.21: 32793641-32793641 on Build GRCh38
rs375164231 Chr.21: 32793753-32793753 on Build GRCh38
rs774246169 Chr.21: 32793730-32793730 on Build GRCh38
rs965566215 Chr.21: 32793492-32793492 on Build GRCh38
rs752207127 Chr.21: 32793745-32793745 on Build GRCh38
rs1020444296 Chr.21: 32793497-32793497 on Build GRCh38
rs773532965 Chr.21: 32793714-32793714 on Build GRCh38
rs778965115 Chr.21: 32793709-32793709 on Build GRCh38
rs897194296 Chr.21: 32793379-32793379 on Build GRCh38
rs755735083 Chr.21: 32793653-32793653 on Build GRCh38
rs907085280 Chr.21: 32793352-32793352 on Build GRCh38
rs570939971 Chr.21: 32793628-32793628 on Build GRCh38
rs1031411621 Chr.21: 32793370-32793370 on Build GRCh38
rs4817501 Chr.21: 32793468-32793468 on Build GRCh38
rs780403437 Chr.21: 32793783-32793783 on Build GRCh38
rs764728493 Chr.21: 32793716-32793716 on Build GRCh38
rs575904703 Chr.21: 32793790-32793790 on Build GRCh38
rs750827168 Chr.21: 32793667-32793667 on Build GRCh38
rs776735696 Chr.21: 32793626-32793626 on Build GRCh38
rs929996846 Chr.21: 32793655-32793655 on Build GRCh38
rs985586431 Chr.21: 32793453-32793453 on Build GRCh38
rs754023476 Chr.21: 32793386-32793386 on Build GRCh38
rs1011674294 Chr.21: 32793414-32793414 on Build GRCh38
rs748249766 Chr.21: 32793707-32793707 on Build GRCh38
rs1044323449 Chr.21: 32793557-32793557 on Build GRCh38
rs373448292 Chr.21: 32793388-32793388 on Build GRCh38
rs911355820 Chr.21: 32793602-32793602 on Build GRCh38
rs368511326 Chr.21: 32793656-32793656 on Build GRCh38
rs368281384 Chr.21: 32793763-32793763 on Build GRCh38
rs1038780205 Chr.21: 32793335-32793335 on Build GRCh38

View Full Product Details