Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32807300-32807802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf62
Entrez Gene ID
56245
Gene Name
chromosome 21 open reading frame 62
Gene Aliases
B37, C21orf120, PRED81
UniGene
Hs.517235
Gene Chromosome Location
Chr.21: 32790673-32813808 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 XM_011529653.2

SNP Detail

SNP ID Chromosome Location
rs1057050656 Chr.21: 32807537-32807537 on Build GRCh38
rs983238266 Chr.21: 32807574-32807574 on Build GRCh38
rs998589789 Chr.21: 32807604-32807604 on Build GRCh38
rs920104950 Chr.21: 32807493-32807493 on Build GRCh38
rs909846273 Chr.21: 32807437-32807437 on Build GRCh38
rs1031320294 Chr.21: 32807428-32807428 on Build GRCh38
rs964254141 Chr.21: 32807467-32807467 on Build GRCh38
rs904264899 Chr.21: 32807666-32807666 on Build GRCh38
rs999812497 Chr.21: 32807531-32807531 on Build GRCh38
rs554720066 Chr.21: 32807418-32807418 on Build GRCh38
rs547317632 Chr.21: 32807672-32807672 on Build GRCh38
rs1033658622 Chr.21: 32807766-32807766 on Build GRCh38
rs987023856 Chr.21: 32807457-32807457 on Build GRCh38
rs536038138 Chr.21: 32807408-32807408 on Build GRCh38
rs937276901 Chr.21: 32807495-32807495 on Build GRCh38
rs948667281 Chr.21: 32807735-32807735 on Build GRCh38
rs1054738957 Chr.21: 32807350-32807350 on Build GRCh38
rs912769443 Chr.21: 32807461-32807461 on Build GRCh38
rs938671815 Chr.21: 32807708-32807708 on Build GRCh38
rs7280327 Chr.21: 32807514-32807514 on Build GRCh38
rs1041978715 Chr.21: 32807664-32807664 on Build GRCh38
rs1021380810 Chr.21: 32807381-32807381 on Build GRCh38
rs887235593 Chr.21: 32807700-32807700 on Build GRCh38
rs199682608 Chr.21: 32807346-32807351 on Build GRCh38
rs1013124886 Chr.21: 32807366-32807366 on Build GRCh38
rs113656831 Chr.21: 32807655-32807655 on Build GRCh38
rs7279941 Chr.21: 32807425-32807425 on Build GRCh38
rs543463865 Chr.21: 32807468-32807468 on Build GRCh38
rs552638073 Chr.21: 32807610-32807610 on Build GRCh38
rs11701881 Chr.21: 32807721-32807721 on Build GRCh38
rs904377416 Chr.21: 32807750-32807750 on Build GRCh38
rs975674276 Chr.21: 32807671-32807671 on Build GRCh38
rs148570120 Chr.21: 32807578-32807578 on Build GRCh38
rs149581979 Chr.21: 32807364-32807364 on Build GRCh38
rs980761779 Chr.21: 32807600-32807600 on Build GRCh38
rs564180419 Chr.21: 32807651-32807651 on Build GRCh38
rs963161878 Chr.21: 32807593-32807593 on Build GRCh38
rs941379341 Chr.21: 32807454-32807454 on Build GRCh38
rs7280333 Chr.21: 32807525-32807525 on Build GRCh38
rs796818918 Chr.21: 32807605-32807605 on Build GRCh38
rs530391866 Chr.21: 32807530-32807530 on Build GRCh38
rs998365709 Chr.21: 32807688-32807690 on Build GRCh38
rs952828414 Chr.21: 32807674-32807674 on Build GRCh38
rs894349069 Chr.21: 32807544-32807544 on Build GRCh38
rs189548739 Chr.21: 32807517-32807517 on Build GRCh38
rs998416338 Chr.21: 32807552-32807552 on Build GRCh38
rs9808743 Chr.21: 32807553-32807553 on Build GRCh38
rs569785723 Chr.21: 32807727-32807727 on Build GRCh38
rs114883385 Chr.21: 32807736-32807736 on Build GRCh38
rs931410059 Chr.21: 32807494-32807494 on Build GRCh38
rs536327081 Chr.21: 32807545-32807545 on Build GRCh38
rs964686811 Chr.21: 32807656-32807656 on Build GRCh38
rs1008486041 Chr.21: 32807626-32807626 on Build GRCh38
rs1004381891 Chr.21: 32807705-32807705 on Build GRCh38
rs541784602 Chr.21: 32807524-32807524 on Build GRCh38
rs10688715 Chr.21: 32807346-32807347 on Build GRCh38
rs954362027 Chr.21: 32807433-32807433 on Build GRCh38
rs563867545 Chr.21: 32807572-32807572 on Build GRCh38
rs746187197 Chr.21: 32807350-32807351 on Build GRCh38
rs781370992 Chr.21: 32807685-32807685 on Build GRCh38
rs976022098 Chr.21: 32807491-32807491 on Build GRCh38
rs558587616 Chr.21: 32807507-32807507 on Build GRCh38
rs992755275 Chr.21: 32807647-32807647 on Build GRCh38
rs75774503 Chr.21: 32807699-32807699 on Build GRCh38
rs528553368 Chr.21: 32807334-32807335 on Build GRCh38
rs973277698 Chr.21: 32807483-32807483 on Build GRCh38
rs182208542 Chr.21: 32807723-32807723 on Build GRCh38

View Full Product Details