Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33517265-33517765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GART
Entrez Gene ID
2618
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
Gene Aliases
AIRS, GARS, GARTF, PAIS, PGFT, PRGS
UniGene
Hs.473648
Gene Chromosome Location
Chr.21: 33503931-33542917 on Build GRCh38
Transcripts
NM_000819.4 NM_001136005.1 NM_001136006.1 XM_005260941.1 XM_006723989.1 XM_006723990.1 XM_011529526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5858541 c.1784G>T p.R595L Chr.21: 33517527-33517527 on Build GRCh38
1030311 c.1890C>T p.I630I Chr.21: 33517421-33517421 on Build GRCh38
6669358 c.1739delC p.P580fs*6 Chr.21: 33517572-33517572 on Build GRCh38
1030310 c.1901C>A p.S634Y Chr.21: 33517410-33517410 on Build GRCh38
4644331 c.1803C>T p.H601H Chr.21: 33517508-33517508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041481437 Chr.21: 33517385-33517385 on Build GRCh38
rs759109728 Chr.21: 33517534-33517534 on Build GRCh38
rs145056428 Chr.21: 33517396-33517396 on Build GRCh38
rs139548106 Chr.21: 33517384-33517384 on Build GRCh38
rs776471333 Chr.21: 33517371-33517371 on Build GRCh38
rs907978516 Chr.21: 33517553-33517553 on Build GRCh38
rs151206034 Chr.21: 33517581-33517581 on Build GRCh38
rs867943062 Chr.21: 33517720-33517720 on Build GRCh38
rs557882403 Chr.21: 33517509-33517509 on Build GRCh38
rs375498243 Chr.21: 33517421-33517421 on Build GRCh38
rs754723198 Chr.21: 33517407-33517407 on Build GRCh38
rs747450439 Chr.21: 33517574-33517574 on Build GRCh38
rs771494572 Chr.21: 33517474-33517476 on Build GRCh38
rs752334753 Chr.21: 33517302-33517302 on Build GRCh38
rs762639082 Chr.21: 33517611-33517611 on Build GRCh38
rs971886992 Chr.21: 33517470-33517470 on Build GRCh38
rs548039784 Chr.21: 33517695-33517698 on Build GRCh38
rs142142742 Chr.21: 33517420-33517420 on Build GRCh38
rs572992301 Chr.21: 33517409-33517409 on Build GRCh38
rs953254450 Chr.21: 33517697-33517697 on Build GRCh38
rs766393455 Chr.21: 33517533-33517533 on Build GRCh38
rs1018659221 Chr.21: 33517710-33517710 on Build GRCh38
rs954750300 Chr.21: 33517463-33517463 on Build GRCh38
rs372112153 Chr.21: 33517492-33517492 on Build GRCh38
rs528566554 Chr.21: 33517692-33517692 on Build GRCh38
rs368440911 Chr.21: 33517482-33517482 on Build GRCh38
rs748437484 Chr.21: 33517582-33517582 on Build GRCh38
rs377766009 Chr.21: 33517572-33517572 on Build GRCh38
rs374385313 Chr.21: 33517544-33517544 on Build GRCh38
rs773106095 Chr.21: 33517607-33517607 on Build GRCh38
rs373456316 Chr.21: 33517640-33517640 on Build GRCh38
rs143711739 Chr.21: 33517644-33517644 on Build GRCh38
rs59920090 Chr.21: 33517416-33517416 on Build GRCh38
rs1041759249 Chr.21: 33517737-33517737 on Build GRCh38
rs373647982 Chr.21: 33517339-33517339 on Build GRCh38
rs202015633 Chr.21: 33517527-33517527 on Build GRCh38
rs546649833 Chr.21: 33517719-33517719 on Build GRCh38
rs761668054 Chr.21: 33517380-33517380 on Build GRCh38
rs202108641 Chr.21: 33517580-33517580 on Build GRCh38
rs764282183 Chr.21: 33517364-33517364 on Build GRCh38
rs760413225 Chr.21: 33517642-33517642 on Build GRCh38
rs768740452 Chr.21: 33517575-33517575 on Build GRCh38
rs987126373 Chr.21: 33517701-33517701 on Build GRCh38
rs776843173 Chr.21: 33517503-33517503 on Build GRCh38
rs961936132 Chr.21: 33517716-33517716 on Build GRCh38
rs368955911 Chr.21: 33517528-33517528 on Build GRCh38
rs146301480 Chr.21: 33517358-33517358 on Build GRCh38
rs34588874 Chr.21: 33517390-33517390 on Build GRCh38
rs774913516 Chr.21: 33517326-33517326 on Build GRCh38
rs777620272 Chr.21: 33517399-33517399 on Build GRCh38
rs779364102 Chr.21: 33517566-33517566 on Build GRCh38
rs774430332 Chr.21: 33517629-33517629 on Build GRCh38
rs769656472 Chr.21: 33517525-33517525 on Build GRCh38
rs747078669 Chr.21: 33517491-33517491 on Build GRCh38
rs767157521 Chr.21: 33517633-33517633 on Build GRCh38
rs750734009 Chr.21: 33517567-33517567 on Build GRCh38
rs755672047 Chr.21: 33517563-33517563 on Build GRCh38
rs557676685 Chr.21: 33517328-33517328 on Build GRCh38
rs144089530 Chr.21: 33517677-33517677 on Build GRCh38
rs771055876 Chr.21: 33517622-33517622 on Build GRCh38
rs569448040 Chr.21: 33517333-33517333 on Build GRCh38
rs972001515 Chr.21: 33517739-33517739 on Build GRCh38
rs769878704 Chr.21: 33517587-33517587 on Build GRCh38
rs758130829 Chr.21: 33517344-33517344 on Build GRCh38
rs759488956 Chr.21: 33517630-33517630 on Build GRCh38
rs573920101 Chr.21: 33517446-33517446 on Build GRCh38
rs996623024 Chr.21: 33517682-33517682 on Build GRCh38
rs779114577 Chr.21: 33517645-33517645 on Build GRCh38
rs372179641 Chr.21: 33517389-33517389 on Build GRCh38
rs772727139 Chr.21: 33517642-33517643 on Build GRCh38
rs1056400467 Chr.21: 33517565-33517565 on Build GRCh38
rs772416090 Chr.21: 33517457-33517457 on Build GRCh38
rs544071183 Chr.21: 33517543-33517543 on Build GRCh38
rs748011138 Chr.21: 33517332-33517333 on Build GRCh38

View Full Product Details