Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33577737-33578269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

SNP Detail

SNP ID Chromosome Location
rs967762213 Chr.21: 33578193-33578193 on Build GRCh38
rs1051607965 Chr.21: 33578035-33578035 on Build GRCh38
rs1035260416 Chr.21: 33577885-33577885 on Build GRCh38
rs199567196 Chr.21: 33577854-33577854 on Build GRCh38
rs185644674 Chr.21: 33577911-33577911 on Build GRCh38
rs111856399 Chr.21: 33577861-33577861 on Build GRCh38
rs557571362 Chr.21: 33577966-33577966 on Build GRCh38
rs956406367 Chr.21: 33578168-33578168 on Build GRCh38
rs1016856952 Chr.21: 33578096-33578096 on Build GRCh38
rs139380929 Chr.21: 33578118-33578118 on Build GRCh38
rs962894434 Chr.21: 33577905-33577905 on Build GRCh38
rs1000162766 Chr.21: 33578202-33578203 on Build GRCh38
rs750934956 Chr.21: 33578101-33578105 on Build GRCh38
rs1046831628 Chr.21: 33577778-33577778 on Build GRCh38
rs922754788 Chr.21: 33578060-33578060 on Build GRCh38
rs781444328 Chr.21: 33578166-33578166 on Build GRCh38
rs546376082 Chr.21: 33577908-33577908 on Build GRCh38
rs978773088 Chr.21: 33577959-33577959 on Build GRCh38
rs975721093 Chr.21: 33578200-33578200 on Build GRCh38
rs141846839 Chr.21: 33578047-33578047 on Build GRCh38
rs1003336764 Chr.21: 33577851-33577851 on Build GRCh38
rs753970746 Chr.21: 33577912-33577915 on Build GRCh38
rs971609616 Chr.21: 33577925-33577925 on Build GRCh38
rs755800440 Chr.21: 33577864-33577864 on Build GRCh38
rs927728754 Chr.21: 33577965-33577965 on Build GRCh38
rs181896718 Chr.21: 33577817-33577817 on Build GRCh38
rs1017138148 Chr.21: 33577898-33577898 on Build GRCh38
rs899675041 Chr.21: 33577820-33577820 on Build GRCh38
rs546312943 Chr.21: 33578146-33578146 on Build GRCh38
rs1005193067 Chr.21: 33577797-33577797 on Build GRCh38
rs766987443 Chr.21: 33577785-33577785 on Build GRCh38
rs76002527 Chr.21: 33577913-33577913 on Build GRCh38
rs946585556 Chr.21: 33577986-33577986 on Build GRCh38
rs893764491 Chr.21: 33577774-33577774 on Build GRCh38
rs571040026 Chr.21: 33578107-33578108 on Build GRCh38
rs113014293 Chr.21: 33578025-33578025 on Build GRCh38
rs776350188 Chr.21: 33578178-33578178 on Build GRCh38
rs930173792 Chr.21: 33578102-33578102 on Build GRCh38
rs982251253 Chr.21: 33577929-33577929 on Build GRCh38
rs933815661 Chr.21: 33578244-33578244 on Build GRCh38
rs200055284 Chr.21: 33577851-33577854 on Build GRCh38
rs987847839 Chr.21: 33577968-33577968 on Build GRCh38
rs568067672 Chr.21: 33577780-33577780 on Build GRCh38
rs190203784 Chr.21: 33577996-33577996 on Build GRCh38
rs753730801 Chr.21: 33577976-33577976 on Build GRCh38
rs1004256463 Chr.21: 33578084-33578084 on Build GRCh38
rs998653107 Chr.21: 33577859-33577859 on Build GRCh38
rs1003078348 Chr.21: 33578150-33578150 on Build GRCh38
rs148105472 Chr.21: 33577847-33577847 on Build GRCh38
rs868061978 Chr.21: 33578153-33578153 on Build GRCh38
rs146333314 Chr.21: 33578116-33578116 on Build GRCh38
rs113758763 Chr.21: 33577934-33577934 on Build GRCh38
rs1003860569 Chr.21: 33578125-33578125 on Build GRCh38
rs987899844 Chr.21: 33578183-33578183 on Build GRCh38
rs887095771 Chr.21: 33577871-33577871 on Build GRCh38
rs921223944 Chr.21: 33578204-33578204 on Build GRCh38
rs532257913 Chr.21: 33578189-33578189 on Build GRCh38
rs143316514 Chr.21: 33578135-33578135 on Build GRCh38
rs1004202252 Chr.21: 33577872-33577872 on Build GRCh38
rs747332747 Chr.21: 33578165-33578166 on Build GRCh38
rs190151919 Chr.21: 33577782-33577782 on Build GRCh38
rs1046775004 Chr.21: 33578106-33578107 on Build GRCh38
rs933866883 Chr.21: 33578031-33578031 on Build GRCh38
rs887231731 Chr.21: 33578072-33578072 on Build GRCh38
rs185178624 Chr.21: 33578115-33578115 on Build GRCh38
rs749883164 Chr.21: 33577830-33577830 on Build GRCh38

View Full Product Details