Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33589512-33590012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3799872 c.1025G>C p.R342T Chr.21: 33589847-33589847 on Build GRCh38
2844062 c.976C>T p.L326L Chr.21: 33589896-33589896 on Build GRCh38
4101384 c.1004C>T p.A335V Chr.21: 33589868-33589868 on Build GRCh38
5957050 c.967C>A p.P323T Chr.21: 33589905-33589905 on Build GRCh38
3668521 c.961G>A p.D321N Chr.21: 33589911-33589911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772766101 Chr.21: 33589789-33589789 on Build GRCh38
rs11548181 Chr.21: 33589562-33589562 on Build GRCh38
rs148353071 Chr.21: 33589704-33589704 on Build GRCh38
rs530452800 Chr.21: 33589752-33589752 on Build GRCh38
rs779565849 Chr.21: 33589852-33589855 on Build GRCh38
rs543916907 Chr.21: 33589588-33589588 on Build GRCh38
rs563296479 Chr.21: 33589746-33589746 on Build GRCh38
rs763776784 Chr.21: 33589663-33589663 on Build GRCh38
rs77053739 Chr.21: 33589785-33589785 on Build GRCh38
rs747840611 Chr.21: 33589784-33589784 on Build GRCh38
rs776011182 Chr.21: 33589803-33589803 on Build GRCh38
rs371455049 Chr.21: 33589834-33589834 on Build GRCh38
rs762420783 Chr.21: 33589794-33589794 on Build GRCh38
rs768592710 Chr.21: 33589781-33589781 on Build GRCh38
rs769466510 Chr.21: 33589917-33589917 on Build GRCh38
rs754116746 Chr.21: 33589733-33589733 on Build GRCh38
rs145158429 Chr.21: 33589888-33589888 on Build GRCh38
rs779041997 Chr.21: 33589741-33589741 on Build GRCh38
rs757230148 Chr.21: 33589737-33589737 on Build GRCh38
rs542522364 Chr.21: 33589863-33589863 on Build GRCh38
rs143362717 Chr.21: 33589603-33589603 on Build GRCh38
rs536554932 Chr.21: 33589684-33589684 on Build GRCh38
rs867837110 Chr.21: 33589797-33589797 on Build GRCh38
rs1044784840 Chr.21: 33589856-33589856 on Build GRCh38
rs986798382 Chr.21: 33589693-33589693 on Build GRCh38
rs780089134 Chr.21: 33589884-33589884 on Build GRCh38
rs777332315 Chr.21: 33589929-33589929 on Build GRCh38
rs745989885 Chr.21: 33589742-33589742 on Build GRCh38
rs764459990 Chr.21: 33589813-33589813 on Build GRCh38
rs535374663 Chr.21: 33589951-33589951 on Build GRCh38
rs767983397 Chr.21: 33589796-33589796 on Build GRCh38
rs753461095 Chr.21: 33589958-33589963 on Build GRCh38
rs3761349 Chr.21: 33589660-33589660 on Build GRCh38
rs1046312 Chr.21: 33589659-33589659 on Build GRCh38
rs879200015 Chr.21: 33589541-33589541 on Build GRCh38
rs1009388888 Chr.21: 33589635-33589635 on Build GRCh38
rs754753495 Chr.21: 33589905-33589905 on Build GRCh38
rs1034860700 Chr.21: 33589611-33589611 on Build GRCh38
rs905873724 Chr.21: 33589666-33589666 on Build GRCh38
rs922436202 Chr.21: 33589606-33589606 on Build GRCh38
rs912993018 Chr.21: 33589556-33589556 on Build GRCh38
rs750482662 Chr.21: 33589846-33589846 on Build GRCh38
rs752310275 Chr.21: 33589546-33589546 on Build GRCh38
rs1028751478 Chr.21: 33589676-33589676 on Build GRCh38
rs376870432 Chr.21: 33589620-33589620 on Build GRCh38
rs181673515 Chr.21: 33589828-33589828 on Build GRCh38
rs201105437 Chr.21: 33589838-33589838 on Build GRCh38
rs552133521 Chr.21: 33589791-33589791 on Build GRCh38
rs534712308 Chr.21: 33589829-33589829 on Build GRCh38
rs751189695 Chr.21: 33589738-33589738 on Build GRCh38
rs898942628 Chr.21: 33589553-33589553 on Build GRCh38
rs760982632 Chr.21: 33589807-33589807 on Build GRCh38
rs968421962 Chr.21: 33589808-33589808 on Build GRCh38
rs978395562 Chr.21: 33589587-33589587 on Build GRCh38
rs376636520 Chr.21: 33589910-33589910 on Build GRCh38
rs748759115 Chr.21: 33589936-33589936 on Build GRCh38
rs188922485 Chr.21: 33589569-33589569 on Build GRCh38
rs746759081 Chr.21: 33589757-33589757 on Build GRCh38
rs200628535 Chr.21: 33589868-33589868 on Build GRCh38
rs369802759 Chr.21: 33589968-33589968 on Build GRCh38
rs904826252 Chr.21: 33589593-33589593 on Build GRCh38
rs747952229 Chr.21: 33589912-33589912 on Build GRCh38
rs776089798 Chr.21: 33589783-33589783 on Build GRCh38
rs761179022 Chr.21: 33589973-33589973 on Build GRCh38
rs1047807750 Chr.21: 33589622-33589622 on Build GRCh38
rs375676572 Chr.21: 33589980-33589980 on Build GRCh38

View Full Product Details