Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33599024-33599531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4101386 c.650G>T p.G217V Chr.21: 33599176-33599176 on Build GRCh38
6526246 c.631G>T p.E211* Chr.21: 33599195-33599195 on Build GRCh38
4101385 c.655G>A p.D219N Chr.21: 33599171-33599171 on Build GRCh38
4101388 c.578C>T p.A193V Chr.21: 33599248-33599248 on Build GRCh38
4101387 c.619G>A p.E207K Chr.21: 33599207-33599207 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779038433 Chr.21: 33599214-33599214 on Build GRCh38
rs768263017 Chr.21: 33599255-33599255 on Build GRCh38
rs112659899 Chr.21: 33599488-33599488 on Build GRCh38
rs1049777195 Chr.21: 33599167-33599167 on Build GRCh38
rs761887737 Chr.21: 33599332-33599335 on Build GRCh38
rs1008490508 Chr.21: 33599225-33599225 on Build GRCh38
rs749392156 Chr.21: 33599198-33599198 on Build GRCh38
rs772000544 Chr.21: 33599217-33599217 on Build GRCh38
rs181993996 Chr.21: 33599154-33599154 on Build GRCh38
rs777893406 Chr.21: 33599196-33599196 on Build GRCh38
rs970692949 Chr.21: 33599084-33599085 on Build GRCh38
rs759828788 Chr.21: 33599306-33599306 on Build GRCh38
rs771824686 Chr.21: 33599129-33599129 on Build GRCh38
rs766837748 Chr.21: 33599156-33599156 on Build GRCh38
rs766861602 Chr.21: 33599287-33599287 on Build GRCh38
rs746648810 Chr.21: 33599246-33599246 on Build GRCh38
rs995181708 Chr.21: 33599279-33599279 on Build GRCh38
rs200356570 Chr.21: 33599164-33599164 on Build GRCh38
rs763676367 Chr.21: 33599139-33599139 on Build GRCh38
rs368922716 Chr.21: 33599155-33599155 on Build GRCh38
rs774252592 Chr.21: 33599167-33599168 on Build GRCh38
rs138988095 Chr.21: 33599163-33599163 on Build GRCh38
rs745693377 Chr.21: 33599128-33599128 on Build GRCh38
rs748178422 Chr.21: 33599101-33599101 on Build GRCh38
rs376830789 Chr.21: 33599269-33599269 on Build GRCh38
rs770828350 Chr.21: 33599125-33599125 on Build GRCh38
rs1026681833 Chr.21: 33599289-33599289 on Build GRCh38
rs540623489 Chr.21: 33599451-33599451 on Build GRCh38
rs76733259 Chr.21: 33599103-33599103 on Build GRCh38
rs1014276676 Chr.21: 33599056-33599056 on Build GRCh38
rs756088252 Chr.21: 33599194-33599194 on Build GRCh38
rs917824121 Chr.21: 33599059-33599059 on Build GRCh38
rs753071782 Chr.21: 33599470-33599470 on Build GRCh38
rs941220965 Chr.21: 33599504-33599504 on Build GRCh38
rs775229467 Chr.21: 33599224-33599224 on Build GRCh38
rs61735780 Chr.21: 33599168-33599168 on Build GRCh38
rs745605421 Chr.21: 33599216-33599216 on Build GRCh38
rs752919932 Chr.21: 33599182-33599182 on Build GRCh38
rs777503408 Chr.21: 33599106-33599106 on Build GRCh38
rs897321965 Chr.21: 33599235-33599235 on Build GRCh38
rs775317724 Chr.21: 33599131-33599131 on Build GRCh38
rs376507121 Chr.21: 33599133-33599133 on Build GRCh38
rs766465927 Chr.21: 33599108-33599108 on Build GRCh38
rs185167943 Chr.21: 33599355-33599355 on Build GRCh38
rs765488111 Chr.21: 33599396-33599396 on Build GRCh38
rs1011848616 Chr.21: 33599460-33599460 on Build GRCh38
rs767879423 Chr.21: 33599329-33599329 on Build GRCh38
rs749260127 Chr.21: 33599107-33599107 on Build GRCh38
rs372289371 Chr.21: 33599227-33599227 on Build GRCh38
rs977631547 Chr.21: 33599122-33599122 on Build GRCh38
rs929655428 Chr.21: 33599343-33599343 on Build GRCh38
rs541296743 Chr.21: 33599209-33599209 on Build GRCh38
rs758759727 Chr.21: 33599498-33599498 on Build GRCh38
rs768598739 Chr.21: 33599166-33599166 on Build GRCh38
rs774870409 Chr.21: 33599288-33599288 on Build GRCh38
rs1046999751 Chr.21: 33599423-33599423 on Build GRCh38
rs867205185 Chr.21: 33599506-33599506 on Build GRCh38
rs528448368 Chr.21: 33599366-33599366 on Build GRCh38
rs560475066 Chr.21: 33599273-33599273 on Build GRCh38
rs767686465 Chr.21: 33599181-33599181 on Build GRCh38

View Full Product Details