Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36069929-36070419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR1
Entrez Gene ID
873
Gene Name
carbonyl reductase 1
Gene Aliases
CBR, SDR21C1, hCBR1
UniGene
Hs.88778
Gene Chromosome Location
Chr.21: 36069924-36073177 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Gene Symbol
LOC100133286
Entrez Gene ID
100133286
Gene Name
uncharacterized LOC100133286
Gene Aliases
-
UniGene
Hs.728809
Gene Chromosome Location
Chr.21: 36069642-36126640 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389990 c.267C>T p.N89N Chr.21: 36070382-36070382 on Build GRCh38
7220713 c.273G>A p.A91A Chr.21: 36070388-36070388 on Build GRCh38
5974224 c.105C>T p.L35L Chr.21: 36070220-36070220 on Build GRCh38
382250 c.136G>T p.A46S Chr.21: 36070251-36070251 on Build GRCh38
2844382 c.266A>G p.N89S Chr.21: 36070381-36070381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777221378 Chr.21: 36070357-36070357 on Build GRCh38
rs746699955 Chr.21: 36070226-36070226 on Build GRCh38
rs1012466885 Chr.21: 36070154-36070156 on Build GRCh38
rs757086930 Chr.21: 36070207-36070207 on Build GRCh38
rs151043730 Chr.21: 36070360-36070360 on Build GRCh38
rs781129548 Chr.21: 36069970-36069970 on Build GRCh38
rs750447177 Chr.21: 36069967-36069968 on Build GRCh38
rs763746901 Chr.21: 36070156-36070156 on Build GRCh38
rs201829813 Chr.21: 36069962-36069963 on Build GRCh38
rs777670435 Chr.21: 36070118-36070118 on Build GRCh38
rs994064884 Chr.21: 36069988-36069988 on Build GRCh38
rs767914019 Chr.21: 36070249-36070249 on Build GRCh38
rs200223739 Chr.21: 36070254-36070254 on Build GRCh38
rs536395967 Chr.21: 36069957-36069987 on Build GRCh38
rs766432246 Chr.21: 36070344-36070344 on Build GRCh38
rs751812271 Chr.21: 36070170-36070170 on Build GRCh38
rs1036385951 Chr.21: 36070115-36070118 on Build GRCh38
rs756250127 Chr.21: 36070103-36070103 on Build GRCh38
rs1029025368 Chr.21: 36069956-36069956 on Build GRCh38
rs745692534 Chr.21: 36070141-36070141 on Build GRCh38
rs559267091 Chr.21: 36070338-36070338 on Build GRCh38
rs760841760 Chr.21: 36070261-36070261 on Build GRCh38
rs968775154 Chr.21: 36069948-36069948 on Build GRCh38
rs758236840 Chr.21: 36070285-36070285 on Build GRCh38
rs766929443 Chr.21: 36070167-36070167 on Build GRCh38
rs117272030 Chr.21: 36070336-36070336 on Build GRCh38
rs944810092 Chr.21: 36070322-36070322 on Build GRCh38
rs757487232 Chr.21: 36069956-36069959 on Build GRCh38
rs555028304 Chr.21: 36070302-36070302 on Build GRCh38
rs768497282 Chr.21: 36069965-36069965 on Build GRCh38
rs768453981 Chr.21: 36070227-36070227 on Build GRCh38
rs11542168 Chr.21: 36070068-36070068 on Build GRCh38
rs25678 Chr.21: 36070334-36070334 on Build GRCh38
rs1023783646 Chr.21: 36070022-36070022 on Build GRCh38
rs777679109 Chr.21: 36070199-36070199 on Build GRCh38
rs566688474 Chr.21: 36070138-36070138 on Build GRCh38
rs576524713 Chr.21: 36070313-36070313 on Build GRCh38
rs776688059 Chr.21: 36070381-36070381 on Build GRCh38
rs41540715 Chr.21: 36070298-36070298 on Build GRCh38
rs989682886 Chr.21: 36070059-36070059 on Build GRCh38
rs142778878 Chr.21: 36070343-36070343 on Build GRCh38
rs758768231 Chr.21: 36070082-36070082 on Build GRCh38
rs1143663 Chr.21: 36070377-36070377 on Build GRCh38
rs911814829 Chr.21: 36069991-36069991 on Build GRCh38
rs897824559 Chr.21: 36070119-36070119 on Build GRCh38
rs369228069 Chr.21: 36070125-36070125 on Build GRCh38
rs191556963 Chr.21: 36070388-36070388 on Build GRCh38
rs987469756 Chr.21: 36069981-36069981 on Build GRCh38
rs146570341 Chr.21: 36070182-36070182 on Build GRCh38
rs371089618 Chr.21: 36070129-36070129 on Build GRCh38
rs762056579 Chr.21: 36070385-36070385 on Build GRCh38
rs773365505 Chr.21: 36070390-36070390 on Build GRCh38
rs779831065 Chr.21: 36070224-36070224 on Build GRCh38
rs769114657 Chr.21: 36070364-36070364 on Build GRCh38
rs748156211 Chr.21: 36070102-36070102 on Build GRCh38
rs753977576 Chr.21: 36070201-36070201 on Build GRCh38
rs751331798 Chr.21: 36070349-36070349 on Build GRCh38
rs769401188 Chr.21: 36070310-36070310 on Build GRCh38
rs780692500 Chr.21: 36070352-36070352 on Build GRCh38
rs768928349 Chr.21: 36070304-36070304 on Build GRCh38
rs754868460 Chr.21: 36070351-36070351 on Build GRCh38
rs539224993 Chr.21: 36070148-36070148 on Build GRCh38
rs930610071 Chr.21: 36070346-36070346 on Build GRCh38
rs772592919 Chr.21: 36070328-36070328 on Build GRCh38
rs533734780 Chr.21: 36070271-36070271 on Build GRCh38
rs772053364 Chr.21: 36070217-36070217 on Build GRCh38
rs930462697 Chr.21: 36070031-36070031 on Build GRCh38
rs532348271 Chr.21: 36070009-36070009 on Build GRCh38
rs746822383 Chr.21: 36070297-36070297 on Build GRCh38
rs569886457 Chr.21: 36070046-36070046 on Build GRCh38
rs536331017 Chr.21: 36070263-36070263 on Build GRCh38
rs778704295 Chr.21: 36070275-36070275 on Build GRCh38
rs745656182 Chr.21: 36070209-36070209 on Build GRCh38
rs867212750 Chr.21: 36069977-36069977 on Build GRCh38
rs756161777 Chr.21: 36070196-36070196 on Build GRCh38
rs766413880 Chr.21: 36070391-36070391 on Build GRCh38
rs766693597 Chr.21: 36070083-36070083 on Build GRCh38
rs972016400 Chr.21: 36070262-36070262 on Build GRCh38
rs558896125 Chr.21: 36070332-36070332 on Build GRCh38
rs143215551 Chr.21: 36070134-36070134 on Build GRCh38
rs915420485 Chr.21: 36070062-36070062 on Build GRCh38
rs759895472 Chr.21: 36070172-36070172 on Build GRCh38
rs748457628 Chr.21: 36069963-36069993 on Build GRCh38
rs748009980 Chr.21: 36069971-36069971 on Build GRCh38
rs77573804 Chr.21: 36070098-36070098 on Build GRCh38
rs11542167 Chr.21: 36070286-36070286 on Build GRCh38
rs775955102 Chr.21: 36070228-36070228 on Build GRCh38
rs1000335693 Chr.21: 36070386-36070386 on Build GRCh38
rs373500404 Chr.21: 36070100-36070100 on Build GRCh38
rs41549312 Chr.21: 36070042-36070042 on Build GRCh38
rs200695117 Chr.21: 36070359-36070359 on Build GRCh38
rs773738111 Chr.21: 36070160-36070160 on Build GRCh38
rs756016071 Chr.21: 36069977-36069984 on Build GRCh38
rs1009289627 Chr.21: 36069954-36069954 on Build GRCh38
rs890506301 Chr.21: 36070094-36070094 on Build GRCh38
rs869145162 Chr.21: 36069959-36069989 on Build GRCh38
rs529825233 Chr.21: 36070356-36070356 on Build GRCh38
rs978855172 Chr.21: 36069955-36069955 on Build GRCh38
rs750434431 Chr.21: 36070280-36070280 on Build GRCh38
rs1003256032 Chr.21: 36070369-36070369 on Build GRCh38
rs571566347 Chr.21: 36070142-36070142 on Build GRCh38
rs540823495 Chr.21: 36070010-36070010 on Build GRCh38
rs747881279 Chr.21: 36070355-36070355 on Build GRCh38
rs370920023 Chr.21: 36070305-36070305 on Build GRCh38
rs375697371 Chr.21: 36070238-36070238 on Build GRCh38
rs1048322369 Chr.21: 36070348-36070348 on Build GRCh38
rs761398228 Chr.21: 36070233-36070233 on Build GRCh38
rs9979814 Chr.21: 36070307-36070307 on Build GRCh38
rs775778302 Chr.21: 36070329-36070329 on Build GRCh38
rs566205268 Chr.21: 36070208-36070208 on Build GRCh38
rs920390177 Chr.21: 36070030-36070030 on Build GRCh38
rs951888249 Chr.21: 36070288-36070288 on Build GRCh38
rs890463460 Chr.21: 36069952-36069952 on Build GRCh38
rs377645409 Chr.21: 36070101-36070101 on Build GRCh38
rs904733513 Chr.21: 36070116-36070116 on Build GRCh38
rs1023456554 Chr.21: 36070161-36070161 on Build GRCh38
rs559448728 Chr.21: 36070011-36070011 on Build GRCh38
rs759878913 Chr.21: 36070242-36070242 on Build GRCh38
rs749509888 Chr.21: 36070206-36070207 on Build GRCh38
rs779953785 Chr.21: 36070294-36070294 on Build GRCh38
rs763329185 Chr.21: 36070162-36070162 on Build GRCh38
rs780185893 Chr.21: 36070097-36070098 on Build GRCh38
rs751985377 Chr.21: 36070088-36070088 on Build GRCh38
rs529746829 Chr.21: 36070020-36070020 on Build GRCh38
rs1030669258 Chr.21: 36070384-36070384 on Build GRCh38
rs760249754 Chr.21: 36070151-36070151 on Build GRCh38
rs752970795 Chr.21: 36070185-36070185 on Build GRCh38
rs367857649 Chr.21: 36070272-36070272 on Build GRCh38
rs543800905 Chr.21: 36070321-36070321 on Build GRCh38

View Full Product Details