Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36200331-36200873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY2
Entrez Gene ID
9980
Gene Name
dopey family member 2
Gene Aliases
21orf5, C21orf5
UniGene
Hs.204575
Gene Chromosome Location
Chr.21: 36156781-36294274 on Build GRCh38
Transcripts
NM_001320714.1 NM_005128.3 XM_017028509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319985 c.325G>T p.G109W Chr.21: 36200335-36200335 on Build GRCh38
5858586 c.376C>T p.L126F Chr.21: 36200386-36200386 on Build GRCh38
4439161 c.359C>T p.S120L Chr.21: 36200369-36200369 on Build GRCh38
6316171 c.343G>T p.A115S Chr.21: 36200353-36200353 on Build GRCh38
2844423 c.491+4C>T p.? Chr.21: 36200505-36200505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555036917 Chr.21: 36200711-36200711 on Build GRCh38
rs772436813 Chr.21: 36200377-36200377 on Build GRCh38
rs746027069 Chr.21: 36200371-36200371 on Build GRCh38
rs895985277 Chr.21: 36200713-36200713 on Build GRCh38
rs909087984 Chr.21: 36200732-36200732 on Build GRCh38
rs762797534 Chr.21: 36200470-36200470 on Build GRCh38
rs1000188814 Chr.21: 36200742-36200742 on Build GRCh38
rs747974617 Chr.21: 36200459-36200459 on Build GRCh38
rs910576353 Chr.21: 36200478-36200478 on Build GRCh38
rs368105209 Chr.21: 36200379-36200379 on Build GRCh38
rs757642641 Chr.21: 36200475-36200475 on Build GRCh38
rs768813541 Chr.21: 36200454-36200454 on Build GRCh38
rs966064103 Chr.21: 36200700-36200700 on Build GRCh38
rs151160436 Chr.21: 36200364-36200364 on Build GRCh38
rs985095542 Chr.21: 36200718-36200718 on Build GRCh38
rs986218700 Chr.21: 36200647-36200647 on Build GRCh38
rs9979031 Chr.21: 36200512-36200512 on Build GRCh38
rs375157280 Chr.21: 36200803-36200803 on Build GRCh38
rs568389060 Chr.21: 36200788-36200788 on Build GRCh38
rs929269412 Chr.21: 36200712-36200712 on Build GRCh38
rs772682598 Chr.21: 36200382-36200383 on Build GRCh38
rs764556098 Chr.21: 36200415-36200415 on Build GRCh38
rs201491075 Chr.21: 36200369-36200369 on Build GRCh38
rs369377073 Chr.21: 36200528-36200528 on Build GRCh38
rs776751796 Chr.21: 36200380-36200380 on Build GRCh38
rs140230955 Chr.21: 36200370-36200370 on Build GRCh38
rs750730233 Chr.21: 36200505-36200505 on Build GRCh38
rs770674531 Chr.21: 36200535-36200535 on Build GRCh38
rs1033209999 Chr.21: 36200617-36200617 on Build GRCh38
rs774848932 Chr.21: 36200551-36200551 on Build GRCh38
rs954721214 Chr.21: 36200805-36200805 on Build GRCh38
rs758094859 Chr.21: 36200353-36200353 on Build GRCh38
rs372560410 Chr.21: 36200774-36200774 on Build GRCh38
rs765630183 Chr.21: 36200553-36200553 on Build GRCh38
rs118183027 Chr.21: 36200559-36200559 on Build GRCh38
rs749491792 Chr.21: 36200840-36200840 on Build GRCh38
rs764894734 Chr.21: 36200390-36200390 on Build GRCh38
rs770551732 Chr.21: 36200481-36200481 on Build GRCh38
rs564178092 Chr.21: 36200518-36200518 on Build GRCh38
rs769771725 Chr.21: 36200467-36200467 on Build GRCh38
rs538083366 Chr.21: 36200739-36200739 on Build GRCh38
rs1048903882 Chr.21: 36200521-36200521 on Build GRCh38
rs749639655 Chr.21: 36200367-36200367 on Build GRCh38
rs747053271 Chr.21: 36200452-36200452 on Build GRCh38
rs747145267 Chr.21: 36200378-36200378 on Build GRCh38
rs940363653 Chr.21: 36200642-36200642 on Build GRCh38
rs367759390 Chr.21: 36200692-36200692 on Build GRCh38
rs368487458 Chr.21: 36200506-36200506 on Build GRCh38
rs912307042 Chr.21: 36200800-36200800 on Build GRCh38
rs887536405 Chr.21: 36200374-36200374 on Build GRCh38
rs111869439 Chr.21: 36200735-36200735 on Build GRCh38
rs566532533 Chr.21: 36200672-36200672 on Build GRCh38
rs557877553 Chr.21: 36200670-36200670 on Build GRCh38
rs377244247 Chr.21: 36200522-36200522 on Build GRCh38
rs372225329 Chr.21: 36200538-36200538 on Build GRCh38
rs200319331 Chr.21: 36200488-36200488 on Build GRCh38
rs147574672 Chr.21: 36200398-36200398 on Build GRCh38
rs145005014 Chr.21: 36200385-36200385 on Build GRCh38
rs944654306 Chr.21: 36200363-36200363 on Build GRCh38
rs1017633216 Chr.21: 36200699-36200699 on Build GRCh38
rs369129523 Chr.21: 36200537-36200537 on Build GRCh38
rs964041275 Chr.21: 36200760-36200760 on Build GRCh38
rs147103456 Chr.21: 36200460-36200460 on Build GRCh38
rs141847036 Chr.21: 36200448-36200448 on Build GRCh38
rs940418907 Chr.21: 36200820-36200820 on Build GRCh38
rs542433080 Chr.21: 36200362-36200362 on Build GRCh38
rs1001743678 Chr.21: 36200439-36200439 on Build GRCh38
rs564240269 Chr.21: 36200814-36200814 on Build GRCh38
rs1021000794 Chr.21: 36200582-36200582 on Build GRCh38
rs757806323 Chr.21: 36200441-36200441 on Build GRCh38
rs774034819 Chr.21: 36200483-36200483 on Build GRCh38
rs374871839 Chr.21: 36200545-36200545 on Build GRCh38
rs999482853 Chr.21: 36200610-36200610 on Build GRCh38
rs550784577 Chr.21: 36200566-36200566 on Build GRCh38
rs751780174 Chr.21: 36200508-36200508 on Build GRCh38
rs559705741 Chr.21: 36200447-36200447 on Build GRCh38
rs772883237 Chr.21: 36200469-36200469 on Build GRCh38
rs754260693 Chr.21: 36200428-36200428 on Build GRCh38
rs780706144 Chr.21: 36200565-36200565 on Build GRCh38
rs752265236 Chr.21: 36200496-36200496 on Build GRCh38
rs546628519 Chr.21: 36200552-36200552 on Build GRCh38
rs781367855 Chr.21: 36200497-36200497 on Build GRCh38
rs1013266732 Chr.21: 36200575-36200575 on Build GRCh38
rs1020294699 Chr.21: 36200511-36200511 on Build GRCh38
rs773077505 Chr.21: 36200392-36200392 on Build GRCh38
rs779090354 Chr.21: 36200442-36200442 on Build GRCh38
rs191568912 Chr.21: 36200583-36200583 on Build GRCh38
rs575432973 Chr.21: 36200782-36200782 on Build GRCh38
rs761313679 Chr.21: 36200628-36200628 on Build GRCh38
rs869110443 Chr.21: 36200852-36200853 on Build GRCh38
rs762451256 Chr.21: 36200397-36200397 on Build GRCh38
rs149183598 Chr.21: 36200499-36200499 on Build GRCh38
rs570562377 Chr.21: 36200704-36200704 on Build GRCh38
rs957516747 Chr.21: 36200644-36200644 on Build GRCh38
rs754534115 Chr.21: 36200399-36200399 on Build GRCh38
rs557162780 Chr.21: 36200781-36200781 on Build GRCh38
rs774155385 Chr.21: 36200536-36200536 on Build GRCh38
rs781350961 Chr.21: 36200457-36200457 on Build GRCh38
rs957488679 Chr.21: 36200468-36200468 on Build GRCh38
rs545947957 Chr.21: 36200811-36200811 on Build GRCh38
rs758679723 Chr.21: 36200445-36200445 on Build GRCh38
rs574986601 Chr.21: 36200358-36200358 on Build GRCh38
rs12482623 Chr.21: 36200687-36200687 on Build GRCh38
rs369032268 Chr.21: 36200361-36200361 on Build GRCh38
rs528474098 Chr.21: 36200526-36200526 on Build GRCh38
rs896187787 Chr.21: 36200572-36200572 on Build GRCh38
rs777818707 Chr.21: 36200531-36200531 on Build GRCh38
rs968001015 Chr.21: 36200403-36200403 on Build GRCh38
rs2409761 Chr.21: 36200750-36200750 on Build GRCh38
rs749053706 Chr.21: 36200533-36200533 on Build GRCh38
rs539588065 Chr.21: 36200650-36200650 on Build GRCh38
rs746310122 Chr.21: 36200443-36200443 on Build GRCh38
rs116743448 Chr.21: 36200560-36200560 on Build GRCh38
rs1051963375 Chr.21: 36200581-36200581 on Build GRCh38

View Full Product Details