Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36208600-36209096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY2
Entrez Gene ID
9980
Gene Name
dopey family member 2
Gene Aliases
21orf5, C21orf5
UniGene
Hs.204575
Gene Chromosome Location
Chr.21: 36156781-36294274 on Build GRCh38
Transcripts
NM_001320714.1 NM_005128.3 XM_017028509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6648664 c.615C>T p.G205G Chr.21: 36208838-36208838 on Build GRCh38
4679182 c.518C>T p.S173L Chr.21: 36208741-36208741 on Build GRCh38
236091 c.512G>C p.R171T Chr.21: 36208735-36208735 on Build GRCh38
4101448 c.519G>A p.S173S Chr.21: 36208742-36208742 on Build GRCh38
3550509 c.578C>T p.P193L Chr.21: 36208801-36208801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774847237 Chr.21: 36208779-36208779 on Build GRCh38
rs576774958 Chr.21: 36209039-36209039 on Build GRCh38
rs1022759471 Chr.21: 36208975-36208975 on Build GRCh38
rs532473565 Chr.21: 36209042-36209042 on Build GRCh38
rs759755173 Chr.21: 36208925-36208926 on Build GRCh38
rs772464706 Chr.21: 36208929-36208929 on Build GRCh38
rs374948091 Chr.21: 36208942-36208942 on Build GRCh38
rs760210059 Chr.21: 36208712-36208712 on Build GRCh38
rs142768435 Chr.21: 36208859-36208859 on Build GRCh38
rs2156412 Chr.21: 36208629-36208629 on Build GRCh38
rs753550337 Chr.21: 36208637-36208637 on Build GRCh38
rs988972507 Chr.21: 36208688-36208688 on Build GRCh38
rs768834396 Chr.21: 36208945-36208945 on Build GRCh38
rs899425563 Chr.21: 36208659-36208659 on Build GRCh38
rs200949348 Chr.21: 36208946-36208946 on Build GRCh38
rs148319957 Chr.21: 36208773-36208773 on Build GRCh38
rs1056527204 Chr.21: 36208962-36208962 on Build GRCh38
rs777451213 Chr.21: 36208902-36208902 on Build GRCh38
rs756261149 Chr.21: 36208697-36208697 on Build GRCh38
rs373279668 Chr.21: 36208816-36208816 on Build GRCh38
rs905361605 Chr.21: 36208702-36208702 on Build GRCh38
rs1012077462 Chr.21: 36208861-36208861 on Build GRCh38
rs747714952 Chr.21: 36208829-36208829 on Build GRCh38
rs368854722 Chr.21: 36208946-36208947 on Build GRCh38
rs138179841 Chr.21: 36208802-36208802 on Build GRCh38
rs777748788 Chr.21: 36208706-36208706 on Build GRCh38
rs745730357 Chr.21: 36208748-36208748 on Build GRCh38
rs781037359 Chr.21: 36208882-36208882 on Build GRCh38
rs773580318 Chr.21: 36209056-36209056 on Build GRCh38
rs747125751 Chr.21: 36209063-36209063 on Build GRCh38
rs986646629 Chr.21: 36209046-36209046 on Build GRCh38
rs374828474 Chr.21: 36208776-36208776 on Build GRCh38
rs778871181 Chr.21: 36208801-36208801 on Build GRCh38
rs997220268 Chr.21: 36209007-36209007 on Build GRCh38
rs919635709 Chr.21: 36209067-36209067 on Build GRCh38
rs763575386 Chr.21: 36208669-36208669 on Build GRCh38
rs150581929 Chr.21: 36208864-36208864 on Build GRCh38
rs751973692 Chr.21: 36208678-36208678 on Build GRCh38
rs372486945 Chr.21: 36208770-36208770 on Build GRCh38
rs200130015 Chr.21: 36208877-36208877 on Build GRCh38
rs775041596 Chr.21: 36208835-36208835 on Build GRCh38
rs968394788 Chr.21: 36208787-36208787 on Build GRCh38
rs145634540 Chr.21: 36208788-36208788 on Build GRCh38
rs1009633487 Chr.21: 36208973-36208973 on Build GRCh38
rs985660115 Chr.21: 36209034-36209034 on Build GRCh38
rs376940991 Chr.21: 36208940-36208940 on Build GRCh38
rs779829524 Chr.21: 36208870-36208870 on Build GRCh38
rs142091518 Chr.21: 36208786-36208786 on Build GRCh38
rs767876389 Chr.21: 36208840-36208840 on Build GRCh38
rs753901076 Chr.21: 36208860-36208860 on Build GRCh38
rs759995111 Chr.21: 36208838-36208838 on Build GRCh38
rs201679787 Chr.21: 36208890-36208890 on Build GRCh38
rs573816517 Chr.21: 36208968-36208968 on Build GRCh38
rs371761215 Chr.21: 36208903-36208903 on Build GRCh38
rs766113069 Chr.21: 36208907-36208907 on Build GRCh38
rs540735678 Chr.21: 36209057-36209059 on Build GRCh38
rs751144781 Chr.21: 36208620-36208620 on Build GRCh38
rs754357679 Chr.21: 36208878-36208879 on Build GRCh38
rs776272971 Chr.21: 36208820-36208820 on Build GRCh38
rs780830783 Chr.21: 36208674-36208674 on Build GRCh38
rs930924601 Chr.21: 36208907-36208907 on Build GRCh38
rs754641667 Chr.21: 36208878-36208878 on Build GRCh38
rs752701809 Chr.21: 36208695-36208695 on Build GRCh38
rs918323037 Chr.21: 36208806-36208806 on Build GRCh38
rs777557207 Chr.21: 36208760-36208760 on Build GRCh38
rs760886199 Chr.21: 36208858-36208858 on Build GRCh38
rs753919420 Chr.21: 36208792-36208792 on Build GRCh38
rs761004187 Chr.21: 36208943-36208943 on Build GRCh38
rs746670698 Chr.21: 36208761-36208761 on Build GRCh38
rs757201792 Chr.21: 36208729-36208729 on Build GRCh38
rs368393461 Chr.21: 36208998-36208998 on Build GRCh38
rs758280074 Chr.21: 36208866-36208866 on Build GRCh38
rs955252541 Chr.21: 36209038-36209038 on Build GRCh38
rs758170362 Chr.21: 36208807-36208807 on Build GRCh38
rs376449500 Chr.21: 36208847-36208847 on Build GRCh38
rs200705051 Chr.21: 36208891-36208891 on Build GRCh38
rs768195451 Chr.21: 36208819-36208819 on Build GRCh38
rs772702729 Chr.21: 36209055-36209055 on Build GRCh38
rs200685392 Chr.21: 36208717-36208717 on Build GRCh38
rs745484545 Chr.21: 36208681-36208681 on Build GRCh38
rs766788922 Chr.21: 36208670-36208670 on Build GRCh38
rs757181678 Chr.21: 36208798-36208798 on Build GRCh38
rs766823927 Chr.21: 36208832-36208832 on Build GRCh38
rs752927253 Chr.21: 36208783-36208783 on Build GRCh38
rs748483213 Chr.21: 36209045-36209045 on Build GRCh38
rs750901131 Chr.21: 36208668-36208668 on Build GRCh38
rs201917816 Chr.21: 36208863-36208863 on Build GRCh38
rs771601066 Chr.21: 36208750-36208750 on Build GRCh38
rs1028247907 Chr.21: 36209019-36209019 on Build GRCh38
rs771546721 Chr.21: 36208830-36208830 on Build GRCh38
rs527896160 Chr.21: 36208622-36208622 on Build GRCh38
rs759998080 Chr.21: 36208780-36208780 on Build GRCh38
rs775247057 Chr.21: 36208752-36208752 on Build GRCh38
rs369824664 Chr.21: 36208809-36208809 on Build GRCh38
rs143348611 Chr.21: 36208742-36208742 on Build GRCh38
rs939670383 Chr.21: 36208621-36208621 on Build GRCh38
rs377479673 Chr.21: 36208952-36208952 on Build GRCh38
rs781538980 Chr.21: 36208682-36208682 on Build GRCh38

View Full Product Details