Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36391314-36391718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
UniGene
Hs.75238
Gene Chromosome Location
Chr.21: 36380145-36416827 on Build GRCh38
Transcripts
NM_005441.2 XM_011529755.2 XM_017028477.1 XM_017028478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546797 c.335_344del10 p.Q113fs*7 Chr.21: 36391626-36391635 on Build GRCh38
5015695 c.377G>A p.R126Q Chr.21: 36391668-36391668 on Build GRCh38
725202 c.270C>A p.I90I Chr.21: 36391561-36391561 on Build GRCh38
252434 c.282G>T p.K94N Chr.21: 36391573-36391573 on Build GRCh38
6372999 c.307C>T p.Q103* Chr.21: 36391598-36391598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757870005 Chr.21: 36391650-36391650 on Build GRCh38
rs148959659 Chr.21: 36391415-36391415 on Build GRCh38
rs771187491 Chr.21: 36391564-36391564 on Build GRCh38
rs1042155375 Chr.21: 36391355-36391355 on Build GRCh38
rs771889105 Chr.21: 36391594-36391594 on Build GRCh38
rs1049989308 Chr.21: 36391400-36391400 on Build GRCh38
rs943925393 Chr.21: 36391418-36391418 on Build GRCh38
rs753502442 Chr.21: 36391514-36391514 on Build GRCh38
rs994168933 Chr.21: 36391439-36391439 on Build GRCh38
rs73383788 Chr.21: 36391369-36391369 on Build GRCh38
rs888489590 Chr.21: 36391448-36391448 on Build GRCh38
rs528036557 Chr.21: 36391492-36391492 on Build GRCh38
rs376014763 Chr.21: 36391520-36391520 on Build GRCh38
rs763992472 Chr.21: 36391500-36391500 on Build GRCh38
rs917574985 Chr.21: 36391346-36391346 on Build GRCh38
rs568896091 Chr.21: 36391677-36391677 on Build GRCh38
rs753205356 Chr.21: 36391668-36391668 on Build GRCh38
rs60992072 Chr.21: 36391481-36391481 on Build GRCh38
rs771249427 Chr.21: 36391481-36391481 on Build GRCh38
rs1026950750 Chr.21: 36391447-36391447 on Build GRCh38
rs997777831 Chr.21: 36391463-36391463 on Build GRCh38
rs965784926 Chr.21: 36391411-36391411 on Build GRCh38
rs765053500 Chr.21: 36391638-36391638 on Build GRCh38
rs749891968 Chr.21: 36391642-36391642 on Build GRCh38
rs969984244 Chr.21: 36391692-36391692 on Build GRCh38
rs1052021981 Chr.21: 36391508-36391508 on Build GRCh38
rs1010570982 Chr.21: 36391537-36391537 on Build GRCh38
rs910090564 Chr.21: 36391464-36391471 on Build GRCh38
rs772122350 Chr.21: 36391689-36391689 on Build GRCh38
rs56118104 Chr.21: 36391664-36391664 on Build GRCh38
rs1007927008 Chr.21: 36391370-36391373 on Build GRCh38
rs948239208 Chr.21: 36391477-36391481 on Build GRCh38
rs774263329 Chr.21: 36391575-36391575 on Build GRCh38
rs945046301 Chr.21: 36391347-36391347 on Build GRCh38
rs965963789 Chr.21: 36391639-36391639 on Build GRCh38
rs756632312 Chr.21: 36391673-36391673 on Build GRCh38
rs370898560 Chr.21: 36391458-36391458 on Build GRCh38
rs763933534 Chr.21: 36391621-36391621 on Build GRCh38
rs756994258 Chr.21: 36391519-36391519 on Build GRCh38
rs145410160 Chr.21: 36391442-36391442 on Build GRCh38
rs552063688 Chr.21: 36391369-36391369 on Build GRCh38
rs372344270 Chr.21: 36391556-36391556 on Build GRCh38
rs993106727 Chr.21: 36391345-36391345 on Build GRCh38
rs749460534 Chr.21: 36391559-36391559 on Build GRCh38
rs114867967 Chr.21: 36391495-36391495 on Build GRCh38
rs770075369 Chr.21: 36391553-36391553 on Build GRCh38
rs776617536 Chr.21: 36391629-36391629 on Build GRCh38
rs897144775 Chr.21: 36391420-36391420 on Build GRCh38
rs746025510 Chr.21: 36391593-36391593 on Build GRCh38
rs375331531 Chr.21: 36391487-36391487 on Build GRCh38
rs1003197500 Chr.21: 36391340-36391340 on Build GRCh38
rs78115313 Chr.21: 36391479-36391479 on Build GRCh38
rs367999820 Chr.21: 36391685-36391685 on Build GRCh38
rs757387898 Chr.21: 36391368-36391368 on Build GRCh38
rs924977949 Chr.21: 36391574-36391574 on Build GRCh38
rs1052635334 Chr.21: 36391345-36391346 on Build GRCh38
rs1043451201 Chr.21: 36391538-36391538 on Build GRCh38
rs757466879 Chr.21: 36391684-36391684 on Build GRCh38
rs771423311 Chr.21: 36391662-36391663 on Build GRCh38
rs748618760 Chr.21: 36391545-36391545 on Build GRCh38
rs374395395 Chr.21: 36391651-36391651 on Build GRCh38
rs781753785 Chr.21: 36391544-36391544 on Build GRCh38
rs1018853466 Chr.21: 36391392-36391392 on Build GRCh38
rs368616207 Chr.21: 36391543-36391543 on Build GRCh38
rs998604199 Chr.21: 36391645-36391645 on Build GRCh38
rs192219894 Chr.21: 36391443-36391443 on Build GRCh38
rs775439270 Chr.21: 36391603-36391603 on Build GRCh38
rs560209586 Chr.21: 36391521-36391521 on Build GRCh38
rs201948252 Chr.21: 36391687-36391687 on Build GRCh38
rs147616559 Chr.21: 36391459-36391459 on Build GRCh38
rs761526218 Chr.21: 36391632-36391632 on Build GRCh38
rs1040969992 Chr.21: 36391419-36391419 on Build GRCh38
rs528972595 Chr.21: 36391605-36391605 on Build GRCh38
rs376106303 Chr.21: 36391464-36391464 on Build GRCh38

View Full Product Details