Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36402418-36402957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
UniGene
Hs.75238
Gene Chromosome Location
Chr.21: 36380145-36416827 on Build GRCh38
Transcripts
NM_005441.2 XM_011529755.2 XM_017028477.1 XM_017028478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140043 c.691G>T p.D231Y Chr.21: 36402785-36402785 on Build GRCh38
6627703 c.690C>T p.D230D Chr.21: 36402784-36402784 on Build GRCh38
4671916 c.708C>T p.F236F Chr.21: 36402802-36402802 on Build GRCh38
1200936 c.673T>G p.Y225D Chr.21: 36402767-36402767 on Build GRCh38
6627706 c.677G>A p.R226Q Chr.21: 36402771-36402771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114386479 Chr.21: 36402802-36402802 on Build GRCh38
rs747978369 Chr.21: 36402714-36402717 on Build GRCh38
rs777455472 Chr.21: 36402889-36402889 on Build GRCh38
rs1041246054 Chr.21: 36402645-36402645 on Build GRCh38
rs530506949 Chr.21: 36402910-36402910 on Build GRCh38
rs958239604 Chr.21: 36402808-36402808 on Build GRCh38
rs747178648 Chr.21: 36402497-36402497 on Build GRCh38
rs904568992 Chr.21: 36402695-36402695 on Build GRCh38
rs959430885 Chr.21: 36402465-36402465 on Build GRCh38
rs138882376 Chr.21: 36402781-36402781 on Build GRCh38
rs776269311 Chr.21: 36402734-36402737 on Build GRCh38
rs936762167 Chr.21: 36402515-36402515 on Build GRCh38
rs992015517 Chr.21: 36402485-36402485 on Build GRCh38
rs139980774 Chr.21: 36402829-36402829 on Build GRCh38
rs368588534 Chr.21: 36402661-36402661 on Build GRCh38
rs77117619 Chr.21: 36402459-36402459 on Build GRCh38
rs1033815056 Chr.21: 36402729-36402729 on Build GRCh38
rs746380922 Chr.21: 36402788-36402788 on Build GRCh38
rs542931023 Chr.21: 36402892-36402892 on Build GRCh38
rs371289350 Chr.21: 36402847-36402847 on Build GRCh38
rs747255684 Chr.21: 36402597-36402597 on Build GRCh38
rs760784933 Chr.21: 36402822-36402822 on Build GRCh38
rs142631891 Chr.21: 36402462-36402462 on Build GRCh38
rs201096817 Chr.21: 36402727-36402727 on Build GRCh38
rs71330678 Chr.21: 36402734-36402734 on Build GRCh38
rs1050307786 Chr.21: 36402532-36402532 on Build GRCh38
rs761823048 Chr.21: 36402828-36402828 on Build GRCh38
rs909601322 Chr.21: 36402526-36402526 on Build GRCh38
rs35051347 Chr.21: 36402914-36402914 on Build GRCh38
rs187888857 Chr.21: 36402902-36402902 on Build GRCh38
rs777430234 Chr.21: 36402722-36402722 on Build GRCh38
rs772524408 Chr.21: 36402797-36402797 on Build GRCh38
rs750325132 Chr.21: 36402844-36402844 on Build GRCh38
rs945315501 Chr.21: 36402494-36402494 on Build GRCh38
rs1047413231 Chr.21: 36402722-36402722 on Build GRCh38
rs775264268 Chr.21: 36402917-36402917 on Build GRCh38
rs113382620 Chr.21: 36402619-36402619 on Build GRCh38
rs747758462 Chr.21: 36402774-36402774 on Build GRCh38
rs746327888 Chr.21: 36402890-36402890 on Build GRCh38
rs779933840 Chr.21: 36402745-36402745 on Build GRCh38
rs574249440 Chr.21: 36402872-36402872 on Build GRCh38
rs981050614 Chr.21: 36402558-36402558 on Build GRCh38
rs925969228 Chr.21: 36402912-36402912 on Build GRCh38
rs750374368 Chr.21: 36402732-36402732 on Build GRCh38
rs1048713668 Chr.21: 36402887-36402887 on Build GRCh38
rs199888379 Chr.21: 36402860-36402860 on Build GRCh38
rs879606365 Chr.21: 36402907-36402907 on Build GRCh38
rs762904226 Chr.21: 36402846-36402846 on Build GRCh38
rs373907249 Chr.21: 36402784-36402784 on Build GRCh38
rs746473282 Chr.21: 36402721-36402722 on Build GRCh38
rs111786621 Chr.21: 36402686-36402686 on Build GRCh38
rs939177183 Chr.21: 36402868-36402868 on Build GRCh38
rs1054818591 Chr.21: 36402909-36402909 on Build GRCh38
rs776561332 Chr.21: 36402827-36402827 on Build GRCh38
rs764029869 Chr.21: 36402717-36402717 on Build GRCh38
rs760987630 Chr.21: 36402712-36402712 on Build GRCh38
rs769823299 Chr.21: 36402916-36402916 on Build GRCh38
rs770355851 Chr.21: 36402730-36402730 on Build GRCh38
rs758009458 Chr.21: 36402722-36402725 on Build GRCh38
rs577719192 Chr.21: 36402826-36402826 on Build GRCh38
rs917806500 Chr.21: 36402486-36402486 on Build GRCh38
rs752407141 Chr.21: 36402861-36402861 on Build GRCh38
rs751127310 Chr.21: 36402752-36402752 on Build GRCh38
rs944168801 Chr.21: 36402636-36402636 on Build GRCh38
rs1018530896 Chr.21: 36402449-36402449 on Build GRCh38
rs918785144 Chr.21: 36402869-36402869 on Build GRCh38
rs145654704 Chr.21: 36402852-36402852 on Build GRCh38
rs1024789578 Chr.21: 36402758-36402758 on Build GRCh38
rs758123718 Chr.21: 36402733-36402733 on Build GRCh38
rs947913168 Chr.21: 36402533-36402533 on Build GRCh38
rs374791353 Chr.21: 36402753-36402753 on Build GRCh38
rs73902506 Chr.21: 36402506-36402506 on Build GRCh38
rs879799419 Chr.21: 36402621-36402621 on Build GRCh38
rs780539459 Chr.21: 36402763-36402763 on Build GRCh38
rs372512182 Chr.21: 36402725-36402725 on Build GRCh38
rs946004983 Chr.21: 36402660-36402660 on Build GRCh38
rs368941131 Chr.21: 36402873-36402873 on Build GRCh38
rs913788416 Chr.21: 36402888-36402888 on Build GRCh38
rs891822123 Chr.21: 36402657-36402657 on Build GRCh38
rs959166961 Chr.21: 36402744-36402744 on Build GRCh38
rs535400545 Chr.21: 36402634-36402634 on Build GRCh38

View Full Product Details