Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37267356-37267848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR3
Entrez Gene ID
10311
Gene Name
DSCR3 arrestin fold containing
Gene Aliases
DCRA, DSCRA
UniGene
Hs.369488
Gene Chromosome Location
Chr.21: 37223425-37268124 on Build GRCh38
Transcripts
NM_006052.1 XM_005260909.4 XM_005260911.4 XM_011529423.2

SNP Detail

SNP ID Chromosome Location
rs1013967279 Chr.21: 37267382-37267382 on Build GRCh38
rs969161120 Chr.21: 37267716-37267716 on Build GRCh38
rs35672675 Chr.21: 37267462-37267463 on Build GRCh38
rs930987434 Chr.21: 37267788-37267788 on Build GRCh38
rs898143927 Chr.21: 37267483-37267483 on Build GRCh38
rs973452476 Chr.21: 37267740-37267740 on Build GRCh38
rs891005436 Chr.21: 37267467-37267467 on Build GRCh38
rs932463572 Chr.21: 37267450-37267450 on Build GRCh38
rs200244479 Chr.21: 37267466-37267466 on Build GRCh38
rs142564131 Chr.21: 37267690-37267690 on Build GRCh38
rs995131195 Chr.21: 37267665-37267665 on Build GRCh38
rs536018616 Chr.21: 37267621-37267621 on Build GRCh38
rs376519929 Chr.21: 37267698-37267698 on Build GRCh38
rs774812598 Chr.21: 37267654-37267654 on Build GRCh38
rs539432656 Chr.21: 37267752-37267752 on Build GRCh38
rs397760805 Chr.21: 37267465-37267466 on Build GRCh38
rs112300108 Chr.21: 37267691-37267691 on Build GRCh38
rs1022662079 Chr.21: 37267489-37267489 on Build GRCh38
rs978354705 Chr.21: 37267445-37267445 on Build GRCh38
rs556893158 Chr.21: 37267718-37267718 on Build GRCh38
rs190480421 Chr.21: 37267758-37267758 on Build GRCh38
rs1051848779 Chr.21: 37267488-37267488 on Build GRCh38
rs958283738 Chr.21: 37267594-37267595 on Build GRCh38
rs184370156 Chr.21: 37267505-37267505 on Build GRCh38
rs921974677 Chr.21: 37267610-37267610 on Build GRCh38
rs546951411 Chr.21: 37267563-37267563 on Build GRCh38
rs974731278 Chr.21: 37267471-37267471 on Build GRCh38
rs895513609 Chr.21: 37267655-37267655 on Build GRCh38
rs370620632 Chr.21: 37267400-37267400 on Build GRCh38
rs866985987 Chr.21: 37267615-37267615 on Build GRCh38
rs570324263 Chr.21: 37267730-37267730 on Build GRCh38
rs192629319 Chr.21: 37267493-37267493 on Build GRCh38
rs572803892 Chr.21: 37267815-37267815 on Build GRCh38
rs561800428 Chr.21: 37267524-37267524 on Build GRCh38
rs1015315991 Chr.21: 37267721-37267721 on Build GRCh38
rs974709228 Chr.21: 37267717-37267717 on Build GRCh38
rs1029228525 Chr.21: 37267702-37267702 on Build GRCh38
rs776233091 Chr.21: 37267501-37267501 on Build GRCh38
rs1037557959 Chr.21: 37267637-37267637 on Build GRCh38
rs151095488 Chr.21: 37267657-37267657 on Build GRCh38
rs1037396357 Chr.21: 37267785-37267785 on Build GRCh38
rs1015654471 Chr.21: 37267482-37267482 on Build GRCh38
rs1035911523 Chr.21: 37267529-37267529 on Build GRCh38
rs1014457764 Chr.21: 37267540-37267540 on Build GRCh38
rs920388529 Chr.21: 37267645-37267645 on Build GRCh38
rs1003897503 Chr.21: 37267476-37267476 on Build GRCh38
rs377485760 Chr.21: 37267458-37267458 on Build GRCh38
rs891064781 Chr.21: 37267648-37267648 on Build GRCh38
rs768077836 Chr.21: 37267712-37267712 on Build GRCh38
rs947434078 Chr.21: 37267391-37267391 on Build GRCh38
rs1020032463 Chr.21: 37267547-37267547 on Build GRCh38
rs914578618 Chr.21: 37267555-37267555 on Build GRCh38
rs1022671854 Chr.21: 37267677-37267677 on Build GRCh38
rs905585921 Chr.21: 37267682-37267682 on Build GRCh38
rs548366586 Chr.21: 37267401-37267401 on Build GRCh38
rs899693434 Chr.21: 37267424-37267424 on Build GRCh38
rs925523978 Chr.21: 37267451-37267451 on Build GRCh38
rs974931920 Chr.21: 37267599-37267599 on Build GRCh38
rs919816315 Chr.21: 37267787-37267787 on Build GRCh38
rs537038829 Chr.21: 37267396-37267396 on Build GRCh38
rs921959741 Chr.21: 37267600-37267600 on Build GRCh38
rs115211774 Chr.21: 37267584-37267584 on Build GRCh38
rs999861436 Chr.21: 37267679-37267679 on Build GRCh38
rs373146302 Chr.21: 37267408-37267408 on Build GRCh38
rs937250719 Chr.21: 37267647-37267647 on Build GRCh38
rs530957755 Chr.21: 37267525-37267525 on Build GRCh38
rs899096133 Chr.21: 37267523-37267523 on Build GRCh38
rs989494434 Chr.21: 37267694-37267694 on Build GRCh38
rs932385575 Chr.21: 37267516-37267516 on Build GRCh38
rs939862357 Chr.21: 37267773-37267773 on Build GRCh38
rs559620451 Chr.21: 37267486-37267486 on Build GRCh38
rs1055472961 Chr.21: 37267528-37267528 on Build GRCh38
rs549643566 Chr.21: 37267640-37267640 on Build GRCh38
rs564359364 Chr.21: 37267537-37267537 on Build GRCh38
rs898189454 Chr.21: 37267659-37267659 on Build GRCh38
rs550944427 Chr.21: 37267536-37267536 on Build GRCh38
rs764816641 Chr.21: 37267556-37267556 on Build GRCh38
rs568267537 Chr.21: 37267560-37267560 on Build GRCh38
rs1003008955 Chr.21: 37267687-37267687 on Build GRCh38
rs1050888393 Chr.21: 37267638-37267638 on Build GRCh38
rs561433791 Chr.21: 37267374-37267374 on Build GRCh38
rs564764414 Chr.21: 37267797-37267797 on Build GRCh38
rs1041746168 Chr.21: 37267664-37267664 on Build GRCh38
rs572667026 Chr.21: 37267494-37267494 on Build GRCh38
rs954756022 Chr.21: 37267723-37267723 on Build GRCh38
rs940727414 Chr.21: 37267633-37267633 on Build GRCh38

View Full Product Details