Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38816586-38817112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS2
Entrez Gene ID
2114
Gene Name
ETS proto-oncogene 2, transcription factor
Gene Aliases
ETS2IT1
UniGene
Hs.644231
Gene Chromosome Location
Chr.21: 38805307-38824955 on Build GRCh38
Transcripts
NM_001256295.1 NM_005239.5 XM_005260935.1 XM_017028290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444458 c.554C>T p.T185I Chr.21: 38817056-38817056 on Build GRCh38
4963322 c.552C>T p.L184L Chr.21: 38817054-38817054 on Build GRCh38
5127184 c.578G>A p.S193N Chr.21: 38817080-38817080 on Build GRCh38
4514645 c.562C>T p.P188S Chr.21: 38817064-38817064 on Build GRCh38
4101577 c.575A>G p.N192S Chr.21: 38817077-38817077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757283346 Chr.21: 38816992-38816993 on Build GRCh38
rs192619285 Chr.21: 38816945-38816945 on Build GRCh38
rs1021456471 Chr.21: 38816731-38816731 on Build GRCh38
rs893521276 Chr.21: 38816861-38816861 on Build GRCh38
rs540790590 Chr.21: 38816862-38816862 on Build GRCh38
rs753135993 Chr.21: 38816836-38816836 on Build GRCh38
rs767704722 Chr.21: 38816759-38816759 on Build GRCh38
rs779726526 Chr.21: 38817081-38817081 on Build GRCh38
rs571287030 Chr.21: 38816946-38816946 on Build GRCh38
rs900650451 Chr.21: 38816885-38816885 on Build GRCh38
rs991035570 Chr.21: 38816666-38816666 on Build GRCh38
rs990021036 Chr.21: 38816730-38816730 on Build GRCh38
rs1048686845 Chr.21: 38816929-38816929 on Build GRCh38
rs752338255 Chr.21: 38816959-38816959 on Build GRCh38
rs781393803 Chr.21: 38817024-38817026 on Build GRCh38
rs373754896 Chr.21: 38816752-38816752 on Build GRCh38
rs747434496 Chr.21: 38817018-38817018 on Build GRCh38
rs749216483 Chr.21: 38817086-38817087 on Build GRCh38
rs372145357 Chr.21: 38817061-38817061 on Build GRCh38
rs555576193 Chr.21: 38816628-38816628 on Build GRCh38
rs146177907 Chr.21: 38816932-38816932 on Build GRCh38
rs774398362 Chr.21: 38816738-38816738 on Build GRCh38
rs931431177 Chr.21: 38816903-38816905 on Build GRCh38
rs768857570 Chr.21: 38817035-38817035 on Build GRCh38
rs543698487 Chr.21: 38816627-38816627 on Build GRCh38
rs777032839 Chr.21: 38817060-38817060 on Build GRCh38
rs542510514 Chr.21: 38816791-38816791 on Build GRCh38
rs527378493 Chr.21: 38816947-38816947 on Build GRCh38
rs869231464 Chr.21: 38816942-38816943 on Build GRCh38
rs1039637672 Chr.21: 38816866-38816866 on Build GRCh38
rs938432461 Chr.21: 38816846-38816846 on Build GRCh38
rs777488061 Chr.21: 38816976-38816976 on Build GRCh38
rs879801446 Chr.21: 38816882-38816882 on Build GRCh38
rs923641473 Chr.21: 38816772-38816772 on Build GRCh38
rs755794929 Chr.21: 38816962-38816962 on Build GRCh38
rs764703277 Chr.21: 38816895-38816895 on Build GRCh38
rs949360084 Chr.21: 38816753-38816753 on Build GRCh38
rs562451865 Chr.21: 38816919-38816919 on Build GRCh38
rs767219668 Chr.21: 38816958-38816958 on Build GRCh38
rs36015447 Chr.21: 38816632-38816633 on Build GRCh38
rs953328003 Chr.21: 38816661-38816661 on Build GRCh38
rs374338058 Chr.21: 38816985-38816985 on Build GRCh38
rs368261290 Chr.21: 38816986-38816986 on Build GRCh38

View Full Product Details