Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38818275-38818780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS2
Entrez Gene ID
2114
Gene Name
ETS proto-oncogene 2, transcription factor
Gene Aliases
ETS2IT1
UniGene
Hs.644231
Gene Chromosome Location
Chr.21: 38805307-38824955 on Build GRCh38
Transcripts
NM_001256295.1 NM_005239.5 XM_005260935.1 XM_017028290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841907 c.748A>C p.T250P Chr.21: 38818583-38818583 on Build GRCh38
3357290 c.747C>T p.V249V Chr.21: 38818582-38818582 on Build GRCh38
6656698 c.722A>G p.K241R Chr.21: 38818557-38818557 on Build GRCh38
6053179 c.728G>A p.R243Q Chr.21: 38818563-38818563 on Build GRCh38
6656700 c.712A>T p.M238L Chr.21: 38818547-38818547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771176252 Chr.21: 38818491-38818491 on Build GRCh38
rs981324703 Chr.21: 38818743-38818743 on Build GRCh38
rs543150382 Chr.21: 38818707-38818707 on Build GRCh38
rs1044199002 Chr.21: 38818510-38818510 on Build GRCh38
rs889960236 Chr.21: 38818334-38818334 on Build GRCh38
rs988371237 Chr.21: 38818756-38818756 on Build GRCh38
rs772028653 Chr.21: 38818425-38818425 on Build GRCh38
rs969951821 Chr.21: 38818455-38818455 on Build GRCh38
rs746987804 Chr.21: 38818621-38818621 on Build GRCh38
rs752058266 Chr.21: 38818376-38818376 on Build GRCh38
rs10854389 Chr.21: 38818696-38818696 on Build GRCh38
rs763874169 Chr.21: 38818449-38818449 on Build GRCh38
rs151101102 Chr.21: 38818340-38818340 on Build GRCh38
rs768032585 Chr.21: 38818552-38818552 on Build GRCh38
rs371820145 Chr.21: 38818413-38818413 on Build GRCh38
rs539828835 Chr.21: 38818532-38818532 on Build GRCh38
rs978094744 Chr.21: 38818357-38818357 on Build GRCh38
rs776625757 Chr.21: 38818643-38818643 on Build GRCh38
rs762488396 Chr.21: 38818680-38818680 on Build GRCh38
rs1043593749 Chr.21: 38818339-38818339 on Build GRCh38
rs115786160 Chr.21: 38818519-38818519 on Build GRCh38
rs757431412 Chr.21: 38818579-38818579 on Build GRCh38
rs764505333 Chr.21: 38818572-38818572 on Build GRCh38
rs753252691 Chr.21: 38818554-38818554 on Build GRCh38
rs549885445 Chr.21: 38818404-38818404 on Build GRCh38
rs563439213 Chr.21: 38818730-38818730 on Build GRCh38
rs61735785 Chr.21: 38818484-38818484 on Build GRCh38
rs560938662 Chr.21: 38818336-38818336 on Build GRCh38
rs202193691 Chr.21: 38818669-38818669 on Build GRCh38
rs960975215 Chr.21: 38818750-38818750 on Build GRCh38
rs372641031 Chr.21: 38818523-38818523 on Build GRCh38
rs114460001 Chr.21: 38818488-38818488 on Build GRCh38
rs376514555 Chr.21: 38818422-38818422 on Build GRCh38
rs981053782 Chr.21: 38818456-38818456 on Build GRCh38
rs115908228 Chr.21: 38818481-38818481 on Build GRCh38
rs567494567 Chr.21: 38818360-38818360 on Build GRCh38
rs755992789 Chr.21: 38818548-38818548 on Build GRCh38
rs369614292 Chr.21: 38818438-38818438 on Build GRCh38
rs8134490 Chr.21: 38818296-38818296 on Build GRCh38
rs370030170 Chr.21: 38818480-38818480 on Build GRCh38
rs371568403 Chr.21: 38818538-38818538 on Build GRCh38
rs781476492 Chr.21: 38818388-38818388 on Build GRCh38
rs933669578 Chr.21: 38818710-38818710 on Build GRCh38
rs777930829 Chr.21: 38818405-38818405 on Build GRCh38
rs749391035 Chr.21: 38818408-38818408 on Build GRCh38
rs746824654 Chr.21: 38818504-38818504 on Build GRCh38
rs946543073 Chr.21: 38818335-38818335 on Build GRCh38
rs753494319 Chr.21: 38818472-38818472 on Build GRCh38
rs755496148 Chr.21: 38818475-38818475 on Build GRCh38
rs115297166 Chr.21: 38818500-38818500 on Build GRCh38
rs747921885 Chr.21: 38818659-38818659 on Build GRCh38
rs577157851 Chr.21: 38818520-38818520 on Build GRCh38
rs771905336 Chr.21: 38818502-38818502 on Build GRCh38
rs755480942 Chr.21: 38818377-38818377 on Build GRCh38
rs116698978 Chr.21: 38818580-38818580 on Build GRCh38
rs892527617 Chr.21: 38818333-38818333 on Build GRCh38
rs768711199 Chr.21: 38818626-38818626 on Build GRCh38
rs890802282 Chr.21: 38818374-38818374 on Build GRCh38
rs775434582 Chr.21: 38818443-38818443 on Build GRCh38
rs369448029 Chr.21: 38818691-38818691 on Build GRCh38
rs552586832 Chr.21: 38818476-38818476 on Build GRCh38
rs758506814 Chr.21: 38818594-38818594 on Build GRCh38
rs909710085 Chr.21: 38818622-38818622 on Build GRCh38
rs1024159376 Chr.21: 38818563-38818563 on Build GRCh38
rs138127643 Chr.21: 38818574-38818574 on Build GRCh38
rs751140138 Chr.21: 38818359-38818359 on Build GRCh38
rs939298893 Chr.21: 38818298-38818298 on Build GRCh38
rs571412345 Chr.21: 38818312-38818312 on Build GRCh38
rs563116610 Chr.21: 38818409-38818409 on Build GRCh38
rs777554249 Chr.21: 38818549-38818549 on Build GRCh38
rs200893699 Chr.21: 38818474-38818474 on Build GRCh38
rs1052523864 Chr.21: 38818331-38818331 on Build GRCh38
rs114562289 Chr.21: 38818503-38818503 on Build GRCh38
rs764929763 Chr.21: 38818530-38818530 on Build GRCh38
rs778912960 Chr.21: 38818496-38818496 on Build GRCh38
rs192909371 Chr.21: 38818295-38818295 on Build GRCh38
rs780262985 Chr.21: 38818620-38818620 on Build GRCh38
rs745969118 Chr.21: 38818587-38818587 on Build GRCh38
rs572228891 Chr.21: 38818297-38818297 on Build GRCh38
rs558527007 Chr.21: 38818535-38818535 on Build GRCh38
rs780635928 Chr.21: 38818375-38818375 on Build GRCh38
rs200961208 Chr.21: 38818444-38818444 on Build GRCh38
rs376534683 Chr.21: 38818664-38818664 on Build GRCh38
rs114815090 Chr.21: 38818748-38818748 on Build GRCh38
rs1015316627 Chr.21: 38818410-38818410 on Build GRCh38
rs150430243 Chr.21: 38818562-38818562 on Build GRCh38
rs541233040 Chr.21: 38818328-38818328 on Build GRCh38

View Full Product Details