Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39185325-39185843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD1
Entrez Gene ID
54014
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
C21orf107, DCAF19, N143, WDR9
UniGene
Hs.627139
Gene Chromosome Location
Chr.21: 39184176-39321595 on Build GRCh38
Transcripts
NM_018963.4 NM_033656.3 XM_011529611.1 XM_011529612.2 XM_011529613.1 XM_017028373.1 XM_017028374.1

SNP Detail

SNP ID Chromosome Location
rs13046464 Chr.21: 39185576-39185576 on Build GRCh38
rs758927991 Chr.21: 39185624-39185625 on Build GRCh38
rs529349096 Chr.21: 39185710-39185711 on Build GRCh38
rs878957702 Chr.21: 39185796-39185796 on Build GRCh38
rs937398204 Chr.21: 39185704-39185704 on Build GRCh38
rs576558647 Chr.21: 39185729-39185729 on Build GRCh38
rs374097713 Chr.21: 39185646-39185646 on Build GRCh38
rs781200555 Chr.21: 39185710-39185712 on Build GRCh38
rs902997936 Chr.21: 39185757-39185757 on Build GRCh38
rs967975360 Chr.21: 39185466-39185466 on Build GRCh38
rs202009663 Chr.21: 39185515-39185516 on Build GRCh38
rs888858718 Chr.21: 39185812-39185812 on Build GRCh38
rs949934079 Chr.21: 39185742-39185742 on Build GRCh38
rs543225818 Chr.21: 39185476-39185476 on Build GRCh38
rs201257356 Chr.21: 39185706-39185708 on Build GRCh38
rs1056191839 Chr.21: 39185711-39185711 on Build GRCh38
rs1054233302 Chr.21: 39185779-39185779 on Build GRCh38
rs976925770 Chr.21: 39185607-39185607 on Build GRCh38
rs1007618022 Chr.21: 39185559-39185559 on Build GRCh38
rs112396503 Chr.21: 39185515-39185515 on Build GRCh38
rs200916294 Chr.21: 39185517-39185517 on Build GRCh38
rs568715397 Chr.21: 39185557-39185557 on Build GRCh38
rs916303387 Chr.21: 39185621-39185621 on Build GRCh38
rs774654246 Chr.21: 39185752-39185752 on Build GRCh38
rs11287806 Chr.21: 39185541-39185541 on Build GRCh38
rs142513641 Chr.21: 39185767-39185767 on Build GRCh38
rs573949437 Chr.21: 39185471-39185471 on Build GRCh38
rs116237092 Chr.21: 39185385-39185385 on Build GRCh38
rs368441044 Chr.21: 39185647-39185647 on Build GRCh38
rs747059896 Chr.21: 39185709-39185712 on Build GRCh38
rs1033105988 Chr.21: 39185437-39185437 on Build GRCh38
rs749374844 Chr.21: 39185398-39185398 on Build GRCh38
rs1056230131 Chr.21: 39185543-39185543 on Build GRCh38
rs769625811 Chr.21: 39185711-39185712 on Build GRCh38
rs918719390 Chr.21: 39185636-39185636 on Build GRCh38
rs923789984 Chr.21: 39185502-39185502 on Build GRCh38
rs34806799 Chr.21: 39185709-39185710 on Build GRCh38
rs796751402 Chr.21: 39185544-39185544 on Build GRCh38
rs6517523 Chr.21: 39185406-39185406 on Build GRCh38
rs968404448 Chr.21: 39185807-39185807 on Build GRCh38
rs532237998 Chr.21: 39185395-39185397 on Build GRCh38
rs200869358 Chr.21: 39185708-39185708 on Build GRCh38
rs543647490 Chr.21: 39185709-39185711 on Build GRCh38
rs374539502 Chr.21: 39185681-39185681 on Build GRCh38
rs753365977 Chr.21: 39185560-39185561 on Build GRCh38
rs1021615541 Chr.21: 39185778-39185778 on Build GRCh38
rs397712075 Chr.21: 39185720-39185721 on Build GRCh38
rs760828368 Chr.21: 39185514-39185515 on Build GRCh38
rs1018566498 Chr.21: 39185578-39185578 on Build GRCh38
rs1028993995 Chr.21: 39185373-39185375 on Build GRCh38
rs997552176 Chr.21: 39185350-39185351 on Build GRCh38
rs1041552920 Chr.21: 39185743-39185743 on Build GRCh38
rs1005124972 Chr.21: 39185647-39185648 on Build GRCh38
rs966003370 Chr.21: 39185600-39185600 on Build GRCh38
rs953441006 Chr.21: 39185766-39185767 on Build GRCh38

View Full Product Details