Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39187556-39188063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD1
Entrez Gene ID
54014
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
C21orf107, DCAF19, N143, WDR9
UniGene
Hs.627139
Gene Chromosome Location
Chr.21: 39184176-39321595 on Build GRCh38
Transcripts
NM_018963.4 NM_033656.3 XM_011529611.1 XM_011529612.2 XM_011529613.1 XM_017028373.1 XM_017028374.1

SNP Detail

SNP ID Chromosome Location
rs1053238447 Chr.21: 39187991-39187991 on Build GRCh38
rs188577349 Chr.21: 39187600-39187600 on Build GRCh38
rs770327835 Chr.21: 39187819-39187819 on Build GRCh38
rs973173990 Chr.21: 39187854-39187854 on Build GRCh38
rs11910039 Chr.21: 39187941-39187941 on Build GRCh38
rs893213952 Chr.21: 39187920-39187920 on Build GRCh38
rs904246924 Chr.21: 39187662-39187662 on Build GRCh38
rs1017453061 Chr.21: 39187827-39187827 on Build GRCh38
rs370116366 Chr.21: 39187707-39187710 on Build GRCh38
rs1053281820 Chr.21: 39187852-39187852 on Build GRCh38
rs751158371 Chr.21: 39187807-39187808 on Build GRCh38
rs75487026 Chr.21: 39187874-39187874 on Build GRCh38
rs11909837 Chr.21: 39187986-39187986 on Build GRCh38
rs544817164 Chr.21: 39187850-39187850 on Build GRCh38
rs759180426 Chr.21: 39187901-39187901 on Build GRCh38
rs943172677 Chr.21: 39187654-39187654 on Build GRCh38
rs185154497 Chr.21: 39188021-39188021 on Build GRCh38
rs578177435 Chr.21: 39187703-39187706 on Build GRCh38
rs1000291021 Chr.21: 39187669-39187669 on Build GRCh38
rs906093579 Chr.21: 39188037-39188038 on Build GRCh38
rs934868166 Chr.21: 39187845-39187845 on Build GRCh38
rs981688291 Chr.21: 39187790-39187790 on Build GRCh38
rs117885878 Chr.21: 39187674-39187674 on Build GRCh38
rs918592634 Chr.21: 39187888-39187888 on Build GRCh38
rs943221944 Chr.21: 39187932-39187932 on Build GRCh38
rs535152104 Chr.21: 39187601-39187601 on Build GRCh38
rs950557229 Chr.21: 39187603-39187603 on Build GRCh38
rs375092743 Chr.21: 39188022-39188022 on Build GRCh38
rs1001674641 Chr.21: 39187820-39187821 on Build GRCh38
rs947472926 Chr.21: 39187943-39187943 on Build GRCh38
rs1016463120 Chr.21: 39187661-39187661 on Build GRCh38
rs929216413 Chr.21: 39187591-39187591 on Build GRCh38
rs916222662 Chr.21: 39187711-39187711 on Build GRCh38
rs770884785 Chr.21: 39187816-39187817 on Build GRCh38
rs77444825 Chr.21: 39187635-39187635 on Build GRCh38
rs541328433 Chr.21: 39187851-39187851 on Build GRCh38
rs987356423 Chr.21: 39187904-39187904 on Build GRCh38
rs948984623 Chr.21: 39187787-39187787 on Build GRCh38
rs900530733 Chr.21: 39187985-39187985 on Build GRCh38
rs559162470 Chr.21: 39187584-39187584 on Build GRCh38
rs1014514205 Chr.21: 39187788-39187788 on Build GRCh38
rs1039459255 Chr.21: 39187954-39187954 on Build GRCh38
rs1004727926 Chr.21: 39188032-39188032 on Build GRCh38
rs564771858 Chr.21: 39187903-39187903 on Build GRCh38
rs990893341 Chr.21: 39187709-39187709 on Build GRCh38
rs73221197 Chr.21: 39187844-39187844 on Build GRCh38
rs562640309 Chr.21: 39187760-39187760 on Build GRCh38
rs180715794 Chr.21: 39188015-39188015 on Build GRCh38
rs891471166 Chr.21: 39188017-39188017 on Build GRCh38
rs555095671 Chr.21: 39187611-39187611 on Build GRCh38
rs777302284 Chr.21: 39187615-39187615 on Build GRCh38

View Full Product Details