Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39187818-39188364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD1
Entrez Gene ID
54014
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
C21orf107, DCAF19, N143, WDR9
UniGene
Hs.627139
Gene Chromosome Location
Chr.21: 39184176-39321595 on Build GRCh38
Transcripts
NM_018963.4 NM_033656.3 XM_011529611.1 XM_011529612.2 XM_011529613.1 XM_017028373.1 XM_017028374.1

SNP Detail

SNP ID Chromosome Location
rs1053238447 Chr.21: 39187991-39187991 on Build GRCh38
rs182313771 Chr.21: 39188196-39188196 on Build GRCh38
rs973173990 Chr.21: 39187854-39187854 on Build GRCh38
rs912957457 Chr.21: 39188244-39188244 on Build GRCh38
rs11910039 Chr.21: 39187941-39187941 on Build GRCh38
rs914718704 Chr.21: 39188238-39188238 on Build GRCh38
rs752306443 Chr.21: 39188070-39188070 on Build GRCh38
rs893213952 Chr.21: 39187920-39187920 on Build GRCh38
rs911833440 Chr.21: 39188185-39188185 on Build GRCh38
rs956253013 Chr.21: 39188221-39188221 on Build GRCh38
rs752185243 Chr.21: 39188045-39188045 on Build GRCh38
rs1053281820 Chr.21: 39187852-39187852 on Build GRCh38
rs75487026 Chr.21: 39187874-39187874 on Build GRCh38
rs928924761 Chr.21: 39188211-39188211 on Build GRCh38
rs11909837 Chr.21: 39187986-39187986 on Build GRCh38
rs544817164 Chr.21: 39187850-39187850 on Build GRCh38
rs543947526 Chr.21: 39188074-39188074 on Build GRCh38
rs759180426 Chr.21: 39187901-39187901 on Build GRCh38
rs11910151 Chr.21: 39188336-39188336 on Build GRCh38
rs949719900 Chr.21: 39188245-39188245 on Build GRCh38
rs185154497 Chr.21: 39188021-39188021 on Build GRCh38
rs947529693 Chr.21: 39188307-39188307 on Build GRCh38
rs906093579 Chr.21: 39188037-39188038 on Build GRCh38
rs934868166 Chr.21: 39187845-39187845 on Build GRCh38
rs192187280 Chr.21: 39188172-39188172 on Build GRCh38
rs918592634 Chr.21: 39187888-39187888 on Build GRCh38
rs906144546 Chr.21: 39188252-39188252 on Build GRCh38
rs943221944 Chr.21: 39187932-39187932 on Build GRCh38
rs375092743 Chr.21: 39188022-39188022 on Build GRCh38
rs994264002 Chr.21: 39188105-39188106 on Build GRCh38
rs947472926 Chr.21: 39187943-39187943 on Build GRCh38
rs539355306 Chr.21: 39188294-39188294 on Build GRCh38
rs541328433 Chr.21: 39187851-39187851 on Build GRCh38
rs963157204 Chr.21: 39188104-39188107 on Build GRCh38
rs1026113918 Chr.21: 39188116-39188116 on Build GRCh38
rs987356423 Chr.21: 39187904-39187904 on Build GRCh38
rs763924196 Chr.21: 39188186-39188186 on Build GRCh38
rs925807888 Chr.21: 39188201-39188201 on Build GRCh38
rs900530733 Chr.21: 39187985-39187985 on Build GRCh38
rs140712472 Chr.21: 39188049-39188049 on Build GRCh38
rs562519075 Chr.21: 39188177-39188177 on Build GRCh38
rs1039459255 Chr.21: 39187954-39187954 on Build GRCh38
rs989403094 Chr.21: 39188228-39188228 on Build GRCh38
rs1004727926 Chr.21: 39188032-39188032 on Build GRCh38
rs986739295 Chr.21: 39188183-39188183 on Build GRCh38
rs564771858 Chr.21: 39187903-39187903 on Build GRCh38
rs368315354 Chr.21: 39188142-39188142 on Build GRCh38
rs73221197 Chr.21: 39187844-39187844 on Build GRCh38
rs1045409030 Chr.21: 39188248-39188251 on Build GRCh38
rs180715794 Chr.21: 39188015-39188015 on Build GRCh38
rs891471166 Chr.21: 39188017-39188017 on Build GRCh38
rs964585921 Chr.21: 39188195-39188195 on Build GRCh38
rs377188932 Chr.21: 39188287-39188287 on Build GRCh38
rs757254798 Chr.21: 39188295-39188295 on Build GRCh38

View Full Product Details