Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39192058-39192606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD1
Entrez Gene ID
54014
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
C21orf107, DCAF19, N143, WDR9
UniGene
Hs.627139
Gene Chromosome Location
Chr.21: 39184176-39321595 on Build GRCh38
Transcripts
NM_018963.4 NM_033656.3 XM_011529611.1 XM_011529612.2 XM_011529613.1 XM_017028373.1 XM_017028374.1

SNP Detail

SNP ID Chromosome Location
rs577828015 Chr.21: 39192533-39192533 on Build GRCh38
rs967835201 Chr.21: 39192204-39192204 on Build GRCh38
rs1022187964 Chr.21: 39192396-39192396 on Build GRCh38
rs35945460 Chr.21: 39192388-39192388 on Build GRCh38
rs999337917 Chr.21: 39192435-39192435 on Build GRCh38
rs1036422530 Chr.21: 39192289-39192289 on Build GRCh38
rs555104715 Chr.21: 39192354-39192354 on Build GRCh38
rs552025066 Chr.21: 39192093-39192093 on Build GRCh38
rs563953147 Chr.21: 39192140-39192140 on Build GRCh38
rs1048238855 Chr.21: 39192295-39192295 on Build GRCh38
rs539634246 Chr.21: 39192255-39192255 on Build GRCh38
rs916232161 Chr.21: 39192539-39192539 on Build GRCh38
rs575157079 Chr.21: 39192380-39192380 on Build GRCh38
rs960725643 Chr.21: 39192510-39192511 on Build GRCh38
rs58041256 Chr.21: 39192263-39192267 on Build GRCh38
rs948300149 Chr.21: 39192247-39192247 on Build GRCh38
rs757037637 Chr.21: 39192261-39192262 on Build GRCh38
rs376464102 Chr.21: 39192151-39192151 on Build GRCh38
rs1030782604 Chr.21: 39192453-39192453 on Build GRCh38
rs960380942 Chr.21: 39192222-39192222 on Build GRCh38
rs553107805 Chr.21: 39192263-39192263 on Build GRCh38
rs370727796 Chr.21: 39192449-39192451 on Build GRCh38
rs902777364 Chr.21: 39192511-39192511 on Build GRCh38
rs1032893720 Chr.21: 39192104-39192104 on Build GRCh38
rs918515781 Chr.21: 39192257-39192257 on Build GRCh38
rs2410104 Chr.21: 39192538-39192538 on Build GRCh38
rs992219024 Chr.21: 39192238-39192238 on Build GRCh38
rs937158421 Chr.21: 39192311-39192311 on Build GRCh38
rs535448873 Chr.21: 39192303-39192303 on Build GRCh38
rs2898369 Chr.21: 39192398-39192398 on Build GRCh38
rs531437829 Chr.21: 39192240-39192240 on Build GRCh38
rs2836933 Chr.21: 39192276-39192276 on Build GRCh38
rs200921557 Chr.21: 39192266-39192266 on Build GRCh38
rs1006758938 Chr.21: 39192153-39192153 on Build GRCh38
rs866841149 Chr.21: 39192486-39192486 on Build GRCh38
rs1032575676 Chr.21: 39192577-39192577 on Build GRCh38
rs967888977 Chr.21: 39192417-39192419 on Build GRCh38
rs112629458 Chr.21: 39192262-39192266 on Build GRCh38
rs1050850604 Chr.21: 39192306-39192306 on Build GRCh38
rs550872039 Chr.21: 39192105-39192105 on Build GRCh38
rs192820029 Chr.21: 39192552-39192552 on Build GRCh38
rs969045890 Chr.21: 39192544-39192544 on Build GRCh38
rs977844345 Chr.21: 39192215-39192215 on Build GRCh38
rs1022135461 Chr.21: 39192159-39192159 on Build GRCh38
rs189064105 Chr.21: 39192403-39192403 on Build GRCh38
rs911304162 Chr.21: 39192221-39192221 on Build GRCh38
rs895561620 Chr.21: 39192357-39192357 on Build GRCh38
rs879812576 Chr.21: 39192446-39192446 on Build GRCh38

View Full Product Details