Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40062759-40063306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 XM_017028281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934515 c.4895A>G p.K1632R Chr.21: 40062893-40062893 on Build GRCh38
5858631 c.4917G>A p.M1639I Chr.21: 40062871-40062871 on Build GRCh38
5948675 c.4894_4895delAA p.K1632fs*4 Chr.21: 40062893-40062894 on Build GRCh38
2818498 c.4917G>T p.M1639I Chr.21: 40062871-40062871 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556683191 Chr.21: 40062983-40062983 on Build GRCh38
rs928247156 Chr.21: 40063106-40063106 on Build GRCh38
rs74728842 Chr.21: 40063245-40063245 on Build GRCh38
rs750153687 Chr.21: 40062909-40062909 on Build GRCh38
rs954069100 Chr.21: 40063233-40063233 on Build GRCh38
rs367982690 Chr.21: 40062833-40062833 on Build GRCh38
rs982314377 Chr.21: 40063041-40063041 on Build GRCh38
rs767793306 Chr.21: 40062874-40062874 on Build GRCh38
rs897266036 Chr.21: 40063120-40063120 on Build GRCh38
rs565986526 Chr.21: 40062796-40062796 on Build GRCh38
rs1018901177 Chr.21: 40062964-40062964 on Build GRCh38
rs748067263 Chr.21: 40063158-40063158 on Build GRCh38
rs1029702134 Chr.21: 40063224-40063224 on Build GRCh38
rs1015414451 Chr.21: 40063113-40063113 on Build GRCh38
rs937967885 Chr.21: 40063010-40063010 on Build GRCh38
rs569406885 Chr.21: 40062788-40062788 on Build GRCh38
rs1033271987 Chr.21: 40063256-40063256 on Build GRCh38
rs761867690 Chr.21: 40063261-40063261 on Build GRCh38
rs768797611 Chr.21: 40063011-40063011 on Build GRCh38
rs749841219 Chr.21: 40062949-40062949 on Build GRCh38
rs899169280 Chr.21: 40063142-40063142 on Build GRCh38
rs775493319 Chr.21: 40062883-40062883 on Build GRCh38
rs886259685 Chr.21: 40063244-40063244 on Build GRCh38
rs2837409 Chr.21: 40062914-40062914 on Build GRCh38
rs926153579 Chr.21: 40062785-40062785 on Build GRCh38
rs771871199 Chr.21: 40062842-40062842 on Build GRCh38
rs1055501444 Chr.21: 40063128-40063128 on Build GRCh38
rs754488941 Chr.21: 40062927-40062927 on Build GRCh38
rs765049665 Chr.21: 40062905-40062905 on Build GRCh38
rs780514852 Chr.21: 40062929-40062929 on Build GRCh38
rs774812703 Chr.21: 40062856-40062856 on Build GRCh38
rs534578109 Chr.21: 40062812-40062812 on Build GRCh38
rs371017133 Chr.21: 40062890-40062890 on Build GRCh38
rs945579081 Chr.21: 40063030-40063030 on Build GRCh38
rs964390913 Chr.21: 40062979-40062979 on Build GRCh38
rs763982794 Chr.21: 40062898-40062898 on Build GRCh38
rs751279749 Chr.21: 40062903-40062903 on Build GRCh38
rs938019664 Chr.21: 40063114-40063114 on Build GRCh38
rs573903818 Chr.21: 40062884-40062884 on Build GRCh38
rs576747751 Chr.21: 40063000-40063000 on Build GRCh38
rs998256983 Chr.21: 40063145-40063145 on Build GRCh38
rs930691906 Chr.21: 40063179-40063179 on Build GRCh38
rs757138450 Chr.21: 40062902-40062902 on Build GRCh38
rs545599197 Chr.21: 40063055-40063055 on Build GRCh38
rs751069934 Chr.21: 40062922-40062922 on Build GRCh38
rs914045461 Chr.21: 40063012-40063012 on Build GRCh38
rs888327936 Chr.21: 40063046-40063046 on Build GRCh38
rs777292862 Chr.21: 40062833-40062833 on Build GRCh38
rs746833069 Chr.21: 40062953-40062953 on Build GRCh38
rs761002553 Chr.21: 40062989-40062989 on Build GRCh38
rs374548601 Chr.21: 40062784-40062784 on Build GRCh38
rs554143913 Chr.21: 40062843-40062843 on Build GRCh38

View Full Product Details